Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 134858940-134859194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf24
Entrez Gene ID
134553
Gene Name
chromosome 5 open reading frame 24
Gene Aliases
-
UniGene
Hs.708536
Gene Chromosome Location
Chr.5: 134833603-134859735 on Build GRCh38
Transcripts
NM_001135586.1 NM_001300894.1 NM_152409.3 XM_005271889.3 XM_017009049.1 XM_017009050.1

SNP Detail

SNP ID Chromosome Location
rs939577380 Chr.5: 134859082-134859082 on Build GRCh38
rs145093735 Chr.5: 134858975-134858975 on Build GRCh38
rs62380022 Chr.5: 134859158-134859158 on Build GRCh38
rs4418145 Chr.5: 134859079-134859079 on Build GRCh38
rs971774479 Chr.5: 134858985-134858985 on Build GRCh38
rs1028981965 Chr.5: 134858978-134858978 on Build GRCh38
rs140681325 Chr.5: 134859160-134859160 on Build GRCh38
rs1056504869 Chr.5: 134859100-134859100 on Build GRCh38
rs920802314 Chr.5: 134859017-134859018 on Build GRCh38
rs892460157 Chr.5: 134859129-134859129 on Build GRCh38
rs962323204 Chr.5: 134859151-134859151 on Build GRCh38
rs930831983 Chr.5: 134859024-134859024 on Build GRCh38
rs551734550 Chr.5: 134859106-134859106 on Build GRCh38
rs1045608023 Chr.5: 134859031-134859031 on Build GRCh38
rs529329525 Chr.5: 134859039-134859039 on Build GRCh38
rs371291842 Chr.5: 134859080-134859080 on Build GRCh38
rs563074442 Chr.5: 134859057-134859057 on Build GRCh38
rs1015633180 Chr.5: 134859084-134859084 on Build GRCh38
rs544888379 Chr.5: 134859048-134859048 on Build GRCh38
rs952173976 Chr.5: 134858994-134858994 on Build GRCh38
rs533548967 Chr.5: 134859068-134859068 on Build GRCh38

View Full Product Details