Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 147392108-147392687 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPYSL3
Entrez Gene ID
1809
Gene Name
dihydropyrimidinase like 3
Gene Aliases
CRMP-4, CRMP4, DRP-3, DRP3, LCRMP, ULIP, ULIP-1
UniGene
Hs.519659
Gene Chromosome Location
Chr.5: 147390808-147510056 on Build GRCh38
Transcripts
NM_001197294.1 NM_001387.2 XM_011537574.1 XM_011537577.2

Gene Symbol
STK32A
Entrez Gene ID
202374
Gene Name
serine/threonine kinase 32A
Gene Aliases
YANK1
UniGene
Hs.585069
Gene Chromosome Location
Chr.5: 147234963-147401996 on Build GRCh38
Transcripts
NM_001197294.1 NM_001387.2 XM_011537574.1 XM_011537577.2

SNP Detail

SNP ID Chromosome Location
rs530676830 Chr.5: 147392521-147392521 on Build GRCh38
rs59026412 Chr.5: 147392310-147392310 on Build GRCh38
rs867237370 Chr.5: 147392544-147392544 on Build GRCh38
rs3805533 Chr.5: 147392187-147392187 on Build GRCh38
rs577580337 Chr.5: 147392537-147392537 on Build GRCh38
rs1000391210 Chr.5: 147392520-147392520 on Build GRCh38
rs890032573 Chr.5: 147392323-147392323 on Build GRCh38
rs535017942 Chr.5: 147392289-147392289 on Build GRCh38
rs369757405 Chr.5: 147392557-147392557 on Build GRCh38
rs984127456 Chr.5: 147392605-147392605 on Build GRCh38
rs765804403 Chr.5: 147392442-147392442 on Build GRCh38
rs1048715299 Chr.5: 147392320-147392320 on Build GRCh38
rs1021107170 Chr.5: 147392437-147392437 on Build GRCh38
rs944471309 Chr.5: 147392423-147392423 on Build GRCh38
rs1041217453 Chr.5: 147392372-147392372 on Build GRCh38
rs997145717 Chr.5: 147392167-147392167 on Build GRCh38
rs562019808 Chr.5: 147392508-147392508 on Build GRCh38
rs143682341 Chr.5: 147392413-147392413 on Build GRCh38
rs973198525 Chr.5: 147392281-147392281 on Build GRCh38
rs544437600 Chr.5: 147392536-147392536 on Build GRCh38
rs917297411 Chr.5: 147392276-147392276 on Build GRCh38
rs537653143 Chr.5: 147392367-147392367 on Build GRCh38
rs970468110 Chr.5: 147392319-147392319 on Build GRCh38
rs756774466 Chr.5: 147392579-147392579 on Build GRCh38
rs568629561 Chr.5: 147392313-147392313 on Build GRCh38
rs1043369498 Chr.5: 147392499-147392499 on Build GRCh38
rs1000332437 Chr.5: 147392189-147392189 on Build GRCh38
rs959233139 Chr.5: 147392262-147392262 on Build GRCh38
rs546235095 Chr.5: 147392218-147392218 on Build GRCh38
rs990678010 Chr.5: 147392264-147392264 on Build GRCh38
rs564124459 Chr.5: 147392608-147392608 on Build GRCh38
rs931334957 Chr.5: 147392586-147392586 on Build GRCh38
rs866405044 Chr.5: 147392419-147392419 on Build GRCh38
rs911184698 Chr.5: 147392612-147392612 on Build GRCh38
rs754207295 Chr.5: 147392328-147392328 on Build GRCh38
rs902228015 Chr.5: 147392462-147392462 on Build GRCh38
rs1032219554 Chr.5: 147392243-147392243 on Build GRCh38
rs917703300 Chr.5: 147392274-147392274 on Build GRCh38
rs775342340 Chr.5: 147392278-147392278 on Build GRCh38
rs918619694 Chr.5: 147392287-147392287 on Build GRCh38
rs920522605 Chr.5: 147392362-147392362 on Build GRCh38
rs760878995 Chr.5: 147392152-147392152 on Build GRCh38
rs753606011 Chr.5: 147392581-147392583 on Build GRCh38
rs1047617031 Chr.5: 147392397-147392397 on Build GRCh38
rs557229342 Chr.5: 147392396-147392396 on Build GRCh38
rs956153718 Chr.5: 147392229-147392229 on Build GRCh38
rs1031781681 Chr.5: 147392214-147392214 on Build GRCh38
rs1024819888 Chr.5: 147392572-147392572 on Build GRCh38
rs949514923 Chr.5: 147392279-147392279 on Build GRCh38
rs980094080 Chr.5: 147392353-147392353 on Build GRCh38
rs992865958 Chr.5: 147392255-147392255 on Build GRCh38
rs542218190 Chr.5: 147392496-147392496 on Build GRCh38
rs959286857 Chr.5: 147392515-147392515 on Build GRCh38
rs1014338047 Chr.5: 147392550-147392550 on Build GRCh38
rs573291818 Chr.5: 147392427-147392427 on Build GRCh38
rs1049171 Chr.5: 147392415-147392415 on Build GRCh38
rs1028995254 Chr.5: 147392175-147392175 on Build GRCh38
rs147224404 Chr.5: 147392418-147392418 on Build GRCh38
rs566108899 Chr.5: 147392283-147392283 on Build GRCh38
rs554858336 Chr.5: 147392312-147392312 on Build GRCh38

View Full Product Details