Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 16246644-16247131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMPR
Entrez Gene ID
2766
Gene Name
guanosine monophosphate reductase
Gene Aliases
GMPR 1, GMPR1
UniGene
Hs.484741
Gene Chromosome Location
Chr.6: 16238580-16295549 on Build GRCh38
Transcripts
NM_006877.3 XM_011514508.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1075798 c.101G>A p.R34H Chr.6: 16246855-16246855 on Build GRCh38
1442104 c.110C>T p.T37M Chr.6: 16246864-16246864 on Build GRCh38
5029484 c.207+6G>A p.? Chr.6: 16246967-16246967 on Build GRCh38
4386813 c.134A>G p.Y45C Chr.6: 16246888-16246888 on Build GRCh38
351352 c.150C>T p.I50I Chr.6: 16246904-16246904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773708499 Chr.6: 16247008-16247008 on Build GRCh38
rs1018635459 Chr.6: 16246828-16246828 on Build GRCh38
rs745734586 Chr.6: 16246972-16246972 on Build GRCh38
rs370278724 Chr.6: 16246797-16246797 on Build GRCh38
rs768110482 Chr.6: 16246818-16246818 on Build GRCh38
rs930486183 Chr.6: 16247090-16247090 on Build GRCh38
rs1022965492 Chr.6: 16246925-16246925 on Build GRCh38
rs372790288 Chr.6: 16246870-16246870 on Build GRCh38
rs113260772 Chr.6: 16247107-16247107 on Build GRCh38
rs934114734 Chr.6: 16247030-16247030 on Build GRCh38
rs200089377 Chr.6: 16246909-16246909 on Build GRCh38
rs764448787 Chr.6: 16246810-16246810 on Build GRCh38
rs752136074 Chr.6: 16246934-16246934 on Build GRCh38
rs909909360 Chr.6: 16246970-16246970 on Build GRCh38
rs768164091 Chr.6: 16246902-16246902 on Build GRCh38
rs576991943 Chr.6: 16247053-16247053 on Build GRCh38
rs774714347 Chr.6: 16246708-16246708 on Build GRCh38
rs772259846 Chr.6: 16246753-16246753 on Build GRCh38
rs939338261 Chr.6: 16246670-16246670 on Build GRCh38
rs772248295 Chr.6: 16246922-16246930 on Build GRCh38
rs368446203 Chr.6: 16246813-16246813 on Build GRCh38
rs748432274 Chr.6: 16246823-16246824 on Build GRCh38
rs763236446 Chr.6: 16246805-16246805 on Build GRCh38
rs758121173 Chr.6: 16246941-16246941 on Build GRCh38
rs749587857 Chr.6: 16246865-16246865 on Build GRCh38
rs148924653 Chr.6: 16247042-16247042 on Build GRCh38
rs908571343 Chr.6: 16247011-16247011 on Build GRCh38
rs756997269 Chr.6: 16246961-16246961 on Build GRCh38
rs149627498 Chr.6: 16246950-16246950 on Build GRCh38
rs745970404 Chr.6: 16246859-16246859 on Build GRCh38
rs150485998 Chr.6: 16246854-16246854 on Build GRCh38
rs777683771 Chr.6: 16246852-16246852 on Build GRCh38
rs532720146 Chr.6: 16246869-16246869 on Build GRCh38
rs763620736 Chr.6: 16246945-16246945 on Build GRCh38
rs762028767 Chr.6: 16246729-16246729 on Build GRCh38
rs901266647 Chr.6: 16247026-16247026 on Build GRCh38
rs143425054 Chr.6: 16246980-16246981 on Build GRCh38
rs369777917 Chr.6: 16247002-16247002 on Build GRCh38
rs200506239 Chr.6: 16246907-16246907 on Build GRCh38
rs762437950 Chr.6: 16246814-16246814 on Build GRCh38
rs764908312 Chr.6: 16246917-16246917 on Build GRCh38
rs11538835 Chr.6: 16246891-16246891 on Build GRCh38
rs145730255 Chr.6: 16247028-16247028 on Build GRCh38
rs1037497163 Chr.6: 16247025-16247025 on Build GRCh38
rs749303401 Chr.6: 16246993-16246993 on Build GRCh38
rs138267551 Chr.6: 16246864-16246864 on Build GRCh38
rs773614032 Chr.6: 16247014-16247014 on Build GRCh38
rs773800767 Chr.6: 16246906-16246906 on Build GRCh38
rs1036321831 Chr.6: 16247037-16247037 on Build GRCh38
rs180770339 Chr.6: 16247101-16247101 on Build GRCh38
rs988443571 Chr.6: 16246856-16246856 on Build GRCh38
rs888831802 Chr.6: 16246757-16246757 on Build GRCh38
rs201868333 Chr.6: 16246968-16246968 on Build GRCh38
rs778946935 Chr.6: 16246824-16246824 on Build GRCh38
rs773880662 Chr.6: 16246770-16246770 on Build GRCh38
rs752328522 Chr.6: 16246831-16246831 on Build GRCh38
rs374885877 Chr.6: 16247075-16247075 on Build GRCh38
rs376976957 Chr.6: 16246995-16246995 on Build GRCh38
rs1036676481 Chr.6: 16246683-16246683 on Build GRCh38
rs768957220 Chr.6: 16246996-16246996 on Build GRCh38
rs940644227 Chr.6: 16246682-16246682 on Build GRCh38
rs1028702920 Chr.6: 16246754-16246754 on Build GRCh38
rs753432808 Chr.6: 16246819-16246819 on Build GRCh38
rs764998367 Chr.6: 16246825-16246825 on Build GRCh38
rs778957545 Chr.6: 16246998-16246998 on Build GRCh38
rs376565645 Chr.6: 16246984-16246984 on Build GRCh38
rs754493907 Chr.6: 16246823-16246823 on Build GRCh38
rs1032001970 Chr.6: 16246671-16246671 on Build GRCh38
rs757983536 Chr.6: 16246846-16246846 on Build GRCh38
rs2274152 Chr.6: 16246674-16246674 on Build GRCh38
rs781065689 Chr.6: 16246858-16246858 on Build GRCh38
rs1031949848 Chr.6: 16247108-16247108 on Build GRCh38
rs757224478 Chr.6: 16246855-16246855 on Build GRCh38

View Full Product Details