Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 47576424-47576995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD2AP
Entrez Gene ID
23607
Gene Name
CD2 associated protein
Gene Aliases
CMS
UniGene
Hs.485518
Gene Chromosome Location
Chr.6: 47477746-47627263 on Build GRCh38
Transcripts
NM_012120.2 XM_005248976.1 XM_011514449.2 XM_017010641.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3777723 c.772G>C p.E258Q Chr.6: 47576566-47576566 on Build GRCh38
1079998 c.798T>C p.G266G Chr.6: 47576592-47576592 on Build GRCh38
5619852 c.779C>A p.T260K Chr.6: 47576573-47576573 on Build GRCh38
3353474 c.792C>T p.T264T Chr.6: 47576586-47576586 on Build GRCh38
7360240 c.772G>T p.E258* Chr.6: 47576566-47576566 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750612328 Chr.6: 47576639-47576639 on Build GRCh38
rs574442886 Chr.6: 47576748-47576748 on Build GRCh38
rs1034978918 Chr.6: 47576551-47576551 on Build GRCh38
rs767431919 Chr.6: 47576502-47576502 on Build GRCh38
rs375203011 Chr.6: 47576629-47576629 on Build GRCh38
rs754036606 Chr.6: 47576523-47576523 on Build GRCh38
rs1003417103 Chr.6: 47576817-47576817 on Build GRCh38
rs763886646 Chr.6: 47576457-47576457 on Build GRCh38
rs779242012 Chr.6: 47576535-47576535 on Build GRCh38
rs184327199 Chr.6: 47576651-47576651 on Build GRCh38
rs1005581100 Chr.6: 47576956-47576956 on Build GRCh38
rs1014937533 Chr.6: 47576778-47576778 on Build GRCh38
rs755612631 Chr.6: 47576649-47576649 on Build GRCh38
rs770767773 Chr.6: 47576591-47576591 on Build GRCh38
rs1025577490 Chr.6: 47576944-47576944 on Build GRCh38
rs368514132 Chr.6: 47576517-47576517 on Build GRCh38
rs769883915 Chr.6: 47576602-47576606 on Build GRCh38
rs373185402 Chr.6: 47576696-47576696 on Build GRCh38
rs996231191 Chr.6: 47576707-47576707 on Build GRCh38
rs897708638 Chr.6: 47576906-47576906 on Build GRCh38
rs760653572 Chr.6: 47576608-47576608 on Build GRCh38
rs1044896024 Chr.6: 47576462-47576462 on Build GRCh38
rs535431429 Chr.6: 47576874-47576874 on Build GRCh38
rs572279680 Chr.6: 47576943-47576943 on Build GRCh38
rs556298222 Chr.6: 47576725-47576725 on Build GRCh38
rs144331531 Chr.6: 47576467-47576467 on Build GRCh38
rs979828332 Chr.6: 47576507-47576507 on Build GRCh38
rs775449594 Chr.6: 47576606-47576606 on Build GRCh38
rs780574436 Chr.6: 47576548-47576548 on Build GRCh38
rs780336709 Chr.6: 47576645-47576645 on Build GRCh38
rs766248270 Chr.6: 47576487-47576487 on Build GRCh38
rs925687129 Chr.6: 47576931-47576931 on Build GRCh38
rs1044864690 Chr.6: 47576774-47576774 on Build GRCh38
rs144912461 Chr.6: 47576586-47576586 on Build GRCh38
rs141050155 Chr.6: 47576542-47576542 on Build GRCh38
rs1052071098 Chr.6: 47576739-47576739 on Build GRCh38
rs776297606 Chr.6: 47576557-47576558 on Build GRCh38
rs770411520 Chr.6: 47576507-47576508 on Build GRCh38
rs773889150 Chr.6: 47576593-47576593 on Build GRCh38
rs1025608540 Chr.6: 47576600-47576600 on Build GRCh38
rs747522110 Chr.6: 47576646-47576646 on Build GRCh38
rs750158112 Chr.6: 47576938-47576938 on Build GRCh38
rs977403733 Chr.6: 47576712-47576712 on Build GRCh38
rs113028773 Chr.6: 47576686-47576686 on Build GRCh38
rs759309620 Chr.6: 47576596-47576596 on Build GRCh38
rs969520046 Chr.6: 47576833-47576833 on Build GRCh38
rs552725917 Chr.6: 47576489-47576489 on Build GRCh38
rs112709151 Chr.6: 47576780-47576780 on Build GRCh38
rs979610103 Chr.6: 47576866-47576866 on Build GRCh38
rs1056704487 Chr.6: 47576901-47576901 on Build GRCh38
rs935665835 Chr.6: 47576569-47576569 on Build GRCh38
rs752581251 Chr.6: 47576508-47576508 on Build GRCh38
rs139417092 Chr.6: 47576823-47576826 on Build GRCh38
rs767246659 Chr.6: 47576602-47576602 on Build GRCh38
rs553531189 Chr.6: 47576897-47576897 on Build GRCh38
rs747496653 Chr.6: 47576566-47576566 on Build GRCh38
rs757454906 Chr.6: 47576631-47576631 on Build GRCh38
rs769409950 Chr.6: 47576582-47576582 on Build GRCh38
rs757507978 Chr.6: 47576524-47576524 on Build GRCh38
rs762745837 Chr.6: 47576477-47576477 on Build GRCh38
rs952741281 Chr.6: 47576952-47576952 on Build GRCh38
rs905017385 Chr.6: 47576816-47576816 on Build GRCh38
rs759501591 Chr.6: 47576571-47576576 on Build GRCh38
rs387906340 Chr.6: 47576523-47576523 on Build GRCh38
rs961220165 Chr.6: 47576809-47576809 on Build GRCh38
rs370791689 Chr.6: 47576501-47576501 on Build GRCh38
rs199622885 Chr.6: 47576587-47576587 on Build GRCh38
rs188419618 Chr.6: 47576697-47576697 on Build GRCh38
rs758743814 Chr.6: 47576640-47576640 on Build GRCh38
rs764126956 Chr.6: 47576610-47576610 on Build GRCh38
rs191382971 Chr.6: 47576455-47576455 on Build GRCh38
rs946430291 Chr.6: 47576770-47576770 on Build GRCh38
rs935781862 Chr.6: 47576949-47576952 on Build GRCh38
rs746297717 Chr.6: 47576538-47576538 on Build GRCh38
rs370335632 Chr.6: 47576520-47576520 on Build GRCh38
rs141402367 Chr.6: 47576458-47576458 on Build GRCh38
rs935588296 Chr.6: 47576726-47576726 on Build GRCh38

View Full Product Details