Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 98917276-98917808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL4
Entrez Gene ID
26235
Gene Name
F-box and leucine rich repeat protein 4
Gene Aliases
FBL4, FBL5, MTDPS13
UniGene
Hs.536850
Gene Chromosome Location
Chr.6: 98868535-98948006 on Build GRCh38
Transcripts
NM_001278716.1 NM_012160.4 NR_103836.1 NR_103837.1 XM_005266930.2 XM_011535748.2 XM_017010726.1 XM_017010727.1 XM_017010728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3698012 c.711G>T p.K237N Chr.6: 98917521-98917521 on Build GRCh38
6237239 c.706C>T p.L236F Chr.6: 98917526-98917526 on Build GRCh38
1728321 c.668T>C p.V223A Chr.6: 98917564-98917564 on Build GRCh38
3630716 c.523C>T p.L175F Chr.6: 98917709-98917709 on Build GRCh38
6514240 c.682G>T p.V228L Chr.6: 98917550-98917550 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763612215 Chr.6: 98917450-98917450 on Build GRCh38
rs895268542 Chr.6: 98917649-98917649 on Build GRCh38
rs1004540303 Chr.6: 98917313-98917313 on Build GRCh38
rs1045002743 Chr.6: 98917716-98917716 on Build GRCh38
rs200734083 Chr.6: 98917564-98917564 on Build GRCh38
rs1015958378 Chr.6: 98917693-98917693 on Build GRCh38
rs772730274 Chr.6: 98917763-98917763 on Build GRCh38
rs141193306 Chr.6: 98917501-98917501 on Build GRCh38
rs150748757 Chr.6: 98917691-98917691 on Build GRCh38
rs779833991 Chr.6: 98917625-98917625 on Build GRCh38
rs761491160 Chr.6: 98917542-98917542 on Build GRCh38
rs749286898 Chr.6: 98917595-98917595 on Build GRCh38
rs781164933 Chr.6: 98917735-98917735 on Build GRCh38
rs768763924 Chr.6: 98917349-98917351 on Build GRCh38
rs149549442 Chr.6: 98917488-98917488 on Build GRCh38
rs761384727 Chr.6: 98917459-98917459 on Build GRCh38
rs202033721 Chr.6: 98917550-98917550 on Build GRCh38
rs770348037 Chr.6: 98917768-98917768 on Build GRCh38
rs201149065 Chr.6: 98917706-98917706 on Build GRCh38
rs760396559 Chr.6: 98917435-98917435 on Build GRCh38
rs748899561 Chr.6: 98917766-98917766 on Build GRCh38
rs776805590 Chr.6: 98917352-98917352 on Build GRCh38
rs530909841 Chr.6: 98917773-98917773 on Build GRCh38
rs747923744 Chr.6: 98917749-98917749 on Build GRCh38
rs763549609 Chr.6: 98917556-98917556 on Build GRCh38
rs780661786 Chr.6: 98917326-98917326 on Build GRCh38
rs767346377 Chr.6: 98917418-98917418 on Build GRCh38
rs759388601 Chr.6: 98917414-98917414 on Build GRCh38
rs768714798 Chr.6: 98917349-98917349 on Build GRCh38
rs750971078 Chr.6: 98917477-98917477 on Build GRCh38
rs557769001 Chr.6: 98917483-98917483 on Build GRCh38
rs768342478 Chr.6: 98917642-98917642 on Build GRCh38
rs929955972 Chr.6: 98917452-98917452 on Build GRCh38
rs901074781 Chr.6: 98917701-98917701 on Build GRCh38
rs747270865 Chr.6: 98917343-98917343 on Build GRCh38
rs769532824 Chr.6: 98917666-98917666 on Build GRCh38
rs1055044734 Chr.6: 98917619-98917619 on Build GRCh38
rs1022013838 Chr.6: 98917785-98917785 on Build GRCh38
rs774720010 Chr.6: 98917685-98917685 on Build GRCh38
rs774255399 Chr.6: 98917410-98917410 on Build GRCh38
rs890876800 Chr.6: 98917565-98917565 on Build GRCh38
rs767879225 Chr.6: 98917690-98917690 on Build GRCh38
rs772085661 Chr.6: 98917598-98917598 on Build GRCh38
rs886041625 Chr.6: 98917610-98917611 on Build GRCh38
rs781455701 Chr.6: 98917566-98917566 on Build GRCh38
rs376909719 Chr.6: 98917456-98917456 on Build GRCh38
rs1043639561 Chr.6: 98917522-98917522 on Build GRCh38
rs953775721 Chr.6: 98917470-98917470 on Build GRCh38
rs776419278 Chr.6: 98917518-98917518 on Build GRCh38
rs199923936 Chr.6: 98917700-98917700 on Build GRCh38
rs201858974 Chr.6: 98917705-98917705 on Build GRCh38
rs143154211 Chr.6: 98917495-98917495 on Build GRCh38
rs989651385 Chr.6: 98917778-98917778 on Build GRCh38
rs771854304 Chr.6: 98917502-98917502 on Build GRCh38
rs764855170 Chr.6: 98917544-98917544 on Build GRCh38
rs780218423 Chr.6: 98917337-98917337 on Build GRCh38
rs768403473 Chr.6: 98917510-98917510 on Build GRCh38
rs753998502 Chr.6: 98917703-98917703 on Build GRCh38
rs200592647 Chr.6: 98917732-98917732 on Build GRCh38
rs748199266 Chr.6: 98917383-98917383 on Build GRCh38
rs939377900 Chr.6: 98917602-98917602 on Build GRCh38
rs903771977 Chr.6: 98917528-98917528 on Build GRCh38
rs545027066 Chr.6: 98917560-98917560 on Build GRCh38
rs955115739 Chr.6: 98917307-98917307 on Build GRCh38
rs995708161 Chr.6: 98917655-98917655 on Build GRCh38
rs965056782 Chr.6: 98917342-98917342 on Build GRCh38
rs779530195 Chr.6: 98917332-98917332 on Build GRCh38
rs373308410 Chr.6: 98917667-98917667 on Build GRCh38
rs765912815 Chr.6: 98917466-98917466 on Build GRCh38
rs766767309 Chr.6: 98917559-98917559 on Build GRCh38
rs760018394 Chr.6: 98917687-98917687 on Build GRCh38
rs964532159 Chr.6: 98917616-98917616 on Build GRCh38
rs761581391 Chr.6: 98917665-98917665 on Build GRCh38
rs758776562 Chr.6: 98917480-98917480 on Build GRCh38
rs752917494 Chr.6: 98917587-98917587 on Build GRCh38
rs138206466 Chr.6: 98917485-98917485 on Build GRCh38
rs771755480 Chr.6: 98917689-98917689 on Build GRCh38
rs369242693 Chr.6: 98917497-98917497 on Build GRCh38
rs1030715483 Chr.6: 98917303-98917303 on Build GRCh38
rs766972168 Chr.6: 98917674-98917674 on Build GRCh38
rs760356648 Chr.6: 98917508-98917508 on Build GRCh38
rs78911908 Chr.6: 98917562-98917562 on Build GRCh38
rs760238524 Chr.6: 98917692-98917692 on Build GRCh38
rs776510763 Chr.6: 98917662-98917662 on Build GRCh38
rs1018430772 Chr.6: 98917615-98917615 on Build GRCh38
rs1008415820 Chr.6: 98917599-98917599 on Build GRCh38
rs769621421 Chr.6: 98917752-98917752 on Build GRCh38
rs991952518 Chr.6: 98917781-98917781 on Build GRCh38
rs146999462 Chr.6: 98917419-98917419 on Build GRCh38
rs770907606 Chr.6: 98917398-98917398 on Build GRCh38
rs775384414 Chr.6: 98917507-98917507 on Build GRCh38
rs746878388 Chr.6: 98917631-98917631 on Build GRCh38
rs191658390 Chr.6: 98917695-98917695 on Build GRCh38
rs148621221 Chr.6: 98917496-98917496 on Build GRCh38
rs371516190 Chr.6: 98917589-98917589 on Build GRCh38
rs746293212 Chr.6: 98917335-98917335 on Build GRCh38
rs796181204 Chr.6: 98917539-98917539 on Build GRCh38

View Full Product Details