Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 53272806-53273368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL5
Entrez Gene ID
60481
Gene Name
ELOVL fatty acid elongase 5
Gene Aliases
HELO1, SCA38, dJ483K16.1
UniGene
Hs.520189
Gene Chromosome Location
Chr.6: 53267398-53349179 on Build GRCh38
Transcripts
NM_001242828.1 NM_001242830.1 NM_001301856.1 NM_021814.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5676263 c.613G>T p.V205F Chr.6: 53273309-53273309 on Build GRCh38
3777753 c.537C>T p.L179L Chr.6: 53273304-53273304 on Build GRCh38
4571962 c.519T>A p.N173K Chr.6: 53273322-53273322 on Build GRCh38
4571961 c.600T>A p.N200K Chr.6: 53273322-53273322 on Build GRCh38
4503891 c.569C>T p.P190L Chr.6: 53273272-53273272 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894971196 Chr.6: 53273115-53273115 on Build GRCh38
rs959237977 Chr.6: 53272851-53272851 on Build GRCh38
rs553585093 Chr.6: 53273328-53273328 on Build GRCh38
rs66540961 Chr.6: 53272912-53272921 on Build GRCh38
rs532876575 Chr.6: 53272973-53272973 on Build GRCh38
rs150496797 Chr.6: 53273103-53273103 on Build GRCh38
rs966875761 Chr.6: 53272931-53272932 on Build GRCh38
rs746398884 Chr.6: 53273199-53273199 on Build GRCh38
rs183968702 Chr.6: 53273249-53273249 on Build GRCh38
rs765847164 Chr.6: 53273301-53273301 on Build GRCh38
rs982863469 Chr.6: 53273303-53273303 on Build GRCh38
rs1047053082 Chr.6: 53273047-53273047 on Build GRCh38
rs541453246 Chr.6: 53273023-53273023 on Build GRCh38
rs749381595 Chr.6: 53273189-53273189 on Build GRCh38
rs191773768 Chr.6: 53272832-53272832 on Build GRCh38
rs80193816 Chr.6: 53272921-53272921 on Build GRCh38
rs911388871 Chr.6: 53272941-53272941 on Build GRCh38
rs761391551 Chr.6: 53273264-53273264 on Build GRCh38
rs1011979846 Chr.6: 53272858-53272858 on Build GRCh38
rs915845225 Chr.6: 53272900-53272900 on Build GRCh38
rs772383368 Chr.6: 53273200-53273200 on Build GRCh38
rs561340155 Chr.6: 53273168-53273168 on Build GRCh38
rs751476241 Chr.6: 53273306-53273306 on Build GRCh38
rs1017198515 Chr.6: 53273004-53273004 on Build GRCh38
rs759102900 Chr.6: 53273083-53273083 on Build GRCh38
rs1012507710 Chr.6: 53273131-53273131 on Build GRCh38
rs983314742 Chr.6: 53273090-53273090 on Build GRCh38
rs537497996 Chr.6: 53273173-53273173 on Build GRCh38
rs987030084 Chr.6: 53273178-53273178 on Build GRCh38
rs752544208 Chr.6: 53273320-53273320 on Build GRCh38
rs1052872168 Chr.6: 53272984-53272984 on Build GRCh38
rs904345322 Chr.6: 53272939-53272939 on Build GRCh38
rs754985930 Chr.6: 53273309-53273309 on Build GRCh38
rs930059445 Chr.6: 53273276-53273276 on Build GRCh38
rs763090303 Chr.6: 53272911-53272912 on Build GRCh38
rs756040737 Chr.6: 53273327-53273327 on Build GRCh38
rs772801768 Chr.6: 53273274-53273274 on Build GRCh38
rs1038440246 Chr.6: 53272979-53272979 on Build GRCh38
rs535445676 Chr.6: 53273063-53273064 on Build GRCh38
rs116481773 Chr.6: 53272892-53272892 on Build GRCh38
rs1038702815 Chr.6: 53273296-53273296 on Build GRCh38
rs113464632 Chr.6: 53272918-53272921 on Build GRCh38
rs968543678 Chr.6: 53272909-53272909 on Build GRCh38
rs919982140 Chr.6: 53273207-53273207 on Build GRCh38
rs1008530049 Chr.6: 53272995-53272995 on Build GRCh38
rs929968383 Chr.6: 53273046-53273046 on Build GRCh38
rs774228952 Chr.6: 53273179-53273180 on Build GRCh38
rs1056332844 Chr.6: 53273113-53273113 on Build GRCh38
rs915895962 Chr.6: 53273116-53273116 on Build GRCh38
rs774895342 Chr.6: 53273196-53273196 on Build GRCh38
rs948674001 Chr.6: 53273122-53273122 on Build GRCh38
rs367553728 Chr.6: 53273263-53273263 on Build GRCh38
rs890166328 Chr.6: 53272985-53272985 on Build GRCh38
rs761777839 Chr.6: 53273258-53273265 on Build GRCh38
rs955619058 Chr.6: 53272893-53272893 on Build GRCh38
rs938665278 Chr.6: 53272842-53272842 on Build GRCh38
rs987082382 Chr.6: 53272915-53272915 on Build GRCh38
rs375281159 Chr.6: 53273184-53273184 on Build GRCh38
rs187353151 Chr.6: 53273310-53273310 on Build GRCh38
rs1022493242 Chr.6: 53272859-53272859 on Build GRCh38
rs900114699 Chr.6: 53273022-53273022 on Build GRCh38
rs199770402 Chr.6: 53273280-53273280 on Build GRCh38
rs770971103 Chr.6: 53273191-53273191 on Build GRCh38
rs767015504 Chr.6: 53273143-53273143 on Build GRCh38
rs967711629 Chr.6: 53272870-53272870 on Build GRCh38
rs764868909 Chr.6: 53273266-53273266 on Build GRCh38
rs978181926 Chr.6: 53272890-53272890 on Build GRCh38
rs560747726 Chr.6: 53272925-53272925 on Build GRCh38
rs756416091 Chr.6: 53273187-53273187 on Build GRCh38
rs903604211 Chr.6: 53273159-53273159 on Build GRCh38
rs992847194 Chr.6: 53273104-53273104 on Build GRCh38
rs58581846 Chr.6: 53272921-53272930 on Build GRCh38
rs539911231 Chr.6: 53273087-53273087 on Build GRCh38
rs964214485 Chr.6: 53273186-53273186 on Build GRCh38
rs77341998 Chr.6: 53273161-53273161 on Build GRCh38
rs1030666653 Chr.6: 53273168-53273168 on Build GRCh38
rs920017013 Chr.6: 53273041-53273041 on Build GRCh38
rs1034823347 Chr.6: 53272835-53272835 on Build GRCh38
rs751092138 Chr.6: 53273048-53273048 on Build GRCh38
rs1015694093 Chr.6: 53273098-53273098 on Build GRCh38

View Full Product Details