Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 135387870-135388320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHI1
Entrez Gene ID
54806
Gene Name
Abelson helper integration site 1
Gene Aliases
AHI-1, JBTS3, ORF1, dJ71N10.1
UniGene
Hs.386684
Gene Chromosome Location
Chr.6: 135283972-135497775 on Build GRCh38
Transcripts
NM_001134830.1 NM_001134831.1 NM_001134832.1 NM_017651.4 XM_011535910.2 XM_011535911.2 XM_017010977.1 XM_017010978.1 XM_017010979.1 XM_017010980.1 XM_017010981.1 XM_017010982.1 XM_017010983.1 XM_017010984.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5975792 c.3152A>G p.K1051R Chr.6: 135387991-135387991 on Build GRCh38
1729497 c.3133A>T p.T1045S Chr.6: 135388010-135388010 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746468056 Chr.6: 135388053-135388053 on Build GRCh38
rs1055879335 Chr.6: 135388094-135388094 on Build GRCh38
rs774535934 Chr.6: 135388067-135388067 on Build GRCh38
rs17064487 Chr.6: 135387948-135387948 on Build GRCh38
rs759963348 Chr.6: 135388073-135388073 on Build GRCh38
rs376533791 Chr.6: 135387984-135387984 on Build GRCh38
rs368653066 Chr.6: 135388137-135388148 on Build GRCh38
rs768163296 Chr.6: 135388054-135388054 on Build GRCh38
rs1020543685 Chr.6: 135387907-135387907 on Build GRCh38
rs972189867 Chr.6: 135388193-135388193 on Build GRCh38
rs369930986 Chr.6: 135388285-135388285 on Build GRCh38
rs997206556 Chr.6: 135388095-135388095 on Build GRCh38
rs771807833 Chr.6: 135387936-135387936 on Build GRCh38
rs984854398 Chr.6: 135388168-135388168 on Build GRCh38
rs553193078 Chr.6: 135388042-135388042 on Build GRCh38
rs775146185 Chr.6: 135388051-135388051 on Build GRCh38
rs148672318 Chr.6: 135388135-135388146 on Build GRCh38
rs926121131 Chr.6: 135388000-135388005 on Build GRCh38
rs903205013 Chr.6: 135388173-135388173 on Build GRCh38
rs752771214 Chr.6: 135387998-135387998 on Build GRCh38
rs771505431 Chr.6: 135388066-135388066 on Build GRCh38
rs761913026 Chr.6: 135388057-135388057 on Build GRCh38
rs1028986361 Chr.6: 135388109-135388109 on Build GRCh38
rs1038262941 Chr.6: 135388065-135388065 on Build GRCh38
rs765688177 Chr.6: 135387957-135387957 on Build GRCh38
rs375377363 Chr.6: 135388182-135388182 on Build GRCh38
rs762589948 Chr.6: 135387997-135388005 on Build GRCh38
rs895924922 Chr.6: 135388122-135388122 on Build GRCh38
rs749176640 Chr.6: 135388022-135388022 on Build GRCh38
rs2064429 Chr.6: 135388083-135388083 on Build GRCh38
rs200151742 Chr.6: 135388030-135388030 on Build GRCh38
rs764391581 Chr.6: 135388017-135388018 on Build GRCh38
rs866101022 Chr.6: 135388032-135388032 on Build GRCh38
rs964574900 Chr.6: 135388006-135388006 on Build GRCh38
rs778552866 Chr.6: 135388031-135388031 on Build GRCh38
rs752001838 Chr.6: 135387985-135387985 on Build GRCh38
rs767789516 Chr.6: 135388076-135388076 on Build GRCh38
rs745496007 Chr.6: 135387932-135387932 on Build GRCh38
rs752971408 Chr.6: 135388078-135388078 on Build GRCh38
rs773726424 Chr.6: 135387967-135387967 on Build GRCh38
rs113398452 Chr.6: 135388070-135388071 on Build GRCh38
rs543736818 Chr.6: 135388201-135388201 on Build GRCh38
rs908220451 Chr.6: 135388052-135388052 on Build GRCh38
rs776273631 Chr.6: 135388059-135388059 on Build GRCh38
rs201629742 Chr.6: 135387961-135387961 on Build GRCh38
rs909271495 Chr.6: 135388177-135388177 on Build GRCh38
rs533293205 Chr.6: 135388195-135388195 on Build GRCh38
rs760210121 Chr.6: 135387955-135387955 on Build GRCh38
rs867774458 Chr.6: 135387949-135387949 on Build GRCh38
rs767879728 Chr.6: 135387997-135387997 on Build GRCh38
rs545085352 Chr.6: 135388163-135388163 on Build GRCh38
rs938946304 Chr.6: 135388281-135388281 on Build GRCh38
rs938938708 Chr.6: 135387927-135387927 on Build GRCh38
rs975177867 Chr.6: 135388007-135388007 on Build GRCh38
rs373270480 Chr.6: 135388050-135388050 on Build GRCh38
rs201074308 Chr.6: 135388010-135388010 on Build GRCh38
rs763343744 Chr.6: 135388061-135388061 on Build GRCh38
rs756276732 Chr.6: 135388004-135388004 on Build GRCh38
rs976026901 Chr.6: 135388198-135388198 on Build GRCh38
rs1055423149 Chr.6: 135387941-135387941 on Build GRCh38
rs765333574 Chr.6: 135388127-135388127 on Build GRCh38
rs774387901 Chr.6: 135388030-135388033 on Build GRCh38
rs1014301583 Chr.6: 135387904-135387904 on Build GRCh38
rs1020259198 Chr.6: 135388148-135388148 on Build GRCh38

View Full Product Details