Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 129605622-129606102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP18
Entrez Gene ID
93663
Gene Name
Rho GTPase activating protein 18
Gene Aliases
MacGAP, SENEX, bA307O14.2
UniGene
Hs.486458
Gene Chromosome Location
Chr.6: 129576145-129710225 on Build GRCh38
Transcripts
NM_033515.2 XM_005267212.2 XM_005267213.1 XM_017011494.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3948004 c.1332C>G p.I444M Chr.6: 129605910-129605910 on Build GRCh38
6293984 c.1350C>A p.N450K Chr.6: 129605892-129605892 on Build GRCh38
4934212 c.1350C>T p.N450N Chr.6: 129605892-129605892 on Build GRCh38
7000306 c.1330A>G p.I444V Chr.6: 129605912-129605912 on Build GRCh38
7209529 c.1295A>T p.K432M Chr.6: 129605947-129605947 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs539451121 Chr.6: 129605824-129605824 on Build GRCh38
rs76670714 Chr.6: 129605764-129605764 on Build GRCh38
rs981586849 Chr.6: 129605939-129605939 on Build GRCh38
rs916798977 Chr.6: 129606043-129606043 on Build GRCh38
rs765794990 Chr.6: 129605856-129605856 on Build GRCh38
rs147754877 Chr.6: 129605940-129605940 on Build GRCh38
rs939648656 Chr.6: 129605697-129605697 on Build GRCh38
rs781400499 Chr.6: 129605908-129605908 on Build GRCh38
rs775313640 Chr.6: 129605981-129605981 on Build GRCh38
rs1043800337 Chr.6: 129605756-129605756 on Build GRCh38
rs145138074 Chr.6: 129605799-129605799 on Build GRCh38
rs972415026 Chr.6: 129605963-129605963 on Build GRCh38
rs376956100 Chr.6: 129606065-129606065 on Build GRCh38
rs184996641 Chr.6: 129605776-129605776 on Build GRCh38
rs747202090 Chr.6: 129605970-129605972 on Build GRCh38
rs750958692 Chr.6: 129605868-129605868 on Build GRCh38
rs147822812 Chr.6: 129605692-129605692 on Build GRCh38
rs3813366 Chr.6: 129605869-129605869 on Build GRCh38
rs972901324 Chr.6: 129605669-129605670 on Build GRCh38
rs918247104 Chr.6: 129605978-129605978 on Build GRCh38
rs771783590 Chr.6: 129605973-129605973 on Build GRCh38
rs916620505 Chr.6: 129605741-129605741 on Build GRCh38
rs181017947 Chr.6: 129605719-129605719 on Build GRCh38
rs3829756 Chr.6: 129605657-129605657 on Build GRCh38
rs901653392 Chr.6: 129605788-129605788 on Build GRCh38
rs755138072 Chr.6: 129605898-129605898 on Build GRCh38
rs777550443 Chr.6: 129605923-129605923 on Build GRCh38
rs979647046 Chr.6: 129605802-129605802 on Build GRCh38
rs781747363 Chr.6: 129605743-129605743 on Build GRCh38
rs778221850 Chr.6: 129606048-129606048 on Build GRCh38
rs552580597 Chr.6: 129605713-129605713 on Build GRCh38
rs535178512 Chr.6: 129605770-129605770 on Build GRCh38
rs917705561 Chr.6: 129605930-129605930 on Build GRCh38
rs763634388 Chr.6: 129606010-129606014 on Build GRCh38
rs202043590 Chr.6: 129605938-129605938 on Build GRCh38
rs991065709 Chr.6: 129606016-129606020 on Build GRCh38
rs139614205 Chr.6: 129605766-129605766 on Build GRCh38
rs370858739 Chr.6: 129605922-129605922 on Build GRCh38
rs1014698939 Chr.6: 129605929-129605929 on Build GRCh38
rs745383100 Chr.6: 129605962-129605962 on Build GRCh38
rs773558049 Chr.6: 129606004-129606004 on Build GRCh38
rs751720325 Chr.6: 129605888-129605888 on Build GRCh38
rs191563044 Chr.6: 129606083-129606083 on Build GRCh38
rs754197495 Chr.6: 129605832-129605832 on Build GRCh38
rs762214955 Chr.6: 129605988-129605988 on Build GRCh38
rs555606373 Chr.6: 129606024-129606024 on Build GRCh38
rs531785206 Chr.6: 129605727-129605727 on Build GRCh38
rs902569659 Chr.6: 129605761-129605761 on Build GRCh38
rs773967441 Chr.6: 129605943-129605943 on Build GRCh38
rs1020948090 Chr.6: 129605790-129605790 on Build GRCh38
rs777653808 Chr.6: 129605857-129605857 on Build GRCh38
rs766518423 Chr.6: 129605877-129605877 on Build GRCh38
rs770345629 Chr.6: 129605942-129605942 on Build GRCh38
rs764538304 Chr.6: 129605831-129605831 on Build GRCh38
rs878964422 Chr.6: 129605768-129605768 on Build GRCh38

View Full Product Details