Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44144106-44144703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCK
Entrez Gene ID
2645
Gene Name
glucokinase
Gene Aliases
FGQTL3, GK, GLK, HHF3, HK4, HKIV, HXKP, LGLK, MODY2
UniGene
Hs.1270
Gene Chromosome Location
Chr.7: 44143213-44189439 on Build GRCh38
Transcripts
NM_000162.3 NM_033507.1 NM_033508.1 XM_017011966.1

SNP Detail

SNP ID Chromosome Location
rs746383753 Chr.7: 44144143-44144143 on Build GRCh38
rs904019835 Chr.7: 44144501-44144501 on Build GRCh38
rs749438148 Chr.7: 44144335-44144335 on Build GRCh38
rs972002260 Chr.7: 44144646-44144646 on Build GRCh38
rs55714218 Chr.7: 44144377-44144377 on Build GRCh38
rs761652517 Chr.7: 44144376-44144377 on Build GRCh38
rs185418856 Chr.7: 44144372-44144372 on Build GRCh38
rs1027142666 Chr.7: 44144609-44144609 on Build GRCh38
rs968839682 Chr.7: 44144300-44144300 on Build GRCh38
rs781494372 Chr.7: 44144172-44144172 on Build GRCh38
rs1013826118 Chr.7: 44144177-44144177 on Build GRCh38
rs961851617 Chr.7: 44144206-44144206 on Build GRCh38
rs1056498201 Chr.7: 44144415-44144415 on Build GRCh38
rs920044787 Chr.7: 44144245-44144245 on Build GRCh38
rs912546717 Chr.7: 44144292-44144292 on Build GRCh38
rs2908276 Chr.7: 44144339-44144339 on Build GRCh38
rs939314970 Chr.7: 44144413-44144413 on Build GRCh38
rs570413232 Chr.7: 44144383-44144383 on Build GRCh38
rs556996030 Chr.7: 44144401-44144401 on Build GRCh38
rs780243846 Chr.7: 44144290-44144290 on Build GRCh38
rs987066442 Chr.7: 44144582-44144582 on Build GRCh38
rs568995709 Chr.7: 44144445-44144445 on Build GRCh38
rs954295362 Chr.7: 44144617-44144617 on Build GRCh38
rs905303097 Chr.7: 44144408-44144408 on Build GRCh38
rs892745593 Chr.7: 44144197-44144197 on Build GRCh38
rs756513678 Chr.7: 44144196-44144196 on Build GRCh38
rs141645300 Chr.7: 44144626-44144626 on Build GRCh38
rs530604939 Chr.7: 44144362-44144362 on Build GRCh38
rs555058443 Chr.7: 44144458-44144458 on Build GRCh38
rs987128405 Chr.7: 44144349-44144349 on Build GRCh38
rs1018570440 Chr.7: 44144600-44144600 on Build GRCh38
rs2908275 Chr.7: 44144659-44144659 on Build GRCh38
rs1035257023 Chr.7: 44144180-44144180 on Build GRCh38
rs953157470 Chr.7: 44144562-44144562 on Build GRCh38
rs892694860 Chr.7: 44144438-44144438 on Build GRCh38
rs200628800 Chr.7: 44144377-44144377 on Build GRCh38
rs35548117 Chr.7: 44144277-44144278 on Build GRCh38
rs974230278 Chr.7: 44144242-44144242 on Build GRCh38
rs886062344 Chr.7: 44144292-44144292 on Build GRCh38
rs536317133 Chr.7: 44144429-44144429 on Build GRCh38
rs1028731343 Chr.7: 44144554-44144554 on Build GRCh38
rs76374134 Chr.7: 44144289-44144289 on Build GRCh38
rs528412344 Chr.7: 44144397-44144397 on Build GRCh38
rs886062346 Chr.7: 44144415-44144415 on Build GRCh38
rs1015826606 Chr.7: 44144200-44144200 on Build GRCh38
rs34322372 Chr.7: 44144357-44144358 on Build GRCh38
rs960976935 Chr.7: 44144170-44144170 on Build GRCh38
rs944020614 Chr.7: 44144300-44144300 on Build GRCh38
rs138562953 Chr.7: 44144403-44144403 on Build GRCh38
rs886062345 Chr.7: 44144413-44144413 on Build GRCh38
rs961647908 Chr.7: 44144466-44144466 on Build GRCh38
rs752178730 Chr.7: 44144151-44144153 on Build GRCh38
rs997459290 Chr.7: 44144517-44144517 on Build GRCh38
rs746199815 Chr.7: 44144176-44144176 on Build GRCh38
rs570194904 Chr.7: 44144527-44144527 on Build GRCh38
rs886062347 Chr.7: 44144588-44144588 on Build GRCh38
rs1009716250 Chr.7: 44144259-44144259 on Build GRCh38
rs1022847499 Chr.7: 44144480-44144480 on Build GRCh38
rs978538605 Chr.7: 44144302-44144302 on Build GRCh38
rs911794202 Chr.7: 44144369-44144369 on Build GRCh38
rs1052976869 Chr.7: 44144321-44144321 on Build GRCh38

View Full Product Details