Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 36894651-36895188 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMO1
Entrez Gene ID
9844
Gene Name
engulfment and cell motility 1
Gene Aliases
CED-12, CED12, ELMO-1
UniGene
Hs.434989
Gene Chromosome Location
Chr.7: 36852906-37449409 on Build GRCh38
Transcripts
NM_001039459.2 NM_001206480.2 NM_001206482.1 NM_014800.10 NM_130442.3 NR_038120.1 XM_005249919.2 XM_006715805.1 XM_011515654.1 XM_017012838.1 XM_017012839.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4814729 c.658C>T p.R220C Chr.7: 36894903-36894903 on Build GRCh38
4469138 c.1584C>T p.F528F Chr.7: 36894871-36894871 on Build GRCh38
294349 c.1560C>T p.S520S Chr.7: 36894895-36894895 on Build GRCh38
4469139 c.690C>T p.F230F Chr.7: 36894871-36894871 on Build GRCh38
3229002 c.1443G>T p.M481I Chr.7: 36895012-36895012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11766181 Chr.7: 36894912-36894912 on Build GRCh38
rs770672251 Chr.7: 36894809-36894809 on Build GRCh38
rs1020708375 Chr.7: 36895022-36895022 on Build GRCh38
rs917443601 Chr.7: 36894772-36894772 on Build GRCh38
rs184229988 Chr.7: 36895121-36895121 on Build GRCh38
rs926392786 Chr.7: 36895091-36895091 on Build GRCh38
rs1048575731 Chr.7: 36894789-36894789 on Build GRCh38
rs11766187 Chr.7: 36894968-36894968 on Build GRCh38
rs756595816 Chr.7: 36894808-36894813 on Build GRCh38
rs769542184 Chr.7: 36894839-36894839 on Build GRCh38
rs956711929 Chr.7: 36894965-36894965 on Build GRCh38
rs559855594 Chr.7: 36895124-36895124 on Build GRCh38
rs1029574627 Chr.7: 36894860-36894860 on Build GRCh38
rs1044803006 Chr.7: 36894775-36894775 on Build GRCh38
rs949567001 Chr.7: 36894765-36894765 on Build GRCh38
rs959018235 Chr.7: 36895104-36895104 on Build GRCh38
rs937570634 Chr.7: 36894670-36894670 on Build GRCh38
rs749843341 Chr.7: 36894829-36894829 on Build GRCh38
rs751696667 Chr.7: 36895066-36895066 on Build GRCh38
rs759991890 Chr.7: 36895008-36895008 on Build GRCh38
rs779233099 Chr.7: 36894894-36894894 on Build GRCh38
rs777772565 Chr.7: 36895063-36895063 on Build GRCh38
rs780950923 Chr.7: 36894943-36894943 on Build GRCh38
rs570950972 Chr.7: 36894786-36894786 on Build GRCh38
rs574320132 Chr.7: 36895084-36895084 on Build GRCh38
rs369838763 Chr.7: 36894859-36894859 on Build GRCh38
rs767053616 Chr.7: 36894994-36894994 on Build GRCh38
rs953621268 Chr.7: 36894868-36894868 on Build GRCh38
rs761954839 Chr.7: 36894865-36894865 on Build GRCh38
rs759128899 Chr.7: 36894834-36894834 on Build GRCh38
rs754271181 Chr.7: 36894863-36894863 on Build GRCh38
rs780516391 Chr.7: 36894819-36894819 on Build GRCh38
rs1052637554 Chr.7: 36894729-36894729 on Build GRCh38
rs868556780 Chr.7: 36894903-36894903 on Build GRCh38
rs950246342 Chr.7: 36894783-36894783 on Build GRCh38
rs754220393 Chr.7: 36894996-36894996 on Build GRCh38
rs770868534 Chr.7: 36894937-36894937 on Build GRCh38
rs777742599 Chr.7: 36894919-36894919 on Build GRCh38
rs931017490 Chr.7: 36895093-36895093 on Build GRCh38
rs200161329 Chr.7: 36894829-36894829 on Build GRCh38
rs141005773 Chr.7: 36895148-36895148 on Build GRCh38
rs755241179 Chr.7: 36894884-36894884 on Build GRCh38
rs750912430 Chr.7: 36894858-36894858 on Build GRCh38
rs6462722 Chr.7: 36894843-36894843 on Build GRCh38
rs996674526 Chr.7: 36895156-36895156 on Build GRCh38
rs138056213 Chr.7: 36894762-36894762 on Build GRCh38
rs1009805134 Chr.7: 36894964-36894964 on Build GRCh38
rs868099906 Chr.7: 36894822-36894822 on Build GRCh38
rs769301419 Chr.7: 36894975-36894975 on Build GRCh38
rs192804513 Chr.7: 36894982-36894982 on Build GRCh38
rs887184687 Chr.7: 36894790-36894790 on Build GRCh38
rs575519644 Chr.7: 36894988-36894988 on Build GRCh38
rs965183279 Chr.7: 36894813-36894813 on Build GRCh38
rs1040964567 Chr.7: 36895144-36895144 on Build GRCh38
rs917486982 Chr.7: 36895146-36895146 on Build GRCh38
rs746880873 Chr.7: 36894921-36894921 on Build GRCh38
rs537716479 Chr.7: 36895048-36895048 on Build GRCh38
rs991724834 Chr.7: 36895122-36895122 on Build GRCh38
rs979084001 Chr.7: 36895083-36895083 on Build GRCh38
rs765537908 Chr.7: 36895013-36895013 on Build GRCh38
rs376740137 Chr.7: 36895060-36895060 on Build GRCh38
rs765555514 Chr.7: 36894895-36894895 on Build GRCh38
rs758450898 Chr.7: 36894901-36894901 on Build GRCh38
rs746967955 Chr.7: 36894804-36894804 on Build GRCh38
rs762566756 Chr.7: 36894844-36894844 on Build GRCh38
rs372182988 Chr.7: 36894977-36894977 on Build GRCh38
rs745728435 Chr.7: 36894973-36894973 on Build GRCh38

View Full Product Details