Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 130506110-130506667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPG2
Entrez Gene ID
26958
Gene Name
coatomer protein complex subunit gamma 2
Gene Aliases
2-COP, gamma-2-COP
UniGene
Hs.6421
Gene Chromosome Location
Chr.7: 130506238-130668758 on Build GRCh38
Transcripts
NM_012133.5 NM_001253900.1 NM_001253901.1 NM_001253902.1 NM_002402.3 NM_177524.2 NM_177525.2 XM_011516222.1 XM_017012218.1

Gene Symbol
MEST
Entrez Gene ID
4232
Gene Name
mesoderm specific transcript
Gene Aliases
PEG1
UniGene
Hs.270978
Gene Chromosome Location
Chr.7: 130486175-130506297 on Build GRCh38
Transcripts
NM_012133.5 NM_001253900.1 NM_001253901.1 NM_001253902.1 NM_002402.3 NM_177524.2 NM_177525.2 XM_011516222.1 XM_017012218.1

SNP Detail

SNP ID Chromosome Location
rs377671413 Chr.7: 130506407-130506407 on Build GRCh38
rs564333410 Chr.7: 130506488-130506488 on Build GRCh38
rs13312599 Chr.7: 130506486-130506486 on Build GRCh38
rs572304750 Chr.7: 130506595-130506595 on Build GRCh38
rs557024417 Chr.7: 130506412-130506412 on Build GRCh38
rs782729071 Chr.7: 130506588-130506588 on Build GRCh38
rs201408676 Chr.7: 130506325-130506325 on Build GRCh38
rs970776937 Chr.7: 130506573-130506573 on Build GRCh38
rs574415040 Chr.7: 130506318-130506318 on Build GRCh38
rs575309397 Chr.7: 130506455-130506455 on Build GRCh38
rs13234058 Chr.7: 130506320-130506320 on Build GRCh38
rs367603516 Chr.7: 130506501-130506501 on Build GRCh38
rs199960907 Chr.7: 130506501-130506501 on Build GRCh38
rs540028714 Chr.7: 130506510-130506510 on Build GRCh38
rs373685210 Chr.7: 130506134-130506134 on Build GRCh38
rs914452120 Chr.7: 130506625-130506625 on Build GRCh38
rs7779408 Chr.7: 130506473-130506473 on Build GRCh38
rs1019674621 Chr.7: 130506509-130506509 on Build GRCh38
rs952859446 Chr.7: 130506531-130506531 on Build GRCh38
rs371696273 Chr.7: 130506639-130506639 on Build GRCh38
rs782816719 Chr.7: 130506325-130506326 on Build GRCh38
rs1011318833 Chr.7: 130506423-130506423 on Build GRCh38
rs182560786 Chr.7: 130506628-130506628 on Build GRCh38
rs528294422 Chr.7: 130506508-130506508 on Build GRCh38
rs982913290 Chr.7: 130506537-130506537 on Build GRCh38
rs902906247 Chr.7: 130506492-130506492 on Build GRCh38
rs782487183 Chr.7: 130506457-130506458 on Build GRCh38
rs1062058 Chr.7: 130506485-130506485 on Build GRCh38
rs1803442 Chr.7: 130506153-130506153 on Build GRCh38
rs530172596 Chr.7: 130506577-130506577 on Build GRCh38
rs1056116350 Chr.7: 130506621-130506621 on Build GRCh38
rs997498779 Chr.7: 130506511-130506511 on Build GRCh38
rs1027194664 Chr.7: 130506518-130506518 on Build GRCh38
rs144636401 Chr.7: 130506541-130506541 on Build GRCh38
rs1055658200 Chr.7: 130506294-130506294 on Build GRCh38
rs61354155 Chr.7: 130506486-130506486 on Build GRCh38
rs1050667 Chr.7: 130506152-130506152 on Build GRCh38
rs565317091 Chr.7: 130506500-130506500 on Build GRCh38

View Full Product Details