Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 128837255-128837839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLNC
Entrez Gene ID
2318
Gene Name
filamin C
Gene Aliases
ABP-280, ABP280A, ABPA, ABPL, FLN2, MFM5, MPD4
UniGene
Hs.58414
Gene Chromosome Location
Chr.7: 128830429-128859274 on Build GRCh38
Transcripts
NM_001127487.1 NM_001458.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6212496 c.699+1G>T p.? Chr.7: 128837258-128837258 on Build GRCh38
5531344 c.915G>A p.A305A Chr.7: 128837701-128837701 on Build GRCh38
1447939 c.771C>T p.F257F Chr.7: 128837469-128837469 on Build GRCh38
1447938 c.741G>T p.E247D Chr.7: 128837439-128837439 on Build GRCh38
6210078 c.873C>T p.T291T Chr.7: 128837659-128837659 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs906826355 Chr.7: 128837297-128837297 on Build GRCh38
rs918460564 Chr.7: 128837809-128837809 on Build GRCh38
rs756104407 Chr.7: 128837729-128837729 on Build GRCh38
rs927080864 Chr.7: 128837816-128837816 on Build GRCh38
rs1020489966 Chr.7: 128837428-128837428 on Build GRCh38
rs749769428 Chr.7: 128837749-128837749 on Build GRCh38
rs745442558 Chr.7: 128837393-128837393 on Build GRCh38
rs966097091 Chr.7: 128837700-128837700 on Build GRCh38
rs199809140 Chr.7: 128837788-128837788 on Build GRCh38
rs776362922 Chr.7: 128837691-128837691 on Build GRCh38
rs201906839 Chr.7: 128837650-128837650 on Build GRCh38
rs758216356 Chr.7: 128837632-128837632 on Build GRCh38
rs984720410 Chr.7: 128837557-128837557 on Build GRCh38
rs200545315 Chr.7: 128837612-128837612 on Build GRCh38
rs751202105 Chr.7: 128837369-128837369 on Build GRCh38
rs540753816 Chr.7: 128837641-128837641 on Build GRCh38
rs2291559 Chr.7: 128837291-128837291 on Build GRCh38
rs2291560 Chr.7: 128837418-128837418 on Build GRCh38
rs778645211 Chr.7: 128837626-128837626 on Build GRCh38
rs1010523382 Chr.7: 128837703-128837703 on Build GRCh38
rs367600557 Chr.7: 128837322-128837322 on Build GRCh38
rs13237814 Chr.7: 128837491-128837491 on Build GRCh38
rs755583250 Chr.7: 128837503-128837503 on Build GRCh38
rs751466929 Chr.7: 128837709-128837709 on Build GRCh38
rs765604975 Chr.7: 128837475-128837475 on Build GRCh38
rs377172731 Chr.7: 128837445-128837445 on Build GRCh38
rs780677141 Chr.7: 128837388-128837388 on Build GRCh38
rs2291561 Chr.7: 128837493-128837493 on Build GRCh38
rs367579987 Chr.7: 128837292-128837292 on Build GRCh38
rs774743325 Chr.7: 128837682-128837682 on Build GRCh38
rs776614185 Chr.7: 128837567-128837567 on Build GRCh38
rs761442321 Chr.7: 128837484-128837484 on Build GRCh38
rs971262870 Chr.7: 128837810-128837810 on Build GRCh38
rs575141899 Chr.7: 128837683-128837683 on Build GRCh38
rs752652391 Chr.7: 128837701-128837701 on Build GRCh38
rs771789906 Chr.7: 128837405-128837405 on Build GRCh38
rs372747855 Chr.7: 128837633-128837633 on Build GRCh38
rs765545958 Chr.7: 128837769-128837769 on Build GRCh38
rs755636271 Chr.7: 128837394-128837394 on Build GRCh38
rs909002315 Chr.7: 128837541-128837541 on Build GRCh38
rs754527070 Chr.7: 128837624-128837624 on Build GRCh38
rs566461523 Chr.7: 128837430-128837430 on Build GRCh38
rs763968152 Chr.7: 128837705-128837705 on Build GRCh38
rs778626535 Chr.7: 128837417-128837417 on Build GRCh38
rs753795226 Chr.7: 128837526-128837526 on Build GRCh38
rs773579752 Chr.7: 128837429-128837429 on Build GRCh38
rs866326517 Chr.7: 128837715-128837715 on Build GRCh38
rs763238819 Chr.7: 128837569-128837569 on Build GRCh38
rs895373758 Chr.7: 128837671-128837671 on Build GRCh38
rs746121984 Chr.7: 128837728-128837728 on Build GRCh38
rs770479236 Chr.7: 128837596-128837596 on Build GRCh38
rs757478509 Chr.7: 128837639-128837639 on Build GRCh38
rs368602593 Chr.7: 128837552-128837552 on Build GRCh38
rs387906586 Chr.7: 128837450-128837450 on Build GRCh38
rs765885585 Chr.7: 128837684-128837684 on Build GRCh38
rs976043816 Chr.7: 128837489-128837489 on Build GRCh38
rs760219261 Chr.7: 128837278-128837278 on Build GRCh38
rs953269055 Chr.7: 128837521-128837521 on Build GRCh38
rs781212262 Chr.7: 128837711-128837711 on Build GRCh38
rs750082280 Chr.7: 128837502-128837502 on Build GRCh38
rs768452500 Chr.7: 128837410-128837410 on Build GRCh38
rs953435954 Chr.7: 128837759-128837759 on Build GRCh38
rs55907818 Chr.7: 128837566-128837566 on Build GRCh38
rs976229219 Chr.7: 128837481-128837481 on Build GRCh38
rs766932770 Chr.7: 128837307-128837307 on Build GRCh38
rs541392206 Chr.7: 128837710-128837710 on Build GRCh38
rs749217540 Chr.7: 128837409-128837409 on Build GRCh38
rs370330345 Chr.7: 128837480-128837480 on Build GRCh38
rs774847804 Chr.7: 128837768-128837768 on Build GRCh38
rs780144802 Chr.7: 128837741-128837741 on Build GRCh38
rs113049908 Chr.7: 128837524-128837524 on Build GRCh38
rs576908770 Chr.7: 128837630-128837630 on Build GRCh38
rs779455589 Chr.7: 128837513-128837513 on Build GRCh38
rs778751919 Chr.7: 128837536-128837536 on Build GRCh38
rs749823519 Chr.7: 128837659-128837659 on Build GRCh38
rs893364981 Chr.7: 128837381-128837381 on Build GRCh38
rs764476412 Chr.7: 128837573-128837573 on Build GRCh38
rs755329347 Chr.7: 128837367-128837367 on Build GRCh38
rs774154344 Chr.7: 128837302-128837302 on Build GRCh38
rs768975126 Chr.7: 128837665-128837665 on Build GRCh38
rs80097455 Chr.7: 128837422-128837422 on Build GRCh38
rs928331823 Chr.7: 128837591-128837591 on Build GRCh38
rs775809908 Chr.7: 128837653-128837653 on Build GRCh38
rs774595051 Chr.7: 128837282-128837282 on Build GRCh38
rs7787924 Chr.7: 128837811-128837811 on Build GRCh38
rs375465331 Chr.7: 128837602-128837602 on Build GRCh38
rs754916664 Chr.7: 128837535-128837535 on Build GRCh38
rs776079280 Chr.7: 128837568-128837568 on Build GRCh38
rs1054100328 Chr.7: 128837695-128837695 on Build GRCh38
rs765885505 Chr.7: 128837613-128837613 on Build GRCh38
rs1035650211 Chr.7: 128837312-128837312 on Build GRCh38
rs949853392 Chr.7: 128837357-128837357 on Build GRCh38
rs771164451 Chr.7: 128837439-128837439 on Build GRCh38
rs747255701 Chr.7: 128837561-128837561 on Build GRCh38
rs769206104 Chr.7: 128837755-128837755 on Build GRCh38
rs765541921 Chr.7: 128837368-128837368 on Build GRCh38
rs753342024 Chr.7: 128837616-128837616 on Build GRCh38
rs372769089 Chr.7: 128837303-128837303 on Build GRCh38
rs758924780 Chr.7: 128837692-128837692 on Build GRCh38
rs754303576 Chr.7: 128837366-128837366 on Build GRCh38
rs200471132 Chr.7: 128837704-128837704 on Build GRCh38
rs768342823 Chr.7: 128837279-128837279 on Build GRCh38
rs551132755 Chr.7: 128837372-128837372 on Build GRCh38
rs747837157 Chr.7: 128837545-128837545 on Build GRCh38
rs112479041 Chr.7: 128837550-128837550 on Build GRCh38
rs953416778 Chr.7: 128837765-128837765 on Build GRCh38
rs972658922 Chr.7: 128837802-128837802 on Build GRCh38
rs778100014 Chr.7: 128837556-128837556 on Build GRCh38
rs760009430 Chr.7: 128837465-128837465 on Build GRCh38
rs201239973 Chr.7: 128837698-128837698 on Build GRCh38
rs779902112 Chr.7: 128837403-128837403 on Build GRCh38

View Full Product Details