Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150692317-150692908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIMAP2
Entrez Gene ID
26157
Gene Name
GTPase, IMAP family member 2
Gene Aliases
HIMAP2, IAN12, IMAP2
UniGene
Hs.647071
Gene Chromosome Location
Chr.7: 150685706-150693641 on Build GRCh38
Transcripts
NM_015660.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7134205 c.548A>G p.N183S Chr.7: 150692834-150692834 on Build GRCh38
3636058 c.258C>T p.D86D Chr.7: 150692544-150692544 on Build GRCh38
485042 c.464C>A p.S155Y Chr.7: 150692750-150692750 on Build GRCh38
1087251 c.484G>A p.D162N Chr.7: 150692770-150692770 on Build GRCh38
3636060 c.411C>T p.A137A Chr.7: 150692697-150692697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372328919 Chr.7: 150692701-150692701 on Build GRCh38
rs755275076 Chr.7: 150692425-150692425 on Build GRCh38
rs375623579 Chr.7: 150692728-150692728 on Build GRCh38
rs745906034 Chr.7: 150692727-150692727 on Build GRCh38
rs772943762 Chr.7: 150692840-150692840 on Build GRCh38
rs267601408 Chr.7: 150692361-150692361 on Build GRCh38
rs753743840 Chr.7: 150692669-150692669 on Build GRCh38
rs892557833 Chr.7: 150692456-150692456 on Build GRCh38
rs763264836 Chr.7: 150692576-150692576 on Build GRCh38
rs777993215 Chr.7: 150692625-150692625 on Build GRCh38
rs752650056 Chr.7: 150692860-150692860 on Build GRCh38
rs758719032 Chr.7: 150692438-150692438 on Build GRCh38
rs927523889 Chr.7: 150692529-150692529 on Build GRCh38
rs146048659 Chr.7: 150692587-150692587 on Build GRCh38
rs573362845 Chr.7: 150692579-150692579 on Build GRCh38
rs940856948 Chr.7: 150692343-150692343 on Build GRCh38
rs918789969 Chr.7: 150692764-150692764 on Build GRCh38
rs142628430 Chr.7: 150692365-150692365 on Build GRCh38
rs759125462 Chr.7: 150692852-150692852 on Build GRCh38
rs562177098 Chr.7: 150692635-150692635 on Build GRCh38
rs769276327 Chr.7: 150692733-150692733 on Build GRCh38
rs144177217 Chr.7: 150692663-150692663 on Build GRCh38
rs751426384 Chr.7: 150692871-150692871 on Build GRCh38
rs769005291 Chr.7: 150692702-150692703 on Build GRCh38
rs774847313 Chr.7: 150692734-150692734 on Build GRCh38
rs370076034 Chr.7: 150692823-150692823 on Build GRCh38
rs932242424 Chr.7: 150692408-150692408 on Build GRCh38
rs201164211 Chr.7: 150692394-150692394 on Build GRCh38
rs760718539 Chr.7: 150692404-150692404 on Build GRCh38
rs985562937 Chr.7: 150692749-150692750 on Build GRCh38
rs767700202 Chr.7: 150692357-150692357 on Build GRCh38
rs2075078 Chr.7: 150692768-150692768 on Build GRCh38
rs775889884 Chr.7: 150692645-150692645 on Build GRCh38
rs913885573 Chr.7: 150692843-150692843 on Build GRCh38
rs61730412 Chr.7: 150692748-150692748 on Build GRCh38
rs141987826 Chr.7: 150692819-150692819 on Build GRCh38
rs977224341 Chr.7: 150692671-150692685 on Build GRCh38
rs572566547 Chr.7: 150692567-150692567 on Build GRCh38
rs938916927 Chr.7: 150692530-150692530 on Build GRCh38
rs761756140 Chr.7: 150692382-150692382 on Build GRCh38
rs746101312 Chr.7: 150692795-150692795 on Build GRCh38
rs756109142 Chr.7: 150692592-150692592 on Build GRCh38
rs914273730 Chr.7: 150692521-150692521 on Build GRCh38
rs371665331 Chr.7: 150692368-150692368 on Build GRCh38
rs760238269 Chr.7: 150692844-150692844 on Build GRCh38
rs200095405 Chr.7: 150692346-150692346 on Build GRCh38
rs11558054 Chr.7: 150692506-150692506 on Build GRCh38
rs1017549041 Chr.7: 150692626-150692626 on Build GRCh38
rs765788019 Chr.7: 150692561-150692561 on Build GRCh38
rs1050669351 Chr.7: 150692422-150692422 on Build GRCh38
rs576183114 Chr.7: 150692659-150692659 on Build GRCh38
rs146916816 Chr.7: 150692435-150692435 on Build GRCh38
rs1048532412 Chr.7: 150692771-150692771 on Build GRCh38
rs749095988 Chr.7: 150692497-150692497 on Build GRCh38
rs1044676631 Chr.7: 150692524-150692524 on Build GRCh38
rs756663731 Chr.7: 150692366-150692368 on Build GRCh38
rs372417348 Chr.7: 150692861-150692861 on Build GRCh38
rs868338737 Chr.7: 150692708-150692708 on Build GRCh38
rs781429107 Chr.7: 150692710-150692710 on Build GRCh38
rs765115379 Chr.7: 150692854-150692854 on Build GRCh38
rs753069853 Chr.7: 150692427-150692427 on Build GRCh38
rs142150251 Chr.7: 150692742-150692742 on Build GRCh38
rs754363729 Chr.7: 150692421-150692421 on Build GRCh38
rs150902506 Chr.7: 150692785-150692785 on Build GRCh38
rs776009895 Chr.7: 150692729-150692729 on Build GRCh38
rs765810038 Chr.7: 150692759-150692759 on Build GRCh38
rs994229863 Chr.7: 150692418-150692418 on Build GRCh38
rs749080530 Chr.7: 150692612-150692612 on Build GRCh38
rs779905535 Chr.7: 150692603-150692603 on Build GRCh38
rs766842234 Chr.7: 150692420-150692420 on Build GRCh38
rs141540795 Chr.7: 150692487-150692487 on Build GRCh38
rs778027630 Chr.7: 150692479-150692479 on Build GRCh38
rs1860871 Chr.7: 150692505-150692505 on Build GRCh38
rs763759831 Chr.7: 150692584-150692584 on Build GRCh38
rs368735702 Chr.7: 150692339-150692339 on Build GRCh38
rs770303254 Chr.7: 150692473-150692474 on Build GRCh38
rs780740133 Chr.7: 150692369-150692369 on Build GRCh38
rs912122284 Chr.7: 150692802-150692802 on Build GRCh38
rs775981582 Chr.7: 150692848-150692848 on Build GRCh38
rs748115373 Chr.7: 150692629-150692629 on Build GRCh38
rs550994929 Chr.7: 150692818-150692818 on Build GRCh38
rs776678996 Chr.7: 150692634-150692634 on Build GRCh38
rs777336192 Chr.7: 150692436-150692436 on Build GRCh38
rs756855631 Chr.7: 150692461-150692461 on Build GRCh38
rs752453732 Chr.7: 150692777-150692777 on Build GRCh38
rs1052411631 Chr.7: 150692825-150692825 on Build GRCh38
rs751159496 Chr.7: 150692654-150692654 on Build GRCh38
rs756154784 Chr.7: 150692807-150692807 on Build GRCh38
rs746494122 Chr.7: 150692455-150692455 on Build GRCh38
rs772114017 Chr.7: 150692880-150692914 on Build GRCh38
rs145172862 Chr.7: 150692781-150692781 on Build GRCh38
rs200801747 Chr.7: 150692345-150692345 on Build GRCh38
rs988560266 Chr.7: 150692698-150692698 on Build GRCh38
rs770299527 Chr.7: 150692642-150692642 on Build GRCh38
rs76850019 Chr.7: 150692522-150692522 on Build GRCh38
rs375147167 Chr.7: 150692544-150692544 on Build GRCh38
rs778956614 Chr.7: 150692750-150692750 on Build GRCh38
rs747233976 Chr.7: 150692628-150692628 on Build GRCh38
rs748484310 Chr.7: 150692826-150692826 on Build GRCh38
rs35516375 Chr.7: 150692731-150692732 on Build GRCh38
rs146899946 Chr.7: 150692839-150692839 on Build GRCh38
rs368046535 Chr.7: 150692590-150692590 on Build GRCh38
rs765269305 Chr.7: 150692772-150692772 on Build GRCh38
rs773329340 Chr.7: 150692387-150692387 on Build GRCh38
rs17173567 Chr.7: 150692741-150692741 on Build GRCh38
rs6952002 Chr.7: 150692589-150692589 on Build GRCh38
rs766111422 Chr.7: 150692846-150692846 on Build GRCh38
rs779995737 Chr.7: 150692808-150692808 on Build GRCh38
rs760769724 Chr.7: 150692519-150692519 on Build GRCh38
rs749632810 Chr.7: 150692700-150692701 on Build GRCh38
rs201449718 Chr.7: 150692699-150692699 on Build GRCh38
rs754733684 Chr.7: 150692675-150692675 on Build GRCh38
rs865867848 Chr.7: 150692482-150692482 on Build GRCh38
rs763378192 Chr.7: 150692732-150692732 on Build GRCh38
rs866038452 Chr.7: 150692426-150692426 on Build GRCh38
rs760030527 Chr.7: 150692546-150692546 on Build GRCh38
rs267601409 Chr.7: 150692758-150692758 on Build GRCh38
rs964597045 Chr.7: 150692470-150692470 on Build GRCh38
rs748392759 Chr.7: 150692848-150692848 on Build GRCh38
rs888485556 Chr.7: 150692595-150692595 on Build GRCh38
rs143364322 Chr.7: 150692486-150692486 on Build GRCh38
rs766954394 Chr.7: 150692449-150692449 on Build GRCh38
rs768369164 Chr.7: 150692618-150692618 on Build GRCh38
rs952654407 Chr.7: 150692429-150692429 on Build GRCh38
rs780589798 Chr.7: 150692497-150692498 on Build GRCh38
rs556326889 Chr.7: 150692652-150692652 on Build GRCh38
rs762317176 Chr.7: 150692736-150692736 on Build GRCh38
rs754857410 Chr.7: 150692769-150692769 on Build GRCh38
rs763883328 Chr.7: 150692869-150692869 on Build GRCh38
rs144149379 Chr.7: 150692660-150692660 on Build GRCh38
rs771115951 Chr.7: 150692705-150692705 on Build GRCh38

View Full Product Details