Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 25469639-25470122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCA2
Entrez Gene ID
157313
Gene Name
cell division cycle associated 2
Gene Aliases
PPP1R81, Repo-Man
UniGene
Hs.33366
Gene Chromosome Location
Chr.8: 25458569-25511308 on Build GRCh38
Transcripts
NM_001317906.1 NM_001317907.1 NM_152562.3 XM_006716294.2 XM_011544415.1 XM_011544416.2 XM_011544417.2 XM_011544418.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6894796 c.736-1G>T p.? Chr.8: 25469895-25469895 on Build GRCh38
4952866 c.740C>T p.S247L Chr.8: 25469900-25469900 on Build GRCh38
6472499 c.790C>T p.P264S Chr.8: 25469950-25469950 on Build GRCh38
1200392 c.754T>C p.S252P Chr.8: 25469914-25469914 on Build GRCh38
486333 c.736A>G p.I246V Chr.8: 25469896-25469896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024110257 Chr.8: 25469816-25469816 on Build GRCh38
rs920438885 Chr.8: 25469822-25469822 on Build GRCh38
rs754321913 Chr.8: 25469944-25469944 on Build GRCh38
rs374118135 Chr.8: 25469760-25469760 on Build GRCh38
rs377071707 Chr.8: 25469971-25469971 on Build GRCh38
rs759882033 Chr.8: 25469936-25469936 on Build GRCh38
rs559100249 Chr.8: 25469692-25469692 on Build GRCh38
rs149146216 Chr.8: 25469905-25469905 on Build GRCh38
rs780717343 Chr.8: 25469962-25469962 on Build GRCh38
rs749290315 Chr.8: 25469989-25469989 on Build GRCh38
rs111985141 Chr.8: 25469752-25469752 on Build GRCh38
rs922533253 Chr.8: 25469932-25469932 on Build GRCh38
rs138388292 Chr.8: 25469762-25469763 on Build GRCh38
rs770965988 Chr.8: 25470000-25470000 on Build GRCh38
rs760900612 Chr.8: 25470027-25470027 on Build GRCh38
rs376009989 Chr.8: 25469917-25469917 on Build GRCh38
rs539271312 Chr.8: 25469843-25469845 on Build GRCh38
rs779198779 Chr.8: 25469852-25469852 on Build GRCh38
rs553809791 Chr.8: 25469898-25469898 on Build GRCh38
rs78337462 Chr.8: 25469705-25469762 on Build GRCh38
rs753922023 Chr.8: 25470030-25470030 on Build GRCh38
rs111539508 Chr.8: 25469706-25469706 on Build GRCh38
rs929184400 Chr.8: 25469840-25469840 on Build GRCh38
rs111337819 Chr.8: 25469765-25469766 on Build GRCh38
rs869064541 Chr.8: 25469706-25469706 on Build GRCh38
rs757647642 Chr.8: 25469850-25469850 on Build GRCh38
rs372954389 Chr.8: 25469752-25469752 on Build GRCh38
rs781030066 Chr.8: 25469845-25469845 on Build GRCh38
rs184433724 Chr.8: 25469953-25469953 on Build GRCh38
rs752880487 Chr.8: 25469939-25469939 on Build GRCh38
rs770100369 Chr.8: 25469705-25469706 on Build GRCh38
rs7817239 Chr.8: 25469668-25469668 on Build GRCh38
rs113161284 Chr.8: 25469761-25469761 on Build GRCh38
rs764128356 Chr.8: 25470028-25470028 on Build GRCh38
rs960288065 Chr.8: 25469978-25469978 on Build GRCh38
rs370458379 Chr.8: 25469846-25469846 on Build GRCh38
rs189118174 Chr.8: 25469787-25469787 on Build GRCh38
rs771539879 Chr.8: 25469877-25469878 on Build GRCh38
rs890418298 Chr.8: 25469881-25469881 on Build GRCh38
rs754679065 Chr.8: 25469955-25469955 on Build GRCh38
rs952146588 Chr.8: 25469824-25469824 on Build GRCh38
rs528863533 Chr.8: 25470093-25470093 on Build GRCh38
rs370349316 Chr.8: 25469751-25469751 on Build GRCh38
rs112567636 Chr.8: 25469706-25469711 on Build GRCh38
rs944341772 Chr.8: 25469912-25469912 on Build GRCh38
rs372838340 Chr.8: 25470017-25470017 on Build GRCh38
rs904669969 Chr.8: 25469957-25469957 on Build GRCh38
rs371818154 Chr.8: 25470016-25470016 on Build GRCh38
rs764170658 Chr.8: 25469941-25469941 on Build GRCh38
rs188480168 Chr.8: 25470041-25470041 on Build GRCh38
rs372257172 Chr.8: 25469923-25469923 on Build GRCh38
rs757737568 Chr.8: 25469947-25469947 on Build GRCh38
rs200437030 Chr.8: 25469705-25469710 on Build GRCh38
rs920715500 Chr.8: 25470059-25470059 on Build GRCh38
rs776267395 Chr.8: 25469872-25469872 on Build GRCh38
rs775581196 Chr.8: 25470019-25470019 on Build GRCh38
rs191970295 Chr.8: 25469883-25469883 on Build GRCh38
rs62502024 Chr.8: 25470055-25470055 on Build GRCh38
rs929129351 Chr.8: 25470083-25470083 on Build GRCh38
rs756163402 Chr.8: 25469940-25469940 on Build GRCh38
rs372654767 Chr.8: 25469706-25469762 on Build GRCh38
rs574458102 Chr.8: 25469860-25469860 on Build GRCh38
rs556148517 Chr.8: 25469803-25469803 on Build GRCh38
rs902152802 Chr.8: 25470072-25470072 on Build GRCh38
rs570659813 Chr.8: 25469698-25469698 on Build GRCh38
rs768265765 Chr.8: 25469866-25469866 on Build GRCh38
rs111841366 Chr.8: 25469762-25469762 on Build GRCh38
rs561679144 Chr.8: 25470070-25470070 on Build GRCh38
rs370100608 Chr.8: 25469759-25469759 on Build GRCh38
rs115799637 Chr.8: 25469970-25469970 on Build GRCh38
rs985320803 Chr.8: 25469843-25469843 on Build GRCh38
rs773025919 Chr.8: 25469899-25469899 on Build GRCh38
rs778492535 Chr.8: 25469707-25469712 on Build GRCh38
rs368609661 Chr.8: 25470013-25470013 on Build GRCh38
rs747766762 Chr.8: 25469966-25469966 on Build GRCh38
rs774268934 Chr.8: 25469921-25469921 on Build GRCh38
rs369940388 Chr.8: 25469722-25469723 on Build GRCh38
rs999149317 Chr.8: 25470077-25470077 on Build GRCh38
rs112227549 Chr.8: 25469707-25469707 on Build GRCh38
rs762373408 Chr.8: 25470032-25470032 on Build GRCh38

View Full Product Details