Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 52538119-52538691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM150A
Entrez Gene ID
389658
Gene Name
family with sequence similarity 150 member A
Gene Aliases
UNQ9433
UniGene
Hs.527111
Gene Chromosome Location
Chr.8: 52534037-52565461 on Build GRCh38
Transcripts
NM_207413.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5011912 c.359T>G p.L120* Chr.8: 52538474-52538474 on Build GRCh38
6456048 c.349T>A p.L117I Chr.8: 52538484-52538484 on Build GRCh38
4924190 c.337T>C p.C113R Chr.8: 52538496-52538496 on Build GRCh38
3716070 c.342T>C p.A114A Chr.8: 52538491-52538491 on Build GRCh38
7160334 c.366G>C p.V122V Chr.8: 52538467-52538467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989323900 Chr.8: 52538600-52538600 on Build GRCh38
rs750974368 Chr.8: 52538442-52538442 on Build GRCh38
rs927720846 Chr.8: 52538632-52538632 on Build GRCh38
rs757531984 Chr.8: 52538399-52538399 on Build GRCh38
rs28694761 Chr.8: 52538618-52538618 on Build GRCh38
rs543774462 Chr.8: 52538323-52538323 on Build GRCh38
rs985251567 Chr.8: 52538243-52538243 on Build GRCh38
rs936560149 Chr.8: 52538340-52538340 on Build GRCh38
rs989351388 Chr.8: 52538342-52538342 on Build GRCh38
rs781644301 Chr.8: 52538431-52538432 on Build GRCh38
rs761026019 Chr.8: 52538422-52538422 on Build GRCh38
rs772040481 Chr.8: 52538532-52538532 on Build GRCh38
rs945137107 Chr.8: 52538337-52538337 on Build GRCh38
rs765626939 Chr.8: 52538555-52538555 on Build GRCh38
rs916451995 Chr.8: 52538625-52538625 on Build GRCh38
rs981848868 Chr.8: 52538629-52538629 on Build GRCh38
rs769736765 Chr.8: 52538508-52538508 on Build GRCh38
rs189118210 Chr.8: 52538441-52538441 on Build GRCh38
rs764234832 Chr.8: 52538424-52538424 on Build GRCh38
rs763025988 Chr.8: 52538522-52538522 on Build GRCh38
rs572031058 Chr.8: 52538311-52538311 on Build GRCh38
rs757647599 Chr.8: 52538440-52538440 on Build GRCh38
rs773316292 Chr.8: 52538403-52538403 on Build GRCh38
rs894718346 Chr.8: 52538468-52538468 on Build GRCh38
rs1046487085 Chr.8: 52538382-52538382 on Build GRCh38
rs373158587 Chr.8: 52538476-52538476 on Build GRCh38
rs563973612 Chr.8: 52538416-52538416 on Build GRCh38
rs564200296 Chr.8: 52538188-52538188 on Build GRCh38
rs750923236 Chr.8: 52538559-52538559 on Build GRCh38
rs986478638 Chr.8: 52538201-52538201 on Build GRCh38
rs779678030 Chr.8: 52538473-52538473 on Build GRCh38
rs13280500 Chr.8: 52538409-52538409 on Build GRCh38
rs957984380 Chr.8: 52538598-52538598 on Build GRCh38
rs957680278 Chr.8: 52538595-52538595 on Build GRCh38
rs910458673 Chr.8: 52538296-52538296 on Build GRCh38
rs947865292 Chr.8: 52538372-52538372 on Build GRCh38
rs985976964 Chr.8: 52538576-52538576 on Build GRCh38
rs531708639 Chr.8: 52538320-52538320 on Build GRCh38
rs760673860 Chr.8: 52538537-52538537 on Build GRCh38
rs995111659 Chr.8: 52538520-52538520 on Build GRCh38
rs1030651386 Chr.8: 52538591-52538591 on Build GRCh38
rs938883123 Chr.8: 52538655-52538655 on Build GRCh38
rs183239937 Chr.8: 52538170-52538170 on Build GRCh38
rs747915262 Chr.8: 52538488-52538488 on Build GRCh38
rs554327449 Chr.8: 52538160-52538160 on Build GRCh38
rs752104284 Chr.8: 52538480-52538480 on Build GRCh38
rs752831019 Chr.8: 52538420-52538420 on Build GRCh38
rs572352926 Chr.8: 52538169-52538169 on Build GRCh38
rs972101399 Chr.8: 52538156-52538156 on Build GRCh38
rs1019362566 Chr.8: 52538556-52538556 on Build GRCh38
rs755621671 Chr.8: 52538517-52538517 on Build GRCh38
rs754164128 Chr.8: 52538427-52538427 on Build GRCh38
rs758856914 Chr.8: 52538349-52538349 on Build GRCh38
rs759640619 Chr.8: 52538405-52538405 on Build GRCh38
rs111987541 Chr.8: 52538580-52538580 on Build GRCh38
rs193045818 Chr.8: 52538539-52538539 on Build GRCh38
rs532840607 Chr.8: 52538575-52538575 on Build GRCh38
rs945020761 Chr.8: 52538623-52538623 on Build GRCh38
rs866998488 Chr.8: 52538147-52538147 on Build GRCh38
rs762142173 Chr.8: 52538542-52538542 on Build GRCh38
rs953668996 Chr.8: 52538236-52538236 on Build GRCh38
rs778513846 Chr.8: 52538322-52538322 on Build GRCh38
rs1019770320 Chr.8: 52538252-52538253 on Build GRCh38
rs954538300 Chr.8: 52538175-52538175 on Build GRCh38
rs1015201798 Chr.8: 52538162-52538162 on Build GRCh38
rs146291795 Chr.8: 52538667-52538667 on Build GRCh38
rs780717044 Chr.8: 52538478-52538478 on Build GRCh38
rs757870287 Chr.8: 52538470-52538470 on Build GRCh38
rs1029195944 Chr.8: 52538218-52538218 on Build GRCh38
rs775698873 Chr.8: 52538535-52538535 on Build GRCh38
rs768301155 Chr.8: 52538477-52538477 on Build GRCh38

View Full Product Details