Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 104355833-104356345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCSTAMP
Entrez Gene ID
81501
Gene Name
dendrocyte expressed seven transmembrane protein
Gene Aliases
FIND, TM7SF4, hDC-STAMP
UniGene
Hs.652230
Gene Chromosome Location
Chr.8: 104339087-104356874 on Build GRCh38
Transcripts
NM_001257317.1 NM_030788.3 XM_011517321.1 XM_011517324.1 XM_017013878.1 XM_017013879.1 XM_017013880.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7114884 c.1405A>C p.K469Q Chr.8: 104356190-104356190 on Build GRCh38
4626101 c.1355C>T p.P452L Chr.8: 104356140-104356140 on Build GRCh38
3643679 c.1359C>T p.V453V Chr.8: 104356144-104356144 on Build GRCh38
4394932 c.1374G>A p.R458R Chr.8: 104356159-104356159 on Build GRCh38
323853 c.1373G>C p.R458T Chr.8: 104356158-104356158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021161866 Chr.8: 104356310-104356310 on Build GRCh38
rs527713692 Chr.8: 104356058-104356058 on Build GRCh38
rs765732707 Chr.8: 104356241-104356241 on Build GRCh38
rs1007229559 Chr.8: 104356020-104356020 on Build GRCh38
rs898086164 Chr.8: 104355902-104355902 on Build GRCh38
rs139019600 Chr.8: 104355876-104355876 on Build GRCh38
rs770721642 Chr.8: 104356227-104356227 on Build GRCh38
rs769735055 Chr.8: 104356097-104356097 on Build GRCh38
rs146388338 Chr.8: 104356228-104356228 on Build GRCh38
rs371471567 Chr.8: 104356231-104356231 on Build GRCh38
rs760216940 Chr.8: 104356239-104356239 on Build GRCh38
rs897024088 Chr.8: 104356174-104356174 on Build GRCh38
rs756451858 Chr.8: 104356069-104356069 on Build GRCh38
rs371947817 Chr.8: 104356196-104356196 on Build GRCh38
rs958501278 Chr.8: 104355960-104355960 on Build GRCh38
rs373966410 Chr.8: 104356195-104356195 on Build GRCh38
rs572674739 Chr.8: 104356303-104356303 on Build GRCh38
rs750595152 Chr.8: 104355937-104355937 on Build GRCh38
rs901484447 Chr.8: 104356056-104356056 on Build GRCh38
rs1031573219 Chr.8: 104356067-104356067 on Build GRCh38
rs749267145 Chr.8: 104356199-104356200 on Build GRCh38
rs989994384 Chr.8: 104356105-104356105 on Build GRCh38
rs557699044 Chr.8: 104356314-104356314 on Build GRCh38
rs759127030 Chr.8: 104356144-104356144 on Build GRCh38
rs1056696252 Chr.8: 104355875-104355875 on Build GRCh38
rs953730142 Chr.8: 104355857-104355857 on Build GRCh38
rs989732613 Chr.8: 104355972-104355972 on Build GRCh38
rs1017310189 Chr.8: 104356049-104356049 on Build GRCh38
rs781456540 Chr.8: 104356206-104356206 on Build GRCh38
rs762265625 Chr.8: 104356172-104356172 on Build GRCh38
rs541715391 Chr.8: 104356281-104356281 on Build GRCh38
rs762789596 Chr.8: 104356102-104356102 on Build GRCh38
rs748576990 Chr.8: 104356291-104356291 on Build GRCh38
rs755206468 Chr.8: 104356203-104356203 on Build GRCh38
rs4734780 Chr.8: 104356180-104356180 on Build GRCh38
rs772923786 Chr.8: 104356098-104356098 on Build GRCh38
rs768862327 Chr.8: 104356078-104356078 on Build GRCh38
rs771680242 Chr.8: 104356232-104356232 on Build GRCh38
rs758786657 Chr.8: 104356190-104356190 on Build GRCh38
rs765750442 Chr.8: 104356179-104356179 on Build GRCh38
rs748231261 Chr.8: 104356207-104356207 on Build GRCh38
rs547681147 Chr.8: 104355987-104355987 on Build GRCh38
rs185869734 Chr.8: 104356312-104356312 on Build GRCh38
rs535760250 Chr.8: 104356288-104356288 on Build GRCh38
rs1055507800 Chr.8: 104356103-104356103 on Build GRCh38
rs773993853 Chr.8: 104356136-104356136 on Build GRCh38
rs781362692 Chr.8: 104356086-104356086 on Build GRCh38
rs777759392 Chr.8: 104356211-104356211 on Build GRCh38
rs779965454 Chr.8: 104356073-104356074 on Build GRCh38
rs71520881 Chr.8: 104356126-104356126 on Build GRCh38
rs992597329 Chr.8: 104356184-104356184 on Build GRCh38
rs565949296 Chr.8: 104356027-104356027 on Build GRCh38
rs115926403 Chr.8: 104356071-104356071 on Build GRCh38
rs560113860 Chr.8: 104356297-104356297 on Build GRCh38
rs1036937032 Chr.8: 104356166-104356166 on Build GRCh38
rs767032498 Chr.8: 104356147-104356147 on Build GRCh38
rs536009866 Chr.8: 104356226-104356226 on Build GRCh38
rs1006569406 Chr.8: 104356275-104356275 on Build GRCh38
rs139972199 Chr.8: 104356152-104356152 on Build GRCh38
rs985083652 Chr.8: 104355862-104355862 on Build GRCh38
rs369750036 Chr.8: 104356237-104356237 on Build GRCh38
rs945542930 Chr.8: 104355998-104355998 on Build GRCh38
rs910811459 Chr.8: 104355957-104355957 on Build GRCh38

View Full Product Details