Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207898844-207899374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD34
Entrez Gene ID
947
Gene Name
CD34 molecule
Gene Aliases
-
UniGene
Hs.374990
Gene Chromosome Location
Chr.1: 207886538-207911338 on Build GRCh38
Transcripts
NM_001025109.1 NM_001773.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4135944 c.298A>C p.T100P Chr.1: 207899191-207899191 on Build GRCh38
903226 c.378C>T p.N126N Chr.1: 207899111-207899111 on Build GRCh38
1928771 c.330C>T p.V110V Chr.1: 207899159-207899159 on Build GRCh38
6460009 c.263-1G>A p.? Chr.1: 207899227-207899227 on Build GRCh38
1179012 c.499A>G p.I167V Chr.1: 207898990-207898990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901349924 Chr.1: 207899296-207899296 on Build GRCh38
rs963310956 Chr.1: 207898884-207898884 on Build GRCh38
rs554528108 Chr.1: 207899351-207899351 on Build GRCh38
rs1050022972 Chr.1: 207899342-207899342 on Build GRCh38
rs148688256 Chr.1: 207899091-207899091 on Build GRCh38
rs767549906 Chr.1: 207899212-207899212 on Build GRCh38
rs868211780 Chr.1: 207898992-207898992 on Build GRCh38
rs747179677 Chr.1: 207898994-207898996 on Build GRCh38
rs367645253 Chr.1: 207899198-207899198 on Build GRCh38
rs144312803 Chr.1: 207899222-207899222 on Build GRCh38
rs749484739 Chr.1: 207899011-207899011 on Build GRCh38
rs62620011 Chr.1: 207898960-207898960 on Build GRCh38
rs35659831 Chr.1: 207899001-207899002 on Build GRCh38
rs376393304 Chr.1: 207898942-207898942 on Build GRCh38
rs556737151 Chr.1: 207899217-207899217 on Build GRCh38
rs12029239 Chr.1: 207899281-207899281 on Build GRCh38
rs1026255950 Chr.1: 207899315-207899315 on Build GRCh38
rs757085668 Chr.1: 207898987-207898987 on Build GRCh38
rs560113399 Chr.1: 207899005-207899005 on Build GRCh38
rs752810474 Chr.1: 207899269-207899269 on Build GRCh38
rs774973450 Chr.1: 207899164-207899164 on Build GRCh38
rs184763697 Chr.1: 207899118-207899118 on Build GRCh38
rs755837615 Chr.1: 207899223-207899223 on Build GRCh38
rs913887476 Chr.1: 207899244-207899244 on Build GRCh38
rs199980221 Chr.1: 207898936-207898936 on Build GRCh38
rs149445501 Chr.1: 207899075-207899075 on Build GRCh38
rs755312965 Chr.1: 207899132-207899132 on Build GRCh38
rs942542419 Chr.1: 207899024-207899024 on Build GRCh38
rs368829114 Chr.1: 207899136-207899136 on Build GRCh38
rs754126325 Chr.1: 207899124-207899124 on Build GRCh38
rs146237257 Chr.1: 207899110-207899110 on Build GRCh38
rs772025818 Chr.1: 207899147-207899147 on Build GRCh38
rs778363505 Chr.1: 207899148-207899148 on Build GRCh38
rs768891729 Chr.1: 207899180-207899182 on Build GRCh38
rs777644382 Chr.1: 207899272-207899272 on Build GRCh38
rs772955563 Chr.1: 207899219-207899220 on Build GRCh38
rs535806154 Chr.1: 207899321-207899321 on Build GRCh38
rs542222417 Chr.1: 207899082-207899082 on Build GRCh38
rs371116376 Chr.1: 207899032-207899032 on Build GRCh38
rs374693020 Chr.1: 207898916-207898916 on Build GRCh38
rs758534429 Chr.1: 207899271-207899271 on Build GRCh38
rs773563879 Chr.1: 207899049-207899049 on Build GRCh38
rs201672357 Chr.1: 207899084-207899084 on Build GRCh38
rs772704456 Chr.1: 207899043-207899043 on Build GRCh38
rs751495564 Chr.1: 207899112-207899112 on Build GRCh38
rs757151224 Chr.1: 207899116-207899116 on Build GRCh38
rs763474226 Chr.1: 207899177-207899177 on Build GRCh38
rs949326279 Chr.1: 207898882-207898882 on Build GRCh38
rs370364374 Chr.1: 207899111-207899111 on Build GRCh38
rs748164623 Chr.1: 207899034-207899034 on Build GRCh38
rs780448744 Chr.1: 207899000-207899000 on Build GRCh38
rs933405933 Chr.1: 207898946-207898946 on Build GRCh38
rs766062512 Chr.1: 207898941-207898941 on Build GRCh38
rs746886232 Chr.1: 207898968-207898968 on Build GRCh38
rs761074750 Chr.1: 207899057-207899057 on Build GRCh38
rs757840744 Chr.1: 207899107-207899107 on Build GRCh38
rs758294686 Chr.1: 207898961-207898961 on Build GRCh38
rs200978602 Chr.1: 207899256-207899256 on Build GRCh38
rs1016943516 Chr.1: 207898867-207898867 on Build GRCh38
rs762571077 Chr.1: 207899173-207899173 on Build GRCh38
rs769556201 Chr.1: 207899159-207899159 on Build GRCh38
rs1002790192 Chr.1: 207899162-207899162 on Build GRCh38
rs773952663 Chr.1: 207899284-207899284 on Build GRCh38
rs747471462 Chr.1: 207899153-207899153 on Build GRCh38
rs779002641 Chr.1: 207899134-207899134 on Build GRCh38
rs759880989 Chr.1: 207899156-207899156 on Build GRCh38
rs568752718 Chr.1: 207899308-207899308 on Build GRCh38
rs373027811 Chr.1: 207899076-207899076 on Build GRCh38
rs376801128 Chr.1: 207899014-207899014 on Build GRCh38
rs368478737 Chr.1: 207899275-207899275 on Build GRCh38
rs777554605 Chr.1: 207898967-207898967 on Build GRCh38
rs201559434 Chr.1: 207899206-207899206 on Build GRCh38
rs373770382 Chr.1: 207899154-207899154 on Build GRCh38
rs766678767 Chr.1: 207899070-207899070 on Build GRCh38
rs2724368 Chr.1: 207899352-207899352 on Build GRCh38
rs370014441 Chr.1: 207898948-207898948 on Build GRCh38
rs972049398 Chr.1: 207898895-207898895 on Build GRCh38
rs759544403 Chr.1: 207899204-207899204 on Build GRCh38

View Full Product Details