Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 15778036-15778611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM188A
Entrez Gene ID
80013
Gene Name
family with sequence similarity 188 member A
Gene Aliases
C10orf97, CARP, DERP5, MST126, MSTP126, my042
UniGene
Hs.158870
Gene Chromosome Location
Chr.10: 15778169-15860533 on Build GRCh38
Transcripts
NM_001318330.1 NM_024948.3 XM_005252600.2 XM_005252602.2 XM_006717508.2 XM_011519693.1 XM_011519694.1 XM_017016672.1 XM_017016673.1 XM_017016674.1 XM_017016675.1 XM_017016676.1

SNP Detail

SNP ID Chromosome Location
rs966462377 Chr.10: 15778123-15778123 on Build GRCh38
rs748096161 Chr.10: 15778133-15778134 on Build GRCh38
rs572598589 Chr.10: 15778066-15778066 on Build GRCh38
rs1041720864 Chr.10: 15778131-15778131 on Build GRCh38
rs181089347 Chr.10: 15778101-15778101 on Build GRCh38
rs982152396 Chr.10: 15778273-15778273 on Build GRCh38
rs562907997 Chr.10: 15778250-15778252 on Build GRCh38
rs957668385 Chr.10: 15778107-15778107 on Build GRCh38
rs971157431 Chr.10: 15778267-15778267 on Build GRCh38
rs929134935 Chr.10: 15778153-15778153 on Build GRCh38
rs577275959 Chr.10: 15778225-15778225 on Build GRCh38
rs1026336188 Chr.10: 15778505-15778505 on Build GRCh38
rs999621514 Chr.10: 15778152-15778152 on Build GRCh38
rs1049403971 Chr.10: 15778090-15778090 on Build GRCh38
rs1042151882 Chr.10: 15778262-15778262 on Build GRCh38
rs776556593 Chr.10: 15778148-15778148 on Build GRCh38
rs574486092 Chr.10: 15778329-15778329 on Build GRCh38
rs147524178 Chr.10: 15778340-15778340 on Build GRCh38
rs986524558 Chr.10: 15778108-15778108 on Build GRCh38
rs999006342 Chr.10: 15778290-15778290 on Build GRCh38
rs894004393 Chr.10: 15778541-15778541 on Build GRCh38
rs531308639 Chr.10: 15778497-15778497 on Build GRCh38
rs55753158 Chr.10: 15778253-15778255 on Build GRCh38
rs565333871 Chr.10: 15778219-15778219 on Build GRCh38
rs552514596 Chr.10: 15778506-15778506 on Build GRCh38
rs570298301 Chr.10: 15778251-15778251 on Build GRCh38
rs919640919 Chr.10: 15778075-15778075 on Build GRCh38
rs143312581 Chr.10: 15778226-15778226 on Build GRCh38
rs868814032 Chr.10: 15778420-15778420 on Build GRCh38
rs866864602 Chr.10: 15778096-15778096 on Build GRCh38
rs182118735 Chr.10: 15778466-15778466 on Build GRCh38
rs188038464 Chr.10: 15778580-15778580 on Build GRCh38
rs990717630 Chr.10: 15778524-15778524 on Build GRCh38
rs972573480 Chr.10: 15778061-15778061 on Build GRCh38
rs1018278107 Chr.10: 15778362-15778362 on Build GRCh38
rs751027297 Chr.10: 15778407-15778407 on Build GRCh38
rs909107417 Chr.10: 15778217-15778217 on Build GRCh38
rs138558162 Chr.10: 15778517-15778517 on Build GRCh38
rs778451190 Chr.10: 15778244-15778244 on Build GRCh38
rs984851547 Chr.10: 15778515-15778515 on Build GRCh38
rs185528232 Chr.10: 15778277-15778277 on Build GRCh38
rs966040266 Chr.10: 15778369-15778369 on Build GRCh38
rs893308739 Chr.10: 15778332-15778332 on Build GRCh38
rs775300097 Chr.10: 15778291-15778291 on Build GRCh38
rs562125019 Chr.10: 15778288-15778288 on Build GRCh38
rs750991237 Chr.10: 15778585-15778585 on Build GRCh38
rs879604399 Chr.10: 15778163-15778163 on Build GRCh38
rs770854212 Chr.10: 15778110-15778110 on Build GRCh38
rs371376516 Chr.10: 15778250-15778250 on Build GRCh38
rs952487861 Chr.10: 15778365-15778365 on Build GRCh38
rs747953002 Chr.10: 15778385-15778385 on Build GRCh38
rs772992536 Chr.10: 15778582-15778585 on Build GRCh38
rs1012312033 Chr.10: 15778554-15778554 on Build GRCh38
rs931053898 Chr.10: 15778078-15778078 on Build GRCh38
rs190229074 Chr.10: 15778428-15778428 on Build GRCh38
rs768077823 Chr.10: 15778436-15778436 on Build GRCh38
rs781069962 Chr.10: 15778364-15778364 on Build GRCh38
rs943680905 Chr.10: 15778403-15778403 on Build GRCh38
rs1007788828 Chr.10: 15778349-15778349 on Build GRCh38
rs910525516 Chr.10: 15778114-15778114 on Build GRCh38
rs961290900 Chr.10: 15778302-15778302 on Build GRCh38
rs541902761 Chr.10: 15778338-15778338 on Build GRCh38
rs371696730 Chr.10: 15778558-15778558 on Build GRCh38
rs1023620361 Chr.10: 15778265-15778265 on Build GRCh38
rs1033210293 Chr.10: 15778300-15778300 on Build GRCh38
rs910873655 Chr.10: 15778387-15778387 on Build GRCh38
rs944289582 Chr.10: 15778126-15778126 on Build GRCh38
rs762486275 Chr.10: 15778359-15778359 on Build GRCh38
rs541551767 Chr.10: 15778521-15778521 on Build GRCh38

View Full Product Details