Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 59245683-59246250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM13C
Entrez Gene ID
220965
Gene Name
family with sequence similarity 13 member C
Gene Aliases
FAM13C1
UniGene
Hs.607594
Gene Chromosome Location
Chr.10: 59245933-59363186 on Build GRCh38
Transcripts
NM_001001971.2 NM_001143773.1 NM_001166698.1 NM_198215.3 XM_005269618.4 XM_005269619.4 XM_006717702.3 XM_006717703.3 XM_011539490.2 XM_011539491.2 XM_017015885.1 XM_017015886.1 XM_017015887.1 XM_017015888.1 XM_017015889.1 XM_017015890.1 XM_017015891.1 XM_017015892.1 XM_017015893.1 NM_001143774.1 NM_032439.3 XM_011540275.2 XM_011540276.2 XM_017016782.1 XM_017016783.1

Gene Symbol
PHYHIPL
Entrez Gene ID
84457
Gene Name
phytanoyl-CoA 2-hydroxylase interacting protein like
Gene Aliases
-
UniGene
Hs.499704
Gene Chromosome Location
Chr.10: 59174811-59247774 on Build GRCh38
Transcripts
NM_001001971.2 NM_001143773.1 NM_001166698.1 NM_198215.3 XM_005269618.4 XM_005269619.4 XM_006717702.3 XM_006717703.3 XM_011539490.2 XM_011539491.2 XM_017015885.1 XM_017015886.1 XM_017015887.1 XM_017015888.1 XM_017015889.1 XM_017015890.1 XM_017015891.1 XM_017015892.1 XM_017015893.1 NM_001143774.1 NM_032439.3 XM_011540275.2 XM_011540276.2 XM_017016782.1 XM_017016783.1

SNP Detail

SNP ID Chromosome Location
rs1005893432 Chr.10: 59245726-59245726 on Build GRCh38
rs183135127 Chr.10: 59245965-59245965 on Build GRCh38
rs746131083 Chr.10: 59245817-59245817 on Build GRCh38
rs190534634 Chr.10: 59245961-59245961 on Build GRCh38
rs914385822 Chr.10: 59245771-59245771 on Build GRCh38
rs988679828 Chr.10: 59245992-59245992 on Build GRCh38
rs150759001 Chr.10: 59245785-59245785 on Build GRCh38
rs979414363 Chr.10: 59245871-59245871 on Build GRCh38
rs918254572 Chr.10: 59246194-59246194 on Build GRCh38
rs905553691 Chr.10: 59246058-59246058 on Build GRCh38
rs886071774 Chr.10: 59245722-59245722 on Build GRCh38
rs1002859214 Chr.10: 59246124-59246124 on Build GRCh38
rs139104646 Chr.10: 59245803-59245803 on Build GRCh38
rs914418168 Chr.10: 59246000-59246000 on Build GRCh38
rs997673472 Chr.10: 59245717-59245717 on Build GRCh38
rs115773882 Chr.10: 59245742-59245742 on Build GRCh38
rs575356985 Chr.10: 59246005-59246005 on Build GRCh38
rs1017177571 Chr.10: 59245734-59245734 on Build GRCh38
rs896769778 Chr.10: 59245826-59245826 on Build GRCh38
rs76341267 Chr.10: 59245856-59245856 on Build GRCh38
rs11006415 Chr.10: 59246020-59246020 on Build GRCh38
rs74794973 Chr.10: 59245795-59245795 on Build GRCh38
rs562128617 Chr.10: 59245805-59245809 on Build GRCh38
rs571403332 Chr.10: 59245893-59245893 on Build GRCh38
rs529595477 Chr.10: 59245720-59245720 on Build GRCh38
rs979893468 Chr.10: 59246060-59246060 on Build GRCh38
rs556755851 Chr.10: 59246003-59246003 on Build GRCh38
rs772540774 Chr.10: 59245900-59245900 on Build GRCh38
rs993731988 Chr.10: 59245829-59245830 on Build GRCh38
rs941651208 Chr.10: 59246040-59246040 on Build GRCh38
rs933176532 Chr.10: 59245984-59245984 on Build GRCh38
rs555329704 Chr.10: 59245935-59245935 on Build GRCh38
rs938398706 Chr.10: 59246129-59246129 on Build GRCh38
rs997446002 Chr.10: 59245964-59245964 on Build GRCh38
rs534242487 Chr.10: 59245890-59245890 on Build GRCh38
rs536754503 Chr.10: 59246159-59246161 on Build GRCh38
rs1033942223 Chr.10: 59246136-59246136 on Build GRCh38
rs1037359529 Chr.10: 59246054-59246054 on Build GRCh38
rs778996468 Chr.10: 59245768-59245768 on Build GRCh38
rs747409173 Chr.10: 59246123-59246123 on Build GRCh38
rs1044504068 Chr.10: 59245791-59245791 on Build GRCh38
rs943270875 Chr.10: 59245917-59245917 on Build GRCh38
rs1018068825 Chr.10: 59245898-59245898 on Build GRCh38
rs886210483 Chr.10: 59246009-59246009 on Build GRCh38
rs774459143 Chr.10: 59245806-59245807 on Build GRCh38
rs764946083 Chr.10: 59246190-59246191 on Build GRCh38
rs539131572 Chr.10: 59245710-59245710 on Build GRCh38
rs753132690 Chr.10: 59245865-59245865 on Build GRCh38
rs1047896185 Chr.10: 59245991-59245991 on Build GRCh38
rs751143064 Chr.10: 59245748-59245748 on Build GRCh38
rs913989259 Chr.10: 59245838-59245838 on Build GRCh38

View Full Product Details