Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 96058475-96059027 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNJ
Entrez Gene ID
54619
Gene Name
cyclin J
Gene Aliases
bA690P14.1
UniGene
Hs.596479
Gene Chromosome Location
Chr.10: 96042848-96060870 on Build GRCh38
Transcripts
NM_001134375.1 NM_001134376.1 NM_019084.4 XM_005269921.3 XM_011539885.2 XM_017016354.1 XM_017016355.1 XM_017016356.1 NR_038444.1 NR_134320.1 NR_134321.1 NR_134322.1

Gene Symbol
ENTPD1-AS1
Entrez Gene ID
728558
Gene Name
ENTPD1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.538374
Gene Chromosome Location
Chr.10: 95753206-96090235 on Build GRCh38
Transcripts
NM_001134375.1 NM_001134376.1 NM_019084.4 XM_005269921.3 XM_011539885.2 XM_017016354.1 XM_017016355.1 XM_017016356.1 NR_038444.1 NR_134320.1 NR_134321.1 NR_134322.1

SNP Detail

SNP ID Chromosome Location
rs528590317 Chr.10: 96058991-96058991 on Build GRCh38
rs41291599 Chr.10: 96058787-96058787 on Build GRCh38
rs184086711 Chr.10: 96058824-96058824 on Build GRCh38
rs942004572 Chr.10: 96058735-96058736 on Build GRCh38
rs566081131 Chr.10: 96058502-96058502 on Build GRCh38
rs552605849 Chr.10: 96058734-96058734 on Build GRCh38
rs909228319 Chr.10: 96058712-96058712 on Build GRCh38
rs1021313451 Chr.10: 96058559-96058559 on Build GRCh38
rs867163285 Chr.10: 96058981-96058981 on Build GRCh38
rs541465787 Chr.10: 96058785-96058785 on Build GRCh38
rs185581339 Chr.10: 96058627-96058627 on Build GRCh38
rs145513285 Chr.10: 96058814-96058814 on Build GRCh38
rs12244026 Chr.10: 96058786-96058786 on Build GRCh38
rs568389771 Chr.10: 96058589-96058589 on Build GRCh38
rs951262045 Chr.10: 96058826-96058826 on Build GRCh38
rs538974494 Chr.10: 96058976-96058976 on Build GRCh38
rs909766354 Chr.10: 96058860-96058860 on Build GRCh38
rs927286508 Chr.10: 96058930-96058930 on Build GRCh38
rs758585452 Chr.10: 96058674-96058674 on Build GRCh38
rs10882713 Chr.10: 96058935-96058935 on Build GRCh38
rs146512667 Chr.10: 96058657-96058657 on Build GRCh38
rs181068005 Chr.10: 96058721-96058721 on Build GRCh38
rs558808350 Chr.10: 96058998-96058998 on Build GRCh38
rs866669750 Chr.10: 96058532-96058532 on Build GRCh38
rs138726081 Chr.10: 96058772-96058773 on Build GRCh38
rs984063905 Chr.10: 96058847-96058847 on Build GRCh38
rs905997599 Chr.10: 96058748-96058748 on Build GRCh38
rs139471024 Chr.10: 96058701-96058701 on Build GRCh38
rs75943857 Chr.10: 96058927-96058927 on Build GRCh38
rs929934010 Chr.10: 96058696-96058696 on Build GRCh38
rs985218452 Chr.10: 96058603-96058603 on Build GRCh38
rs569165448 Chr.10: 96058936-96058936 on Build GRCh38
rs781720596 Chr.10: 96058946-96058946 on Build GRCh38
rs760746003 Chr.10: 96058751-96058752 on Build GRCh38
rs965210496 Chr.10: 96058630-96058630 on Build GRCh38
rs186797953 Chr.10: 96058764-96058764 on Build GRCh38
rs1044465258 Chr.10: 96058737-96058737 on Build GRCh38
rs11596126 Chr.10: 96058583-96058583 on Build GRCh38
rs1017949876 Chr.10: 96058813-96058813 on Build GRCh38
rs180781911 Chr.10: 96058619-96058619 on Build GRCh38
rs780219375 Chr.10: 96058692-96058692 on Build GRCh38
rs189322798 Chr.10: 96058560-96058560 on Build GRCh38
rs188627904 Chr.10: 96058708-96058708 on Build GRCh38
rs765608649 Chr.10: 96058540-96058540 on Build GRCh38
rs1010890545 Chr.10: 96058548-96058548 on Build GRCh38
rs976620313 Chr.10: 96058656-96058656 on Build GRCh38
rs892083172 Chr.10: 96058522-96058523 on Build GRCh38
rs772778424 Chr.10: 96058911-96058911 on Build GRCh38
rs1057456262 Chr.10: 96058990-96058990 on Build GRCh38
rs1057327625 Chr.10: 96058778-96058779 on Build GRCh38
rs969275365 Chr.10: 96058919-96058919 on Build GRCh38
rs536925007 Chr.10: 96058606-96058606 on Build GRCh38
rs559924177 Chr.10: 96058986-96058986 on Build GRCh38
rs904196825 Chr.10: 96058793-96058793 on Build GRCh38
rs998325963 Chr.10: 96058820-96058820 on Build GRCh38
rs543744426 Chr.10: 96058810-96058810 on Build GRCh38
rs187365465 Chr.10: 96058993-96058993 on Build GRCh38

View Full Product Details