Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1517034-1517630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD3A
Entrez Gene ID
55210
Gene Name
ATPase family, AAA domain containing 3A
Gene Aliases
-
UniGene
Hs.23413
Gene Chromosome Location
Chr.1: 1512143-1534687 on Build GRCh38
Transcripts
NM_001170535.1 NM_001170536.1 NM_018188.3 XM_017001628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3746136 c.302G>A p.S101N Chr.1: 1517186-1517186 on Build GRCh38
3976042 c.328T>C p.W110R Chr.1: 1517212-1517212 on Build GRCh38
3976043 c.404C>A p.A135D Chr.1: 1517288-1517288 on Build GRCh38
5816403 c.473C>T p.A158V Chr.1: 1517357-1517357 on Build GRCh38
6226737 c.345C>T p.A115A Chr.1: 1517229-1517229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009306591 Chr.1: 1517290-1517290 on Build GRCh38
rs759611921 Chr.1: 1517357-1517357 on Build GRCh38
rs959613370 Chr.1: 1517408-1517408 on Build GRCh38
rs1007370767 Chr.1: 1517068-1517068 on Build GRCh38
rs111688484 Chr.1: 1517107-1517108 on Build GRCh38
rs780466539 Chr.1: 1517266-1517266 on Build GRCh38
rs867770365 Chr.1: 1517187-1517187 on Build GRCh38
rs992326678 Chr.1: 1517410-1517410 on Build GRCh38
rs751053415 Chr.1: 1517229-1517229 on Build GRCh38
rs540993192 Chr.1: 1517442-1517442 on Build GRCh38
rs1037029795 Chr.1: 1517156-1517156 on Build GRCh38
rs751157483 Chr.1: 1517421-1517421 on Build GRCh38
rs564375426 Chr.1: 1517452-1517452 on Build GRCh38
rs1027518217 Chr.1: 1517063-1517063 on Build GRCh38
rs1040259376 Chr.1: 1517537-1517537 on Build GRCh38
rs749932655 Chr.1: 1517433-1517433 on Build GRCh38
rs571690031 Chr.1: 1517388-1517388 on Build GRCh38
rs995157805 Chr.1: 1517059-1517059 on Build GRCh38
rs918005233 Chr.1: 1517144-1517144 on Build GRCh38
rs563771489 Chr.1: 1517101-1517101 on Build GRCh38
rs777248994 Chr.1: 1517078-1517078 on Build GRCh38
rs994789843 Chr.1: 1517173-1517173 on Build GRCh38
rs1018785302 Chr.1: 1517234-1517234 on Build GRCh38
rs926870459 Chr.1: 1517111-1517111 on Build GRCh38
rs575685262 Chr.1: 1517330-1517330 on Build GRCh38
rs529316879 Chr.1: 1517117-1517117 on Build GRCh38
rs773148798 Chr.1: 1517354-1517354 on Build GRCh38
rs765178275 Chr.1: 1517206-1517206 on Build GRCh38
rs139994378 Chr.1: 1517261-1517261 on Build GRCh38
rs756601598 Chr.1: 1517263-1517263 on Build GRCh38
rs940108737 Chr.1: 1517153-1517153 on Build GRCh38
rs749624303 Chr.1: 1517277-1517277 on Build GRCh38
rs759415572 Chr.1: 1517197-1517197 on Build GRCh38
rs563635730 Chr.1: 1517496-1517496 on Build GRCh38
rs779012956 Chr.1: 1517456-1517456 on Build GRCh38
rs200230792 Chr.1: 1517326-1517326 on Build GRCh38
rs752534140 Chr.1: 1517209-1517209 on Build GRCh38
rs780519970 Chr.1: 1517443-1517443 on Build GRCh38
rs774338831 Chr.1: 1517140-1517140 on Build GRCh38
rs968355850 Chr.1: 1517100-1517100 on Build GRCh38
rs763685989 Chr.1: 1517412-1517412 on Build GRCh38
rs761245214 Chr.1: 1517431-1517431 on Build GRCh38
rs146731359 Chr.1: 1517323-1517323 on Build GRCh38
rs1020728594 Chr.1: 1517595-1517595 on Build GRCh38
rs2478791 Chr.1: 1517235-1517235 on Build GRCh38
rs1009376984 Chr.1: 1517583-1517583 on Build GRCh38
rs35860586 Chr.1: 1517358-1517358 on Build GRCh38
rs192639249 Chr.1: 1517088-1517088 on Build GRCh38
rs550025982 Chr.1: 1517089-1517089 on Build GRCh38
rs61777881 Chr.1: 1517224-1517224 on Build GRCh38
rs142207140 Chr.1: 1517494-1517494 on Build GRCh38
rs931320761 Chr.1: 1517446-1517446 on Build GRCh38
rs145736326 Chr.1: 1517343-1517343 on Build GRCh38
rs762786838 Chr.1: 1517399-1517399 on Build GRCh38
rs764758709 Chr.1: 1517170-1517170 on Build GRCh38
rs528473251 Chr.1: 1517600-1517600 on Build GRCh38
rs529457375 Chr.1: 1517530-1517530 on Build GRCh38
rs926874302 Chr.1: 1517396-1517396 on Build GRCh38
rs1619925 Chr.1: 1517171-1517171 on Build GRCh38
rs577774758 Chr.1: 1517453-1517453 on Build GRCh38
rs1020653590 Chr.1: 1517308-1517308 on Build GRCh38
rs889069966 Chr.1: 1517495-1517495 on Build GRCh38
rs551809867 Chr.1: 1517607-1517607 on Build GRCh38
rs901657167 Chr.1: 1517549-1517549 on Build GRCh38
rs559821283 Chr.1: 1517599-1517599 on Build GRCh38
rs549490670 Chr.1: 1517587-1517587 on Build GRCh38
rs1018719344 Chr.1: 1517076-1517076 on Build GRCh38
rs1619815 Chr.1: 1517212-1517212 on Build GRCh38
rs113256690 Chr.1: 1517102-1517102 on Build GRCh38
rs781555961 Chr.1: 1517246-1517246 on Build GRCh38
rs1619896 Chr.1: 1517186-1517186 on Build GRCh38
rs897788256 Chr.1: 1517058-1517058 on Build GRCh38
rs767115233 Chr.1: 1517432-1517432 on Build GRCh38
rs761933938 Chr.1: 1517164-1517164 on Build GRCh38

View Full Product Details