Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 93355075-93355656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEUP1
Entrez Gene ID
159989
Gene Name
deuterosome assembly protein 1
Gene Aliases
CCDC67
UniGene
Hs.436625
Gene Chromosome Location
Chr.11: 93329079-93438470 on Build GRCh38
Transcripts
NM_181645.3 XM_005273802.1 XM_011542629.2 XM_011542630.1 XM_011542631.2 XM_011542632.2 XM_011542633.2 XM_011542635.2 XM_011542637.2 XM_017017245.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
933326 c.199G>C p.E67Q Chr.11: 93355540-93355540 on Build GRCh38
3454668 c.196C>T p.Q66* Chr.11: 93355537-93355537 on Build GRCh38
7359978 c.187C>A p.Q63K Chr.11: 93355528-93355528 on Build GRCh38
3454669 c.198A>T p.Q66H Chr.11: 93355539-93355539 on Build GRCh38
4037648 c.73A>G p.I25V Chr.11: 93355414-93355414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558510965 Chr.11: 93355277-93355277 on Build GRCh38
rs773985601 Chr.11: 93355321-93355321 on Build GRCh38
rs774202838 Chr.11: 93355429-93355429 on Build GRCh38
rs771992332 Chr.11: 93355577-93355577 on Build GRCh38
rs775320691 Chr.11: 93355498-93355498 on Build GRCh38
rs546997322 Chr.11: 93355112-93355112 on Build GRCh38
rs928400017 Chr.11: 93355520-93355520 on Build GRCh38
rs752469381 Chr.11: 93355207-93355207 on Build GRCh38
rs373323890 Chr.11: 93355326-93355326 on Build GRCh38
rs190325389 Chr.11: 93355109-93355109 on Build GRCh38
rs749812095 Chr.11: 93355553-93355553 on Build GRCh38
rs3019224 Chr.11: 93355153-93355153 on Build GRCh38
rs764499863 Chr.11: 93355464-93355464 on Build GRCh38
rs554716737 Chr.11: 93355208-93355208 on Build GRCh38
rs201641405 Chr.11: 93355358-93355358 on Build GRCh38
rs375780576 Chr.11: 93355489-93355489 on Build GRCh38
rs532159512 Chr.11: 93355560-93355560 on Build GRCh38
rs761532182 Chr.11: 93355446-93355446 on Build GRCh38
rs998191785 Chr.11: 93355239-93355239 on Build GRCh38
rs571303827 Chr.11: 93355117-93355117 on Build GRCh38
rs200134824 Chr.11: 93355546-93355546 on Build GRCh38
rs759460335 Chr.11: 93355102-93355102 on Build GRCh38
rs374648091 Chr.11: 93355477-93355477 on Build GRCh38
rs200488619 Chr.11: 93355462-93355462 on Build GRCh38
rs147207855 Chr.11: 93355212-93355212 on Build GRCh38
rs771869454 Chr.11: 93355512-93355512 on Build GRCh38
rs781219200 Chr.11: 93355380-93355380 on Build GRCh38
rs757377379 Chr.11: 93355376-93355376 on Build GRCh38
rs897971333 Chr.11: 93355282-93355282 on Build GRCh38
rs569038649 Chr.11: 93355164-93355164 on Build GRCh38
rs1045457938 Chr.11: 93355142-93355142 on Build GRCh38
rs957289196 Chr.11: 93355401-93355401 on Build GRCh38
rs201030952 Chr.11: 93355366-93355366 on Build GRCh38
rs778942261 Chr.11: 93355568-93355568 on Build GRCh38
rs758725297 Chr.11: 93355362-93355362 on Build GRCh38
rs750514851 Chr.11: 93355480-93355480 on Build GRCh38
rs769955257 Chr.11: 93355376-93355378 on Build GRCh38
rs775912242 Chr.11: 93355528-93355528 on Build GRCh38
rs765730078 Chr.11: 93355356-93355356 on Build GRCh38
rs751944109 Chr.11: 93355540-93355540 on Build GRCh38
rs557314407 Chr.11: 93355163-93355163 on Build GRCh38
rs1036505201 Chr.11: 93355575-93355575 on Build GRCh38
rs532880533 Chr.11: 93355337-93355337 on Build GRCh38
rs775439544 Chr.11: 93355393-93355393 on Build GRCh38
rs1006418794 Chr.11: 93355209-93355209 on Build GRCh38
rs376502642 Chr.11: 93355415-93355415 on Build GRCh38
rs753756340 Chr.11: 93355554-93355554 on Build GRCh38
rs762993490 Chr.11: 93355507-93355508 on Build GRCh38
rs376905531 Chr.11: 93355484-93355484 on Build GRCh38
rs747221324 Chr.11: 93355370-93355370 on Build GRCh38
rs540410474 Chr.11: 93355217-93355217 on Build GRCh38
rs565421090 Chr.11: 93355184-93355184 on Build GRCh38
rs773396213 Chr.11: 93355456-93355457 on Build GRCh38
rs942278196 Chr.11: 93355593-93355593 on Build GRCh38
rs760209195 Chr.11: 93355457-93355457 on Build GRCh38
rs924457929 Chr.11: 93355378-93355378 on Build GRCh38
rs191542819 Chr.11: 93355547-93355547 on Build GRCh38
rs768491574 Chr.11: 93355422-93355422 on Build GRCh38
rs758750223 Chr.11: 93355463-93355463 on Build GRCh38
rs762107519 Chr.11: 93355542-93355542 on Build GRCh38
rs769641235 Chr.11: 93355389-93355389 on Build GRCh38
rs1036586369 Chr.11: 93355604-93355604 on Build GRCh38
rs766618413 Chr.11: 93355544-93355545 on Build GRCh38
rs898055700 Chr.11: 93355224-93355224 on Build GRCh38
rs757620865 Chr.11: 93355164-93355165 on Build GRCh38
rs770464650 Chr.11: 93355522-93355522 on Build GRCh38
rs780597533 Chr.11: 93355501-93355501 on Build GRCh38
rs201790053 Chr.11: 93355449-93355449 on Build GRCh38
rs78902781 Chr.11: 93355341-93355341 on Build GRCh38
rs747964984 Chr.11: 93355569-93355569 on Build GRCh38
rs370944951 Chr.11: 93355592-93355592 on Build GRCh38
rs968942111 Chr.11: 93355360-93355360 on Build GRCh38
rs1045166386 Chr.11: 93355195-93355195 on Build GRCh38
rs201277348 Chr.11: 93355490-93355490 on Build GRCh38
rs550851882 Chr.11: 93355610-93355610 on Build GRCh38
rs763486417 Chr.11: 93355537-93355537 on Build GRCh38
rs751734785 Chr.11: 93355556-93355576 on Build GRCh38
rs757502099 Chr.11: 93355478-93355478 on Build GRCh38
rs745908412 Chr.11: 93355384-93355384 on Build GRCh38
rs530193317 Chr.11: 93355347-93355347 on Build GRCh38
rs945819976 Chr.11: 93355131-93355131 on Build GRCh38
rs764469678 Chr.11: 93355539-93355539 on Build GRCh38
rs772875843 Chr.11: 93355450-93355450 on Build GRCh38
rs760172818 Chr.11: 93355515-93355515 on Build GRCh38
rs780157573 Chr.11: 93355548-93355548 on Build GRCh38

View Full Product Details