Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 75800232-75800831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGAT2
Entrez Gene ID
84649
Gene Name
diacylglycerol O-acyltransferase 2
Gene Aliases
ARAT, GS1999FULL, HMFN1045
UniGene
Hs.129798
Gene Chromosome Location
Chr.11: 75768733-75801536 on Build GRCh38
Transcripts
NM_001253891.1 NM_032564.4 XM_011545304.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5580806 c.1065C>T p.I355I Chr.11: 75800406-75800406 on Build GRCh38
196235 c.1110C>T p.F370F Chr.11: 75800451-75800451 on Build GRCh38
4677145 c.1134C>T p.G378G Chr.11: 75800475-75800475 on Build GRCh38
1203426 c.1066G>A p.D356N Chr.11: 75800407-75800407 on Build GRCh38
244018 c.1131C>T p.F377F Chr.11: 75800472-75800472 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767963487 Chr.11: 75800499-75800499 on Build GRCh38
rs994388301 Chr.11: 75800703-75800703 on Build GRCh38
rs371424792 Chr.11: 75800704-75800704 on Build GRCh38
rs144285130 Chr.11: 75800458-75800458 on Build GRCh38
rs750873196 Chr.11: 75800439-75800439 on Build GRCh38
rs186425264 Chr.11: 75800664-75800664 on Build GRCh38
rs1028100523 Chr.11: 75800636-75800636 on Build GRCh38
rs148751558 Chr.11: 75800472-75800472 on Build GRCh38
rs962925099 Chr.11: 75800409-75800409 on Build GRCh38
rs777669272 Chr.11: 75800463-75800463 on Build GRCh38
rs752890961 Chr.11: 75800526-75800526 on Build GRCh38
rs539339044 Chr.11: 75800514-75800514 on Build GRCh38
rs138423103 Chr.11: 75800375-75800375 on Build GRCh38
rs770747050 Chr.11: 75800334-75800334 on Build GRCh38
rs781762950 Chr.11: 75800310-75800310 on Build GRCh38
rs773585309 Chr.11: 75800504-75800504 on Build GRCh38
rs745315498 Chr.11: 75800480-75800480 on Build GRCh38
rs891910911 Chr.11: 75800600-75800600 on Build GRCh38
rs745775539 Chr.11: 75800378-75800378 on Build GRCh38
rs973279966 Chr.11: 75800420-75800420 on Build GRCh38
rs749431184 Chr.11: 75800342-75800342 on Build GRCh38
rs373549645 Chr.11: 75800518-75800518 on Build GRCh38
rs558835232 Chr.11: 75800519-75800519 on Build GRCh38
rs916979420 Chr.11: 75800797-75800797 on Build GRCh38
rs775270613 Chr.11: 75800487-75800487 on Build GRCh38
rs1017150946 Chr.11: 75800666-75800666 on Build GRCh38
rs565074034 Chr.11: 75800799-75800799 on Build GRCh38
rs1029608416 Chr.11: 75800369-75800369 on Build GRCh38
rs1006819027 Chr.11: 75800652-75800652 on Build GRCh38
rs948610885 Chr.11: 75800278-75800278 on Build GRCh38
rs753905329 Chr.11: 75800451-75800451 on Build GRCh38
rs376849632 Chr.11: 75800328-75800328 on Build GRCh38
rs200668858 Chr.11: 75800352-75800352 on Build GRCh38
rs112477468 Chr.11: 75800276-75800276 on Build GRCh38
rs769841777 Chr.11: 75800481-75800481 on Build GRCh38
rs772958795 Chr.11: 75800383-75800383 on Build GRCh38
rs369491336 Chr.11: 75800346-75800346 on Build GRCh38
rs912523891 Chr.11: 75800320-75800320 on Build GRCh38
rs768756525 Chr.11: 75800406-75800406 on Build GRCh38
rs376872494 Chr.11: 75800522-75800522 on Build GRCh38
rs973852283 Chr.11: 75800260-75800260 on Build GRCh38
rs776449399 Chr.11: 75800478-75800478 on Build GRCh38
rs371249178 Chr.11: 75800761-75800763 on Build GRCh38
rs974208784 Chr.11: 75800582-75800582 on Build GRCh38
rs575023953 Chr.11: 75800616-75800616 on Build GRCh38
rs752405049 Chr.11: 75800419-75800419 on Build GRCh38
rs1000407142 Chr.11: 75800297-75800297 on Build GRCh38
rs780204650 Chr.11: 75800335-75800335 on Build GRCh38
rs950525818 Chr.11: 75800739-75800739 on Build GRCh38
rs777883845 Chr.11: 75800530-75800530 on Build GRCh38
rs762743938 Chr.11: 75800494-75800494 on Build GRCh38
rs932334534 Chr.11: 75800290-75800290 on Build GRCh38
rs760720423 Chr.11: 75800389-75800389 on Build GRCh38
rs555027391 Chr.11: 75800585-75800585 on Build GRCh38
rs537539523 Chr.11: 75800407-75800407 on Build GRCh38
rs544529156 Chr.11: 75800554-75800554 on Build GRCh38
rs763281362 Chr.11: 75800768-75800768 on Build GRCh38
rs3060 Chr.11: 75800527-75800527 on Build GRCh38
rs756684772 Chr.11: 75800440-75800440 on Build GRCh38
rs998661505 Chr.11: 75800608-75800608 on Build GRCh38
rs1031763032 Chr.11: 75800309-75800309 on Build GRCh38
rs770857243 Chr.11: 75800473-75800473 on Build GRCh38
rs1007075989 Chr.11: 75800386-75800386 on Build GRCh38
rs539516097 Chr.11: 75800707-75800707 on Build GRCh38
rs953774083 Chr.11: 75800641-75800641 on Build GRCh38
rs1044281002 Chr.11: 75800280-75800280 on Build GRCh38
rs992560830 Chr.11: 75800759-75800759 on Build GRCh38
rs374192362 Chr.11: 75800418-75800418 on Build GRCh38
rs757009480 Chr.11: 75800552-75800552 on Build GRCh38
rs746763799 Chr.11: 75800534-75800534 on Build GRCh38
rs545297460 Chr.11: 75800728-75800728 on Build GRCh38
rs969718308 Chr.11: 75800804-75800804 on Build GRCh38
rs770321218 Chr.11: 75800399-75800399 on Build GRCh38
rs889800543 Chr.11: 75800625-75800625 on Build GRCh38
rs1053359570 Chr.11: 75800577-75800577 on Build GRCh38
rs986499621 Chr.11: 75800648-75800649 on Build GRCh38
rs758172784 Chr.11: 75800462-75800462 on Build GRCh38
rs796250734 Chr.11: 75800756-75800758 on Build GRCh38
rs145420090 Chr.11: 75800291-75800291 on Build GRCh38
rs34113941 Chr.11: 75800424-75800424 on Build GRCh38
rs371445822 Chr.11: 75800452-75800452 on Build GRCh38
rs866936335 Chr.11: 75800698-75800698 on Build GRCh38
rs750942390 Chr.11: 75800315-75800315 on Build GRCh38
rs966740757 Chr.11: 75800653-75800653 on Build GRCh38
rs377676292 Chr.11: 75800428-75800428 on Build GRCh38

View Full Product Details