Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 126240665-126241235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM118B
Entrez Gene ID
79607
Gene Name
family with sequence similarity 118 member B
Gene Aliases
-
UniGene
Hs.276315
Gene Chromosome Location
Chr.11: 126211414-126264540 on Build GRCh38
Transcripts
NM_024556.3 XM_005271665.3 XM_011542977.2 XM_011542978.2 XM_017018285.1 XM_017018286.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7269464 c.182C>A p.A61D Chr.11: 126240887-126240887 on Build GRCh38
4145477 c.138G>T p.V46V Chr.11: 126240843-126240843 on Build GRCh38
6566748 c.87-5T>C p.? Chr.11: 126240787-126240787 on Build GRCh38
1638775 c.162A>G p.A54A Chr.11: 126240867-126240867 on Build GRCh38
4019097 c.231T>C p.I77I Chr.11: 126240936-126240936 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764392410 Chr.11: 126241043-126241043 on Build GRCh38
rs569014741 Chr.11: 126241130-126241139 on Build GRCh38
rs190681425 Chr.11: 126241109-126241109 on Build GRCh38
rs776429607 Chr.11: 126240996-126240996 on Build GRCh38
rs935789063 Chr.11: 126240865-126240865 on Build GRCh38
rs767061988 Chr.11: 126240901-126240901 on Build GRCh38
rs534378676 Chr.11: 126241208-126241208 on Build GRCh38
rs749776651 Chr.11: 126240933-126240933 on Build GRCh38
rs112508226 Chr.11: 126240738-126240738 on Build GRCh38
rs775252070 Chr.11: 126240838-126240838 on Build GRCh38
rs761741577 Chr.11: 126240873-126240873 on Build GRCh38
rs1049517253 Chr.11: 126241202-126241202 on Build GRCh38
rs759658414 Chr.11: 126241034-126241034 on Build GRCh38
rs370421247 Chr.11: 126240855-126240855 on Build GRCh38
rs138037012 Chr.11: 126241153-126241153 on Build GRCh38
rs771423560 Chr.11: 126240935-126240935 on Build GRCh38
rs766459848 Chr.11: 126240878-126240878 on Build GRCh38
rs373486807 Chr.11: 126240803-126240803 on Build GRCh38
rs146965322 Chr.11: 126241093-126241093 on Build GRCh38
rs753783931 Chr.11: 126240872-126240872 on Build GRCh38
rs529273134 Chr.11: 126240800-126240800 on Build GRCh38
rs768338635 Chr.11: 126240983-126240983 on Build GRCh38
rs141532978 Chr.11: 126241146-126241147 on Build GRCh38
rs912977861 Chr.11: 126241175-126241175 on Build GRCh38
rs775342326 Chr.11: 126240968-126240968 on Build GRCh38
rs372155646 Chr.11: 126240717-126240717 on Build GRCh38
rs925665228 Chr.11: 126240860-126240860 on Build GRCh38
rs763980731 Chr.11: 126240741-126240741 on Build GRCh38
rs998531476 Chr.11: 126241156-126241156 on Build GRCh38
rs756505078 Chr.11: 126240920-126240920 on Build GRCh38
rs758464482 Chr.11: 126240752-126240752 on Build GRCh38
rs773098505 Chr.11: 126241008-126241008 on Build GRCh38
rs1050845242 Chr.11: 126241134-126241134 on Build GRCh38
rs932409947 Chr.11: 126241125-126241125 on Build GRCh38
rs377443610 Chr.11: 126240826-126240826 on Build GRCh38
rs559821858 Chr.11: 126240743-126240743 on Build GRCh38
rs760438438 Chr.11: 126240846-126240846 on Build GRCh38
rs867630157 Chr.11: 126241009-126241009 on Build GRCh38
rs1052682388 Chr.11: 126241165-126241165 on Build GRCh38
rs377606430 Chr.11: 126241002-126241002 on Build GRCh38
rs866683150 Chr.11: 126240992-126240992 on Build GRCh38
rs748012486 Chr.11: 126241096-126241096 on Build GRCh38
rs140543424 Chr.11: 126241061-126241061 on Build GRCh38
rs1011035671 Chr.11: 126240982-126240982 on Build GRCh38
rs921825160 Chr.11: 126241198-126241198 on Build GRCh38
rs944604118 Chr.11: 126240885-126240885 on Build GRCh38
rs199692306 Chr.11: 126240963-126240963 on Build GRCh38
rs749801351 Chr.11: 126240808-126240808 on Build GRCh38
rs771687839 Chr.11: 126240967-126240967 on Build GRCh38
rs757666351 Chr.11: 126241058-126241058 on Build GRCh38
rs755134067 Chr.11: 126240766-126240766 on Build GRCh38
rs899585317 Chr.11: 126240862-126240862 on Build GRCh38
rs760836255 Chr.11: 126241164-126241166 on Build GRCh38
rs145900366 Chr.11: 126240905-126240905 on Build GRCh38
rs774642462 Chr.11: 126241091-126241091 on Build GRCh38
rs927959173 Chr.11: 126240719-126240719 on Build GRCh38
rs10893489 Chr.11: 126241199-126241199 on Build GRCh38
rs186999900 Chr.11: 126240972-126240972 on Build GRCh38
rs377121357 Chr.11: 126240716-126240719 on Build GRCh38
rs761676919 Chr.11: 126240997-126240997 on Build GRCh38
rs657361 Chr.11: 126241196-126241196 on Build GRCh38
rs74892607 Chr.11: 126240918-126240918 on Build GRCh38
rs748468756 Chr.11: 126240780-126240780 on Build GRCh38
rs759288635 Chr.11: 126240827-126240827 on Build GRCh38
rs767568136 Chr.11: 126241035-126241035 on Build GRCh38
rs778136288 Chr.11: 126240928-126240928 on Build GRCh38
rs3824924 Chr.11: 126241083-126241083 on Build GRCh38
rs771810520 Chr.11: 126240832-126240832 on Build GRCh38
rs927980069 Chr.11: 126241060-126241060 on Build GRCh38
rs965837055 Chr.11: 126241190-126241190 on Build GRCh38
rs890812770 Chr.11: 126241136-126241136 on Build GRCh38
rs913099245 Chr.11: 126240884-126240884 on Build GRCh38
rs12280636 Chr.11: 126241038-126241038 on Build GRCh38
rs1046957093 Chr.11: 126240731-126240731 on Build GRCh38
rs747219132 Chr.11: 126241176-126241176 on Build GRCh38
rs982731726 Chr.11: 126241072-126241072 on Build GRCh38
rs530676295 Chr.11: 126240961-126240961 on Build GRCh38
rs368165315 Chr.11: 126241189-126241190 on Build GRCh38
rs746652521 Chr.11: 126241088-126241088 on Build GRCh38
rs751784789 Chr.11: 126240765-126240765 on Build GRCh38
rs182479791 Chr.11: 126240692-126240692 on Build GRCh38
rs939284884 Chr.11: 126240725-126240725 on Build GRCh38
rs768829135 Chr.11: 126240788-126240788 on Build GRCh38
rs746366577 Chr.11: 126241067-126241067 on Build GRCh38
rs138636455 Chr.11: 126240906-126240906 on Build GRCh38
rs773415519 Chr.11: 126241140-126241140 on Build GRCh38
rs866625269 Chr.11: 126240912-126240912 on Build GRCh38
rs758050584 Chr.11: 126241068-126241068 on Build GRCh38
rs544959184 Chr.11: 126240822-126240822 on Build GRCh38
rs1038165515 Chr.11: 126240824-126240824 on Build GRCh38
rs879436394 Chr.11: 126240919-126240919 on Build GRCh38
rs760220899 Chr.11: 126240967-126240967 on Build GRCh38
rs941165829 Chr.11: 126240805-126240805 on Build GRCh38
rs755219950 Chr.11: 126240916-126240916 on Build GRCh38
rs34620920 Chr.11: 126241145-126241146 on Build GRCh38
rs752090984 Chr.11: 126240947-126240948 on Build GRCh38
rs781555681 Chr.11: 126240773-126240773 on Build GRCh38
rs551729363 Chr.11: 126240753-126240753 on Build GRCh38
rs956911213 Chr.11: 126240980-126240980 on Build GRCh38
rs376772264 Chr.11: 126241055-126241055 on Build GRCh38

View Full Product Details