Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4331019-4331576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIGAR
Entrez Gene ID
57103
Gene Name
TP53 induced glycolysis regulatory phosphatase
Gene Aliases
C12orf5, FR2BP
UniGene
Hs.504545
Gene Chromosome Location
Chr.12: 4321193-4360028 on Build GRCh38
Transcripts
NM_020375.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7192051 c.57G>C p.E19D Chr.12: 4331304-4331304 on Build GRCh38
939385 c.44G>T p.R15I Chr.12: 4331291-4331291 on Build GRCh38
3812030 c.55G>C p.E19Q Chr.12: 4331302-4331302 on Build GRCh38
7119867 c.35G>A p.G12E Chr.12: 4331282-4331282 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs955310840 Chr.12: 4331080-4331080 on Build GRCh38
rs188805211 Chr.12: 4331470-4331470 on Build GRCh38
rs1029113771 Chr.12: 4331519-4331519 on Build GRCh38
rs766696947 Chr.12: 4331247-4331247 on Build GRCh38
rs890946310 Chr.12: 4331399-4331399 on Build GRCh38
rs1048805426 Chr.12: 4331307-4331307 on Build GRCh38
rs1015141964 Chr.12: 4331107-4331107 on Build GRCh38
rs1031978768 Chr.12: 4331472-4331472 on Build GRCh38
rs752205590 Chr.12: 4331547-4331550 on Build GRCh38
rs987868812 Chr.12: 4331526-4331526 on Build GRCh38
rs532463208 Chr.12: 4331421-4331421 on Build GRCh38
rs956450010 Chr.12: 4331208-4331208 on Build GRCh38
rs778405492 Chr.12: 4331314-4331314 on Build GRCh38
rs756815595 Chr.12: 4331312-4331312 on Build GRCh38
rs763569691 Chr.12: 4331347-4331347 on Build GRCh38
rs770356393 Chr.12: 4331229-4331229 on Build GRCh38
rs571465979 Chr.12: 4331059-4331059 on Build GRCh38
rs925289528 Chr.12: 4331213-4331213 on Build GRCh38
rs765119931 Chr.12: 4331262-4331264 on Build GRCh38
rs753339113 Chr.12: 4331306-4331306 on Build GRCh38
rs373922309 Chr.12: 4331234-4331234 on Build GRCh38
rs368097902 Chr.12: 4331239-4331239 on Build GRCh38
rs956678280 Chr.12: 4331503-4331503 on Build GRCh38
rs1045325616 Chr.12: 4331248-4331248 on Build GRCh38
rs373660965 Chr.12: 4331256-4331256 on Build GRCh38
rs773915250 Chr.12: 4331335-4331335 on Build GRCh38
rs1008132858 Chr.12: 4331410-4331410 on Build GRCh38
rs1037667519 Chr.12: 4331269-4331269 on Build GRCh38
rs751776153 Chr.12: 4331261-4331261 on Build GRCh38
rs767181008 Chr.12: 4331277-4331277 on Build GRCh38
rs755979055 Chr.12: 4331298-4331298 on Build GRCh38
rs560986981 Chr.12: 4331114-4331114 on Build GRCh38
rs374820242 Chr.12: 4331327-4331327 on Build GRCh38
rs988320180 Chr.12: 4331216-4331216 on Build GRCh38
rs752563495 Chr.12: 4331284-4331284 on Build GRCh38
rs572435365 Chr.12: 4331071-4331071 on Build GRCh38
rs770198603 Chr.12: 4331332-4331332 on Build GRCh38
rs777723795 Chr.12: 4331299-4331299 on Build GRCh38
rs369713521 Chr.12: 4331330-4331330 on Build GRCh38
rs966622248 Chr.12: 4331132-4331132 on Build GRCh38
rs771787283 Chr.12: 4331328-4331328 on Build GRCh38
rs922043788 Chr.12: 4331150-4331150 on Build GRCh38
rs752906442 Chr.12: 4331167-4331167 on Build GRCh38
rs545599745 Chr.12: 4331058-4331058 on Build GRCh38
rs549860409 Chr.12: 4331367-4331367 on Build GRCh38
rs964044780 Chr.12: 4331121-4331121 on Build GRCh38
rs976253469 Chr.12: 4331134-4331134 on Build GRCh38
rs1015610632 Chr.12: 4331391-4331391 on Build GRCh38
rs994408397 Chr.12: 4331057-4331057 on Build GRCh38
rs777724320 Chr.12: 4331499-4331499 on Build GRCh38
rs1000949767 Chr.12: 4331471-4331471 on Build GRCh38
rs979442068 Chr.12: 4331200-4331200 on Build GRCh38
rs563812892 Chr.12: 4331387-4331387 on Build GRCh38
rs953450590 Chr.12: 4331190-4331190 on Build GRCh38
rs1004524617 Chr.12: 4331356-4331356 on Build GRCh38
rs748586734 Chr.12: 4331331-4331331 on Build GRCh38
rs942023122 Chr.12: 4331259-4331259 on Build GRCh38
rs372295977 Chr.12: 4331357-4331357 on Build GRCh38
rs979246875 Chr.12: 4331162-4331162 on Build GRCh38
rs762848857 Chr.12: 4331367-4331368 on Build GRCh38
rs530101263 Chr.12: 4331224-4331224 on Build GRCh38
rs902076361 Chr.12: 4331455-4331455 on Build GRCh38
rs575329198 Chr.12: 4331532-4331532 on Build GRCh38
rs918065072 Chr.12: 4331235-4331235 on Build GRCh38
rs1053459896 Chr.12: 4331220-4331222 on Build GRCh38
rs371312254 Chr.12: 4331273-4331273 on Build GRCh38
rs3935187 Chr.12: 4331104-4331104 on Build GRCh38
rs754200121 Chr.12: 4331051-4331051 on Build GRCh38

View Full Product Details