Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 15620548-15621042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPS8
Entrez Gene ID
2059
Gene Name
epidermal growth factor receptor pathway substrate 8
Gene Aliases
DFNB102
UniGene
Hs.591160
Gene Chromosome Location
Chr.12: 15620141-15789576 on Build GRCh38
Transcripts
NM_004447.5 XM_011520605.2

SNP Detail

SNP ID Chromosome Location
rs565083448 Chr.12: 15620794-15620794 on Build GRCh38
rs1052086922 Chr.12: 15620838-15620838 on Build GRCh38
rs750633346 Chr.12: 15620979-15620979 on Build GRCh38
rs115824520 Chr.12: 15620800-15620800 on Build GRCh38
rs528982035 Chr.12: 15620598-15620598 on Build GRCh38
rs933687890 Chr.12: 15620835-15620835 on Build GRCh38
rs1026428468 Chr.12: 15620611-15620611 on Build GRCh38
rs980400657 Chr.12: 15620722-15620722 on Build GRCh38
rs12320042 Chr.12: 15620605-15620605 on Build GRCh38
rs1010747816 Chr.12: 15620895-15620895 on Build GRCh38
rs181043196 Chr.12: 15620604-15620604 on Build GRCh38
rs1022069154 Chr.12: 15620578-15620578 on Build GRCh38
rs78748444 Chr.12: 15620723-15620723 on Build GRCh38
rs900912103 Chr.12: 15620812-15620812 on Build GRCh38
rs960133656 Chr.12: 15620991-15620991 on Build GRCh38
rs115056526 Chr.12: 15620684-15620684 on Build GRCh38
rs551663997 Chr.12: 15620716-15620716 on Build GRCh38
rs775612141 Chr.12: 15620606-15620606 on Build GRCh38
rs1002038186 Chr.12: 15620942-15620942 on Build GRCh38
rs187056411 Chr.12: 15620978-15620978 on Build GRCh38
rs368751815 Chr.12: 15620641-15620641 on Build GRCh38
rs1014874150 Chr.12: 15620711-15620711 on Build GRCh38
rs989183360 Chr.12: 15620772-15620772 on Build GRCh38
rs376149089 Chr.12: 15620803-15620803 on Build GRCh38
rs905001806 Chr.12: 15620909-15620909 on Build GRCh38
rs1038973370 Chr.12: 15620995-15620995 on Build GRCh38
rs4304842 Chr.12: 15620717-15620717 on Build GRCh38
rs879933488 Chr.12: 15620795-15620795 on Build GRCh38
rs956618143 Chr.12: 15620571-15620571 on Build GRCh38
rs1022088650 Chr.12: 15620908-15620908 on Build GRCh38
rs892410219 Chr.12: 15620888-15620888 on Build GRCh38
rs984302743 Chr.12: 15620726-15620726 on Build GRCh38
rs761372307 Chr.12: 15620943-15620943 on Build GRCh38
rs993268318 Chr.12: 15621002-15621002 on Build GRCh38
rs1051998823 Chr.12: 15620820-15620820 on Build GRCh38
rs1025622113 Chr.12: 15621018-15621018 on Build GRCh38
rs561465953 Chr.12: 15620575-15620575 on Build GRCh38
rs113738793 Chr.12: 15620761-15620761 on Build GRCh38

View Full Product Details