Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8601760-8602357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AICDA
Entrez Gene ID
57379
Gene Name
activation-induced cytidine deaminase
Gene Aliases
AID, ARP2, CDA2, HEL-S-284, HIGM2
UniGene
Hs.149342
Gene Chromosome Location
Chr.12: 8602166-8612970 on Build GRCh38
Transcripts
NM_020661.2 XM_011520772.2

SNP Detail

SNP ID Chromosome Location
rs7316523 Chr.12: 8601957-8601957 on Build GRCh38
rs1053848812 Chr.12: 8602099-8602099 on Build GRCh38
rs928848719 Chr.12: 8601913-8601913 on Build GRCh38
rs1035965661 Chr.12: 8602187-8602188 on Build GRCh38
rs909907983 Chr.12: 8602164-8602164 on Build GRCh38
rs934031291 Chr.12: 8602234-8602234 on Build GRCh38
rs770049488 Chr.12: 8602029-8602029 on Build GRCh38
rs919083533 Chr.12: 8602003-8602003 on Build GRCh38
rs775117772 Chr.12: 8602285-8602290 on Build GRCh38
rs984682891 Chr.12: 8602148-8602148 on Build GRCh38
rs768873221 Chr.12: 8602176-8602176 on Build GRCh38
rs759982293 Chr.12: 8602321-8602321 on Build GRCh38
rs939988268 Chr.12: 8602171-8602171 on Build GRCh38
rs761908534 Chr.12: 8602270-8602270 on Build GRCh38
rs922706831 Chr.12: 8602222-8602222 on Build GRCh38
rs958738535 Chr.12: 8601915-8601915 on Build GRCh38
rs1016660728 Chr.12: 8602286-8602286 on Build GRCh38
rs889925224 Chr.12: 8602282-8602282 on Build GRCh38
rs893577596 Chr.12: 8602155-8602155 on Build GRCh38
rs75945477 Chr.12: 8601847-8601847 on Build GRCh38
rs55706276 Chr.12: 8602065-8602065 on Build GRCh38
rs140108882 Chr.12: 8601833-8601833 on Build GRCh38
rs185931572 Chr.12: 8602189-8602189 on Build GRCh38
rs767335277 Chr.12: 8601903-8601903 on Build GRCh38
rs561892670 Chr.12: 8602060-8602060 on Build GRCh38
rs930506966 Chr.12: 8601789-8601789 on Build GRCh38
rs1003003617 Chr.12: 8602177-8602177 on Build GRCh38
rs951954876 Chr.12: 8602143-8602143 on Build GRCh38
rs755549431 Chr.12: 8601975-8601975 on Build GRCh38
rs183008338 Chr.12: 8602116-8602116 on Build GRCh38
rs991536190 Chr.12: 8602238-8602238 on Build GRCh38
rs370597790 Chr.12: 8601818-8601818 on Build GRCh38
rs1020444120 Chr.12: 8601832-8601832 on Build GRCh38
rs190411066 Chr.12: 8601948-8601948 on Build GRCh38
rs960857761 Chr.12: 8602063-8602063 on Build GRCh38
rs902165900 Chr.12: 8602044-8602044 on Build GRCh38
rs1011081538 Chr.12: 8602015-8602015 on Build GRCh38
rs111462690 Chr.12: 8602308-8602308 on Build GRCh38
rs778955632 Chr.12: 8602114-8602114 on Build GRCh38
rs760666978 Chr.12: 8601855-8601855 on Build GRCh38
rs946990919 Chr.12: 8602304-8602304 on Build GRCh38
rs972129834 Chr.12: 8602312-8602312 on Build GRCh38
rs780527475 Chr.12: 8602041-8602041 on Build GRCh38
rs999254904 Chr.12: 8602056-8602056 on Build GRCh38
rs1037098768 Chr.12: 8601934-8601934 on Build GRCh38
rs919304677 Chr.12: 8602326-8602326 on Build GRCh38
rs1020501185 Chr.12: 8602168-8602168 on Build GRCh38
rs890040503 Chr.12: 8601965-8601965 on Build GRCh38
rs879871312 Chr.12: 8601947-8601947 on Build GRCh38
rs114391411 Chr.12: 8601919-8601919 on Build GRCh38
rs777357799 Chr.12: 8601904-8601904 on Build GRCh38

View Full Product Details