Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23873987-23874580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNR2
Entrez Gene ID
1269
Gene Name
cannabinoid receptor 2
Gene Aliases
CB-2, CB2, CX5
UniGene
Hs.73037
Gene Chromosome Location
Chr.1: 23870526-23913327 on Build GRCh38
Transcripts
NM_001841.2 XM_011540629.2 XM_017000261.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4931796 c.1057A>G p.R353G Chr.1: 23874561-23874561 on Build GRCh38
6631614 c.1043C>T p.P348L Chr.1: 23874575-23874575 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs201683902 Chr.1: 23874082-23874082 on Build GRCh38
rs1050802239 Chr.1: 23874338-23874338 on Build GRCh38
rs200955792 Chr.1: 23874218-23874218 on Build GRCh38
rs760399323 Chr.1: 23874495-23874495 on Build GRCh38
rs201487654 Chr.1: 23874555-23874555 on Build GRCh38
rs2229582 Chr.1: 23874512-23874512 on Build GRCh38
rs1022074569 Chr.1: 23874312-23874312 on Build GRCh38
rs754195729 Chr.1: 23874504-23874504 on Build GRCh38
rs1053369699 Chr.1: 23874306-23874306 on Build GRCh38
rs927255865 Chr.1: 23874222-23874222 on Build GRCh38
rs1106 Chr.1: 23874282-23874282 on Build GRCh38
rs201940336 Chr.1: 23874457-23874457 on Build GRCh38
rs200330348 Chr.1: 23874265-23874265 on Build GRCh38
rs1105 Chr.1: 23874310-23874310 on Build GRCh38
rs929880494 Chr.1: 23874042-23874042 on Build GRCh38
rs980618266 Chr.1: 23874049-23874049 on Build GRCh38
rs201016530 Chr.1: 23874419-23874419 on Build GRCh38
rs1005203533 Chr.1: 23874089-23874089 on Build GRCh38
rs200527458 Chr.1: 23874066-23874066 on Build GRCh38
rs923397516 Chr.1: 23874467-23874467 on Build GRCh38
rs992070816 Chr.1: 23874111-23874111 on Build GRCh38
rs879246554 Chr.1: 23874421-23874421 on Build GRCh38
rs917053337 Chr.1: 23874038-23874038 on Build GRCh38
rs941252951 Chr.1: 23874302-23874302 on Build GRCh38
rs2229584 Chr.1: 23874479-23874479 on Build GRCh38
rs201694312 Chr.1: 23874374-23874374 on Build GRCh38
rs199630240 Chr.1: 23874383-23874383 on Build GRCh38
rs757586769 Chr.1: 23874211-23874211 on Build GRCh38
rs763896316 Chr.1: 23874496-23874496 on Build GRCh38
rs909781655 Chr.1: 23874300-23874300 on Build GRCh38
rs200571757 Chr.1: 23874021-23874021 on Build GRCh38
rs1030466699 Chr.1: 23874118-23874118 on Build GRCh38
rs143960267 Chr.1: 23874422-23874422 on Build GRCh38
rs2229585 Chr.1: 23874431-23874431 on Build GRCh38
rs199966756 Chr.1: 23874098-23874098 on Build GRCh38
rs937384589 Chr.1: 23874035-23874035 on Build GRCh38
rs1050831547 Chr.1: 23874487-23874487 on Build GRCh38
rs1130316 Chr.1: 23874266-23874266 on Build GRCh38
rs551688994 Chr.1: 23874472-23874472 on Build GRCh38
rs200407932 Chr.1: 23874469-23874469 on Build GRCh38
rs779625711 Chr.1: 23874210-23874210 on Build GRCh38
rs2229586 Chr.1: 23874413-23874413 on Build GRCh38
rs199966509 Chr.1: 23874142-23874142 on Build GRCh38
rs148142264 Chr.1: 23874549-23874549 on Build GRCh38
rs202235055 Chr.1: 23874307-23874307 on Build GRCh38
rs573667686 Chr.1: 23874208-23874208 on Build GRCh38
rs200856456 Chr.1: 23874180-23874180 on Build GRCh38
rs1130320 Chr.1: 23874116-23874116 on Build GRCh38
rs201318944 Chr.1: 23874326-23874326 on Build GRCh38
rs758031503 Chr.1: 23874538-23874538 on Build GRCh38
rs908973572 Chr.1: 23874149-23874149 on Build GRCh38
rs750919886 Chr.1: 23874527-23874527 on Build GRCh38
rs541013896 Chr.1: 23874221-23874221 on Build GRCh38
rs758916991 Chr.1: 23874485-23874485 on Build GRCh38
rs201617271 Chr.1: 23874069-23874069 on Build GRCh38
rs929787100 Chr.1: 23874277-23874277 on Build GRCh38
rs201582730 Chr.1: 23874507-23874507 on Build GRCh38
rs1130321 Chr.1: 23874034-23874034 on Build GRCh38
rs1000489968 Chr.1: 23874539-23874539 on Build GRCh38
rs902045862 Chr.1: 23874272-23874272 on Build GRCh38
rs1033510334 Chr.1: 23874344-23874344 on Build GRCh38
rs202031201 Chr.1: 23874262-23874262 on Build GRCh38
rs1020981020 Chr.1: 23874373-23874373 on Build GRCh38
rs200136483 Chr.1: 23874399-23874399 on Build GRCh38
rs200095777 Chr.1: 23874215-23874215 on Build GRCh38
rs201160547 Chr.1: 23874446-23874446 on Build GRCh38
rs765615838 Chr.1: 23874525-23874525 on Build GRCh38
rs200719571 Chr.1: 23874385-23874385 on Build GRCh38
rs760468108 Chr.1: 23874488-23874488 on Build GRCh38
rs200706567 Chr.1: 23874072-23874072 on Build GRCh38
rs199599665 Chr.1: 23874558-23874558 on Build GRCh38
rs199509097 Chr.1: 23874362-23874362 on Build GRCh38
rs992946797 Chr.1: 23874414-23874414 on Build GRCh38
rs559635272 Chr.1: 23874244-23874244 on Build GRCh38
rs969153044 Chr.1: 23874164-23874164 on Build GRCh38
rs137935661 Chr.1: 23874401-23874401 on Build GRCh38
rs992870194 Chr.1: 23874270-23874270 on Build GRCh38
rs202216673 Chr.1: 23874420-23874420 on Build GRCh38
rs201461373 Chr.1: 23874470-23874470 on Build GRCh38
rs201273734 Chr.1: 23874105-23874105 on Build GRCh38
rs186645513 Chr.1: 23874037-23874037 on Build GRCh38
rs201692255 Chr.1: 23874027-23874027 on Build GRCh38
rs6699937 Chr.1: 23874367-23874367 on Build GRCh38
rs201648173 Chr.1: 23874195-23874195 on Build GRCh38
rs918888834 Chr.1: 23874112-23874112 on Build GRCh38
rs2229583 Chr.1: 23874493-23874493 on Build GRCh38
rs954836833 Chr.1: 23874128-23874128 on Build GRCh38
rs201637175 Chr.1: 23874393-23874393 on Build GRCh38
rs920970865 Chr.1: 23874432-23874432 on Build GRCh38
rs367701893 Chr.1: 23874528-23874528 on Build GRCh38
rs956035121 Chr.1: 23874013-23874013 on Build GRCh38
rs149479710 Chr.1: 23874404-23874404 on Build GRCh38
rs905929650 Chr.1: 23874325-23874325 on Build GRCh38
rs933745453 Chr.1: 23874328-23874328 on Build GRCh38
rs199784805 Chr.1: 23874071-23874071 on Build GRCh38

View Full Product Details