Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32226064-32226545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3I
Entrez Gene ID
8668
Gene Name
eukaryotic translation initiation factor 3 subunit I
Gene Aliases
EIF3S2, PRO2242, TRIP-1, TRIP1, eIF3-beta, eIF3-p36
UniGene
Hs.530096
Gene Chromosome Location
Chr.1: 32221584-32231604 on Build GRCh38
Transcripts
NM_003757.3 XM_017002671.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3977584 c.387T>C p.D129D Chr.1: 32226307-32226307 on Build GRCh38
1296291 c.505G>C p.E169Q Chr.1: 32226507-32226507 on Build GRCh38
1296290 c.396G>C p.Q132H Chr.1: 32226316-32226316 on Build GRCh38
3804957 c.496G>A p.E166K Chr.1: 32226498-32226498 on Build GRCh38
5846390 c.436G>T p.D146Y Chr.1: 32226438-32226438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751405254 Chr.1: 32226357-32226357 on Build GRCh38
rs76933339 Chr.1: 32226345-32226345 on Build GRCh38
rs965760369 Chr.1: 32226327-32226327 on Build GRCh38
rs35594473 Chr.1: 32226125-32226126 on Build GRCh38
rs11554217 Chr.1: 32226278-32226278 on Build GRCh38
rs199681410 Chr.1: 32226310-32226310 on Build GRCh38
rs955659636 Chr.1: 32226095-32226095 on Build GRCh38
rs774203133 Chr.1: 32226476-32226476 on Build GRCh38
rs758637679 Chr.1: 32226410-32226410 on Build GRCh38
rs750288369 Chr.1: 32226218-32226218 on Build GRCh38
rs763686614 Chr.1: 32226203-32226203 on Build GRCh38
rs529108550 Chr.1: 32226104-32226105 on Build GRCh38
rs756108155 Chr.1: 32226238-32226238 on Build GRCh38
rs765426821 Chr.1: 32226506-32226506 on Build GRCh38
rs757619150 Chr.1: 32226246-32226246 on Build GRCh38
rs749554481 Chr.1: 32226130-32226130 on Build GRCh38
rs759377657 Chr.1: 32226486-32226486 on Build GRCh38
rs769222429 Chr.1: 32226420-32226420 on Build GRCh38
rs1056530529 Chr.1: 32226087-32226087 on Build GRCh38
rs1000097457 Chr.1: 32226247-32226247 on Build GRCh38
rs769408795 Chr.1: 32226459-32226459 on Build GRCh38
rs377229890 Chr.1: 32226361-32226361 on Build GRCh38
rs866279978 Chr.1: 32226481-32226481 on Build GRCh38
rs772972113 Chr.1: 32226343-32226343 on Build GRCh38
rs752792250 Chr.1: 32226229-32226229 on Build GRCh38
rs758346269 Chr.1: 32226228-32226228 on Build GRCh38
rs778024392 Chr.1: 32226239-32226239 on Build GRCh38
rs11554219 Chr.1: 32226264-32226264 on Build GRCh38
rs74986471 Chr.1: 32226284-32226284 on Build GRCh38
rs775042459 Chr.1: 32226169-32226169 on Build GRCh38
rs772813256 Chr.1: 32226466-32226466 on Build GRCh38
rs766228235 Chr.1: 32226347-32226347 on Build GRCh38
rs772513365 Chr.1: 32226167-32226167 on Build GRCh38
rs774859478 Chr.1: 32226144-32226144 on Build GRCh38
rs772912286 Chr.1: 32226375-32226375 on Build GRCh38
rs1018242522 Chr.1: 32226129-32226129 on Build GRCh38
rs964406556 Chr.1: 32226131-32226131 on Build GRCh38
rs367916304 Chr.1: 32226152-32226152 on Build GRCh38
rs572553090 Chr.1: 32226358-32226358 on Build GRCh38
rs757579686 Chr.1: 32226212-32226212 on Build GRCh38
rs780478406 Chr.1: 32226414-32226414 on Build GRCh38
rs750551140 Chr.1: 32226514-32226514 on Build GRCh38
rs111982592 Chr.1: 32226322-32226322 on Build GRCh38
rs760139977 Chr.1: 32226179-32226179 on Build GRCh38
rs200845980 Chr.1: 32226479-32226479 on Build GRCh38
rs754027887 Chr.1: 32226371-32226371 on Build GRCh38
rs761452536 Chr.1: 32226339-32226339 on Build GRCh38
rs776214654 Chr.1: 32226211-32226211 on Build GRCh38
rs372175034 Chr.1: 32226301-32226301 on Build GRCh38
rs113367121 Chr.1: 32226321-32226321 on Build GRCh38
rs201776715 Chr.1: 32226395-32226395 on Build GRCh38
rs764948577 Chr.1: 32226217-32226217 on Build GRCh38
rs149707674 Chr.1: 32226115-32226115 on Build GRCh38
rs920032111 Chr.1: 32226173-32226173 on Build GRCh38
rs529692171 Chr.1: 32226488-32226488 on Build GRCh38
rs193247056 Chr.1: 32226214-32226214 on Build GRCh38
rs202084860 Chr.1: 32226261-32226261 on Build GRCh38
rs1042772235 Chr.1: 32226390-32226390 on Build GRCh38
rs375416487 Chr.1: 32226304-32226304 on Build GRCh38
rs780669264 Chr.1: 32226426-32226426 on Build GRCh38
rs573555723 Chr.1: 32226436-32226436 on Build GRCh38
rs776125853 Chr.1: 32226323-32226323 on Build GRCh38
rs114114874 Chr.1: 32226368-32226368 on Build GRCh38
rs779406186 Chr.1: 32226256-32226256 on Build GRCh38
rs746283063 Chr.1: 32226298-32226298 on Build GRCh38
rs199714576 Chr.1: 32226242-32226242 on Build GRCh38
rs771375018 Chr.1: 32226133-32226133 on Build GRCh38
rs764932109 Chr.1: 32226342-32226342 on Build GRCh38
rs747571117 Chr.1: 32226309-32226309 on Build GRCh38
rs769246019 Chr.1: 32226372-32226372 on Build GRCh38
rs896493148 Chr.1: 32226097-32226097 on Build GRCh38
rs902971917 Chr.1: 32226404-32226404 on Build GRCh38

View Full Product Details