Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53466170-53466769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRTB1
Entrez Gene ID
63948
Gene Name
DMRT like family B with proline rich C-terminal 1
Gene Aliases
-
UniGene
Hs.131654
Gene Chromosome Location
Chr.1: 53459399-53467488 on Build GRCh38
Transcripts
NM_033067.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4924942 c.962-2A>T p.? Chr.1: 53466593-53466593 on Build GRCh38
6274526 c.979G>T p.V327L Chr.1: 53466612-53466612 on Build GRCh38
5353794 c.1015G>A p.E339K Chr.1: 53466648-53466648 on Build GRCh38
6056679 c.1015G>C p.E339Q Chr.1: 53466648-53466648 on Build GRCh38
7364651 c.962-1G>T p.? Chr.1: 53466594-53466594 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756357027 Chr.1: 53466531-53466531 on Build GRCh38
rs201751493 Chr.1: 53466657-53466657 on Build GRCh38
rs987768364 Chr.1: 53466490-53466491 on Build GRCh38
rs755649587 Chr.1: 53466612-53466612 on Build GRCh38
rs984016365 Chr.1: 53466475-53466475 on Build GRCh38
rs760889175 Chr.1: 53466241-53466241 on Build GRCh38
rs777043993 Chr.1: 53466575-53466575 on Build GRCh38
rs796437939 Chr.1: 53466499-53466503 on Build GRCh38
rs780080350 Chr.1: 53466587-53466587 on Build GRCh38
rs1053915246 Chr.1: 53466196-53466196 on Build GRCh38
rs1288636 Chr.1: 53466391-53466391 on Build GRCh38
rs999237777 Chr.1: 53466360-53466360 on Build GRCh38
rs200827162 Chr.1: 53466683-53466683 on Build GRCh38
rs942692738 Chr.1: 53466487-53466487 on Build GRCh38
rs555688787 Chr.1: 53466359-53466359 on Build GRCh38
rs930009810 Chr.1: 53466198-53466198 on Build GRCh38
rs111485741 Chr.1: 53466474-53466474 on Build GRCh38
rs899609186 Chr.1: 53466701-53466701 on Build GRCh38
rs78070589 Chr.1: 53466508-53466508 on Build GRCh38
rs772101185 Chr.1: 53466552-53466552 on Build GRCh38
rs754754143 Chr.1: 53466635-53466635 on Build GRCh38
rs1025551301 Chr.1: 53466432-53466432 on Build GRCh38
rs1038137665 Chr.1: 53466699-53466699 on Build GRCh38
rs776006929 Chr.1: 53466675-53466675 on Build GRCh38
rs543861092 Chr.1: 53466707-53466707 on Build GRCh38
rs1025435223 Chr.1: 53466724-53466724 on Build GRCh38
rs12027442 Chr.1: 53466256-53466256 on Build GRCh38
rs1039761245 Chr.1: 53466300-53466300 on Build GRCh38
rs927982466 Chr.1: 53466611-53466611 on Build GRCh38
rs773041973 Chr.1: 53466693-53466693 on Build GRCh38
rs934181208 Chr.1: 53466580-53466580 on Build GRCh38
rs374195119 Chr.1: 53466620-53466620 on Build GRCh38
rs142741634 Chr.1: 53466370-53466370 on Build GRCh38
rs368660189 Chr.1: 53466673-53466673 on Build GRCh38
rs770964936 Chr.1: 53466656-53466656 on Build GRCh38
rs781227554 Chr.1: 53466431-53466431 on Build GRCh38
rs138818482 Chr.1: 53466619-53466619 on Build GRCh38
rs528654976 Chr.1: 53466327-53466327 on Build GRCh38
rs143591589 Chr.1: 53466649-53466649 on Build GRCh38
rs906469101 Chr.1: 53466298-53466298 on Build GRCh38
rs754692027 Chr.1: 53466722-53466722 on Build GRCh38
rs778711933 Chr.1: 53466636-53466636 on Build GRCh38
rs771464263 Chr.1: 53466557-53466557 on Build GRCh38
rs989685051 Chr.1: 53466745-53466745 on Build GRCh38
rs569751218 Chr.1: 53466455-53466455 on Build GRCh38
rs996480735 Chr.1: 53466394-53466394 on Build GRCh38
rs1012425383 Chr.1: 53466385-53466385 on Build GRCh38
rs58259609 Chr.1: 53466310-53466310 on Build GRCh38
rs1047410765 Chr.1: 53466328-53466329 on Build GRCh38
rs998335460 Chr.1: 53466313-53466313 on Build GRCh38
rs915291029 Chr.1: 53466248-53466248 on Build GRCh38
rs557104565 Chr.1: 53466711-53466711 on Build GRCh38
rs774102883 Chr.1: 53466591-53466591 on Build GRCh38
rs747078990 Chr.1: 53466650-53466650 on Build GRCh38
rs759922494 Chr.1: 53466576-53466576 on Build GRCh38
rs761558767 Chr.1: 53466600-53466600 on Build GRCh38
rs942781589 Chr.1: 53466257-53466257 on Build GRCh38
rs1031260971 Chr.1: 53466735-53466735 on Build GRCh38
rs909882440 Chr.1: 53466478-53466478 on Build GRCh38
rs970560276 Chr.1: 53466413-53466413 on Build GRCh38
rs915342747 Chr.1: 53466541-53466541 on Build GRCh38
rs975927030 Chr.1: 53466502-53466502 on Build GRCh38
rs763094935 Chr.1: 53466701-53466702 on Build GRCh38
rs571564290 Chr.1: 53466362-53466362 on Build GRCh38
rs767075900 Chr.1: 53466601-53466601 on Build GRCh38
rs375771195 Chr.1: 53466674-53466674 on Build GRCh38
rs202026130 Chr.1: 53466641-53466641 on Build GRCh38
rs112615087 Chr.1: 53466433-53466433 on Build GRCh38
rs368024889 Chr.1: 53466595-53466595 on Build GRCh38
rs748303555 Chr.1: 53466640-53466640 on Build GRCh38
rs1032423987 Chr.1: 53466364-53466364 on Build GRCh38
rs747148013 Chr.1: 53466556-53466556 on Build GRCh38
rs765832784 Chr.1: 53466710-53466710 on Build GRCh38
rs368433618 Chr.1: 53466604-53466604 on Build GRCh38
rs1048868877 Chr.1: 53466219-53466219 on Build GRCh38
rs529342636 Chr.1: 53466199-53466199 on Build GRCh38
rs1045060817 Chr.1: 53466274-53466274 on Build GRCh38
rs150600400 Chr.1: 53466744-53466744 on Build GRCh38
rs774466915 Chr.1: 53466729-53466729 on Build GRCh38
rs746700140 Chr.1: 53466719-53466720 on Build GRCh38
rs71579971 Chr.1: 53466264-53466265 on Build GRCh38
rs749941816 Chr.1: 53466603-53466603 on Build GRCh38
rs773286654 Chr.1: 53466555-53466555 on Build GRCh38
rs1042020342 Chr.1: 53466338-53466338 on Build GRCh38
rs773454946 Chr.1: 53466685-53466686 on Build GRCh38
rs779504255 Chr.1: 53466715-53466715 on Build GRCh38

View Full Product Details