Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53466568-53467166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRTB1
Entrez Gene ID
63948
Gene Name
DMRT like family B with proline rich C-terminal 1
Gene Aliases
-
UniGene
Hs.131654
Gene Chromosome Location
Chr.1: 53459399-53467488 on Build GRCh38
Transcripts
NM_033067.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4924942 c.962-2A>T p.? Chr.1: 53466593-53466593 on Build GRCh38
6274526 c.979G>T p.V327L Chr.1: 53466612-53466612 on Build GRCh38
5353794 c.1015G>A p.E339K Chr.1: 53466648-53466648 on Build GRCh38
6056679 c.1015G>C p.E339Q Chr.1: 53466648-53466648 on Build GRCh38
7364651 c.962-1G>T p.? Chr.1: 53466594-53466594 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977466876 Chr.1: 53466804-53466804 on Build GRCh38
rs201751493 Chr.1: 53466657-53466657 on Build GRCh38
rs575319017 Chr.1: 53467033-53467033 on Build GRCh38
rs755649587 Chr.1: 53466612-53466612 on Build GRCh38
rs139615523 Chr.1: 53467017-53467017 on Build GRCh38
rs1003254648 Chr.1: 53467037-53467037 on Build GRCh38
rs386631221 Chr.1: 53467145-53467146 on Build GRCh38
rs1046770303 Chr.1: 53467071-53467071 on Build GRCh38
rs200827162 Chr.1: 53466683-53466683 on Build GRCh38
rs977518818 Chr.1: 53467141-53467141 on Build GRCh38
rs899609186 Chr.1: 53466701-53466701 on Build GRCh38
rs934064731 Chr.1: 53466796-53466796 on Build GRCh38
rs754754143 Chr.1: 53466635-53466635 on Build GRCh38
rs1038137665 Chr.1: 53466699-53466699 on Build GRCh38
rs776006929 Chr.1: 53466675-53466675 on Build GRCh38
rs543861092 Chr.1: 53466707-53466707 on Build GRCh38
rs1025435223 Chr.1: 53466724-53466724 on Build GRCh38
rs190565044 Chr.1: 53466835-53466835 on Build GRCh38
rs1031731249 Chr.1: 53467032-53467032 on Build GRCh38
rs559796716 Chr.1: 53466800-53466800 on Build GRCh38
rs927982466 Chr.1: 53466611-53466611 on Build GRCh38
rs374195119 Chr.1: 53466620-53466620 on Build GRCh38
rs773041973 Chr.1: 53466693-53466693 on Build GRCh38
rs368660189 Chr.1: 53466673-53466673 on Build GRCh38
rs770964936 Chr.1: 53466656-53466656 on Build GRCh38
rs372812657 Chr.1: 53466852-53466854 on Build GRCh38
rs1005371841 Chr.1: 53467079-53467079 on Build GRCh38
rs964419431 Chr.1: 53466753-53466753 on Build GRCh38
rs143591589 Chr.1: 53466649-53466649 on Build GRCh38
rs138818482 Chr.1: 53466619-53466619 on Build GRCh38
rs754692027 Chr.1: 53466722-53466722 on Build GRCh38
rs890836659 Chr.1: 53466984-53466984 on Build GRCh38
rs778711933 Chr.1: 53466636-53466636 on Build GRCh38
rs771763316 Chr.1: 53466905-53466905 on Build GRCh38
rs989685051 Chr.1: 53466745-53466745 on Build GRCh38
rs367608121 Chr.1: 53467137-53467137 on Build GRCh38
rs748071522 Chr.1: 53466880-53466880 on Build GRCh38
rs565721796 Chr.1: 53467142-53467142 on Build GRCh38
rs557104565 Chr.1: 53466711-53466711 on Build GRCh38
rs528869832 Chr.1: 53467085-53467085 on Build GRCh38
rs747078990 Chr.1: 53466650-53466650 on Build GRCh38
rs368516471 Chr.1: 53466847-53466849 on Build GRCh38
rs1029675826 Chr.1: 53467115-53467115 on Build GRCh38
rs894715103 Chr.1: 53466898-53466898 on Build GRCh38
rs1038230781 Chr.1: 53466893-53466893 on Build GRCh38
rs761558767 Chr.1: 53466600-53466600 on Build GRCh38
rs1031260971 Chr.1: 53466735-53466735 on Build GRCh38
rs192453375 Chr.1: 53466751-53466751 on Build GRCh38
rs928029903 Chr.1: 53466815-53466815 on Build GRCh38
rs1044709257 Chr.1: 53466965-53466965 on Build GRCh38
rs758406682 Chr.1: 53467022-53467022 on Build GRCh38
rs763094935 Chr.1: 53466701-53466702 on Build GRCh38
rs767075900 Chr.1: 53466601-53466601 on Build GRCh38
rs941251232 Chr.1: 53466879-53466879 on Build GRCh38
rs185643100 Chr.1: 53466829-53466829 on Build GRCh38
rs375771195 Chr.1: 53466674-53466674 on Build GRCh38
rs202026130 Chr.1: 53466641-53466641 on Build GRCh38
rs932478034 Chr.1: 53466959-53466959 on Build GRCh38
rs996725355 Chr.1: 53467097-53467097 on Build GRCh38
rs371048640 Chr.1: 53467072-53467072 on Build GRCh38
rs6684358 Chr.1: 53466836-53466836 on Build GRCh38
rs748303555 Chr.1: 53466640-53466640 on Build GRCh38
rs975403508 Chr.1: 53466784-53466784 on Build GRCh38
rs368024889 Chr.1: 53466595-53466595 on Build GRCh38
rs182399265 Chr.1: 53466848-53466848 on Build GRCh38
rs1011605534 Chr.1: 53467047-53467047 on Build GRCh38
rs772841051 Chr.1: 53466971-53466971 on Build GRCh38
rs765832784 Chr.1: 53466710-53466710 on Build GRCh38
rs79478662 Chr.1: 53466872-53466872 on Build GRCh38
rs368433618 Chr.1: 53466604-53466604 on Build GRCh38
rs998546295 Chr.1: 53467014-53467014 on Build GRCh38
rs963990642 Chr.1: 53467088-53467088 on Build GRCh38
rs150600400 Chr.1: 53466744-53466744 on Build GRCh38
rs774466915 Chr.1: 53466729-53466729 on Build GRCh38
rs74725113 Chr.1: 53466874-53466874 on Build GRCh38
rs746700140 Chr.1: 53466719-53466720 on Build GRCh38
rs749941816 Chr.1: 53466603-53466603 on Build GRCh38
rs773454946 Chr.1: 53466685-53466686 on Build GRCh38
rs779504255 Chr.1: 53466715-53466715 on Build GRCh38
rs922719301 Chr.1: 53466788-53466788 on Build GRCh38

View Full Product Details