Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 74204246-74204812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FPGT
Entrez Gene ID
8790
Gene Name
fucose-1-phosphate guanylyltransferase
Gene Aliases
GFPP
UniGene
Hs.480085
Gene Chromosome Location
Chr.1: 74198212-74208702 on Build GRCh38
Transcripts
NM_001199328.2 NM_001199329.2 NM_003838.4 NM_001112808.2 NM_001199327.1

Gene Symbol
FPGT-TNNI3K
Entrez Gene ID
100526835
Gene Name
FPGT-TNNI3K readthrough
Gene Aliases
-
UniGene
Hs.480085
Gene Chromosome Location
Chr.1: 74198212-74544432 on Build GRCh38
Transcripts
NM_001199328.2 NM_001199329.2 NM_003838.4 NM_001112808.2 NM_001199327.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1252744 c.387A>C p.K129N Chr.1: 74204434-74204434 on Build GRCh38
912068 c.708G>A p.M236I Chr.1: 74204755-74204755 on Build GRCh38
253292 c.388A>T p.I130F Chr.1: 74204435-74204435 on Build GRCh38
912066 c.357A>G p.Q119Q Chr.1: 74204404-74204404 on Build GRCh38
334301 c.690G>T p.K230N Chr.1: 74204737-74204737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752512715 Chr.1: 74204401-74204401 on Build GRCh38
rs748497020 Chr.1: 74204585-74204585 on Build GRCh38
rs763432006 Chr.1: 74204714-74204715 on Build GRCh38
rs768496054 Chr.1: 74204699-74204699 on Build GRCh38
rs1017628181 Chr.1: 74204291-74204291 on Build GRCh38
rs767553427 Chr.1: 74204391-74204391 on Build GRCh38
rs531880048 Chr.1: 74204451-74204451 on Build GRCh38
rs186422789 Chr.1: 74204641-74204641 on Build GRCh38
rs745945484 Chr.1: 74204778-74204778 on Build GRCh38
rs747287145 Chr.1: 74204683-74204683 on Build GRCh38
rs779293454 Chr.1: 74204744-74204744 on Build GRCh38
rs142249154 Chr.1: 74204584-74204584 on Build GRCh38
rs772215421 Chr.1: 74204680-74204680 on Build GRCh38
rs781425224 Chr.1: 74204382-74204383 on Build GRCh38
rs368063418 Chr.1: 74204494-74204494 on Build GRCh38
rs546009671 Chr.1: 74204766-74204766 on Build GRCh38
rs755472590 Chr.1: 74204671-74204671 on Build GRCh38
rs552656123 Chr.1: 74204322-74204322 on Build GRCh38
rs770235361 Chr.1: 74204695-74204696 on Build GRCh38
rs769045501 Chr.1: 74204565-74204565 on Build GRCh38
rs765874898 Chr.1: 74204729-74204729 on Build GRCh38
rs773687144 Chr.1: 74204463-74204463 on Build GRCh38
rs12046751 Chr.1: 74204488-74204488 on Build GRCh38
rs1014391459 Chr.1: 74204276-74204276 on Build GRCh38
rs181649063 Chr.1: 74204567-74204567 on Build GRCh38
rs780804638 Chr.1: 74204522-74204522 on Build GRCh38
rs753200971 Chr.1: 74204353-74204353 on Build GRCh38
rs9425024 Chr.1: 74204289-74204289 on Build GRCh38
rs542066398 Chr.1: 74204735-74204735 on Build GRCh38
rs778158024 Chr.1: 74204764-74204764 on Build GRCh38
rs760747688 Chr.1: 74204576-74204576 on Build GRCh38
rs896423460 Chr.1: 74204282-74204282 on Build GRCh38
rs746705428 Chr.1: 74204444-74204444 on Build GRCh38
rs758939335 Chr.1: 74204376-74204376 on Build GRCh38
rs756775355 Chr.1: 74204446-74204446 on Build GRCh38
rs779763394 Chr.1: 74204651-74204651 on Build GRCh38
rs367800651 Chr.1: 74204571-74204571 on Build GRCh38
rs547793107 Chr.1: 74204553-74204553 on Build GRCh38
rs761951991 Chr.1: 74204388-74204388 on Build GRCh38
rs781280156 Chr.1: 74204776-74204776 on Build GRCh38
rs761475500 Chr.1: 74204633-74204633 on Build GRCh38
rs560259393 Chr.1: 74204283-74204283 on Build GRCh38
rs764738295 Chr.1: 74204299-74204299 on Build GRCh38
rs758748726 Chr.1: 74204753-74204753 on Build GRCh38
rs767482598 Chr.1: 74204718-74204718 on Build GRCh38
rs756626497 Chr.1: 74204534-74204534 on Build GRCh38
rs767286522 Chr.1: 74204640-74204640 on Build GRCh38
rs768859127 Chr.1: 74204363-74204363 on Build GRCh38
rs764393942 Chr.1: 74204375-74204375 on Build GRCh38
rs141519700 Chr.1: 74204405-74204405 on Build GRCh38
rs369885071 Chr.1: 74204352-74204352 on Build GRCh38
rs763852010 Chr.1: 74204513-74204513 on Build GRCh38
rs894558123 Chr.1: 74204276-74204276 on Build GRCh38
rs576522799 Chr.1: 74204590-74204590 on Build GRCh38
rs756896988 Chr.1: 74204517-74204517 on Build GRCh38
rs978021556 Chr.1: 74204742-74204742 on Build GRCh38
rs372462668 Chr.1: 74204655-74204655 on Build GRCh38
rs375088079 Chr.1: 74204783-74204783 on Build GRCh38
rs767555809 Chr.1: 74204493-74204493 on Build GRCh38
rs887616713 Chr.1: 74204302-74204302 on Build GRCh38
rs771631096 Chr.1: 74204775-74204776 on Build GRCh38
rs1005978392 Chr.1: 74204310-74204310 on Build GRCh38
rs752422898 Chr.1: 74204509-74204509 on Build GRCh38
rs755674654 Chr.1: 74204646-74204646 on Build GRCh38
rs1055882317 Chr.1: 74204273-74204273 on Build GRCh38
rs139749795 Chr.1: 74204593-74204593 on Build GRCh38
rs762906455 Chr.1: 74204694-74204694 on Build GRCh38
rs765172400 Chr.1: 74204499-74204499 on Build GRCh38
rs199776247 Chr.1: 74204472-74204472 on Build GRCh38
rs774623275 Chr.1: 74204487-74204487 on Build GRCh38
rs777485615 Chr.1: 74204416-74204416 on Build GRCh38
rs768972029 Chr.1: 74204480-74204480 on Build GRCh38
rs745389270 Chr.1: 74204530-74204530 on Build GRCh38
rs763223014 Chr.1: 74204369-74204369 on Build GRCh38
rs1024494053 Chr.1: 74204279-74204279 on Build GRCh38
rs760256404 Chr.1: 74204781-74204782 on Build GRCh38
rs751198362 Chr.1: 74204516-74204516 on Build GRCh38
rs770258802 Chr.1: 74204349-74204349 on Build GRCh38
rs773753089 Chr.1: 74204712-74204713 on Build GRCh38
rs747300584 Chr.1: 74204358-74204358 on Build GRCh38
rs142435282 Chr.1: 74204411-74204411 on Build GRCh38
rs374701961 Chr.1: 74204710-74204710 on Build GRCh38
rs746432268 Chr.1: 74204343-74204343 on Build GRCh38
rs774134052 Chr.1: 74204705-74204705 on Build GRCh38
rs143587621 Chr.1: 74204685-74204685 on Build GRCh38
rs914786797 Chr.1: 74204445-74204445 on Build GRCh38
rs1028425591 Chr.1: 74204631-74204631 on Build GRCh38
rs572415148 Chr.1: 74204752-74204752 on Build GRCh38
rs753755679 Chr.1: 74204394-74204394 on Build GRCh38
rs925182773 Chr.1: 74204721-74204721 on Build GRCh38
rs1004422299 Chr.1: 74204563-74204563 on Build GRCh38
rs34392787 Chr.1: 74204621-74204622 on Build GRCh38
rs139882690 Chr.1: 74204397-74204397 on Build GRCh38
rs769774345 Chr.1: 74204623-74204623 on Build GRCh38
rs757565795 Chr.1: 74204556-74204556 on Build GRCh38
rs764978201 Chr.1: 74204398-74204398 on Build GRCh38
rs201412617 Chr.1: 74204738-74204738 on Build GRCh38
rs749820622 Chr.1: 74204561-74204561 on Build GRCh38
rs369079941 Chr.1: 74204724-74204724 on Build GRCh38
rs375107941 Chr.1: 74204672-74204672 on Build GRCh38
rs148933608 Chr.1: 74204675-74204675 on Build GRCh38
rs372968994 Chr.1: 74204701-74204701 on Build GRCh38
rs777967379 Chr.1: 74204682-74204682 on Build GRCh38
rs746019668 Chr.1: 74204618-74204618 on Build GRCh38
rs1000629671 Chr.1: 74204269-74204269 on Build GRCh38
rs772303621 Chr.1: 74204570-74204570 on Build GRCh38
rs747199251 Chr.1: 74204765-74204765 on Build GRCh38
rs772018105 Chr.1: 74204723-74204723 on Build GRCh38
rs976175427 Chr.1: 74204387-74204387 on Build GRCh38
rs774978731 Chr.1: 74204780-74204781 on Build GRCh38
rs775943283 Chr.1: 74204479-74204479 on Build GRCh38
rs76458321 Chr.1: 74204587-74204587 on Build GRCh38
rs373185282 Chr.1: 74204382-74204382 on Build GRCh38

View Full Product Details