Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 85581192-85581759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYR61
Entrez Gene ID
3491
Gene Name
cysteine rich angiogenic inducer 61
Gene Aliases
CCN1, GIG1, IGFBP10
UniGene
Hs.8867
Gene Chromosome Location
Chr.1: 85580761-85583967 on Build GRCh38
Transcripts
NM_001554.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4989054 c.200C>T p.T67M Chr.1: 85581501-85581501 on Build GRCh38
6437095 c.272G>A p.C91Y Chr.1: 85581573-85581573 on Build GRCh38
4436794 c.150C>T p.C50C Chr.1: 85581451-85581451 on Build GRCh38
4411120 c.131G>T p.G44V Chr.1: 85581432-85581432 on Build GRCh38
912712 c.189C>G p.D63E Chr.1: 85581490-85581490 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915516451 Chr.1: 85581333-85581333 on Build GRCh38
rs867614780 Chr.1: 85581237-85581237 on Build GRCh38
rs770351633 Chr.1: 85581458-85581458 on Build GRCh38
rs758236830 Chr.1: 85581431-85581431 on Build GRCh38
rs111770855 Chr.1: 85581585-85581585 on Build GRCh38
rs774655685 Chr.1: 85581491-85581491 on Build GRCh38
rs374414176 Chr.1: 85581483-85581483 on Build GRCh38
rs772130312 Chr.1: 85581501-85581501 on Build GRCh38
rs552702347 Chr.1: 85581266-85581266 on Build GRCh38
rs566337737 Chr.1: 85581267-85581267 on Build GRCh38
rs756989998 Chr.1: 85581560-85581560 on Build GRCh38
rs937030530 Chr.1: 85581301-85581301 on Build GRCh38
rs775764118 Chr.1: 85581331-85581331 on Build GRCh38
rs9658584 Chr.1: 85581628-85581628 on Build GRCh38
rs1006894670 Chr.1: 85581558-85581558 on Build GRCh38
rs765658185 Chr.1: 85581323-85581323 on Build GRCh38
rs754828661 Chr.1: 85581394-85581394 on Build GRCh38
rs749989161 Chr.1: 85581562-85581562 on Build GRCh38
rs751253845 Chr.1: 85581557-85581557 on Build GRCh38
rs934065546 Chr.1: 85581377-85581377 on Build GRCh38
rs778276696 Chr.1: 85581397-85581397 on Build GRCh38
rs746698451 Chr.1: 85581445-85581445 on Build GRCh38
rs2297141 Chr.1: 85581241-85581241 on Build GRCh38
rs949286605 Chr.1: 85581237-85581237 on Build GRCh38
rs757430397 Chr.1: 85581606-85581606 on Build GRCh38
rs755595392 Chr.1: 85581565-85581565 on Build GRCh38
rs756662941 Chr.1: 85581356-85581356 on Build GRCh38
rs994150900 Chr.1: 85581721-85581721 on Build GRCh38
rs760669726 Chr.1: 85581507-85581507 on Build GRCh38
rs923762423 Chr.1: 85581244-85581244 on Build GRCh38
rs766274149 Chr.1: 85581380-85581380 on Build GRCh38
rs765306827 Chr.1: 85581513-85581513 on Build GRCh38
rs769081761 Chr.1: 85581601-85581601 on Build GRCh38
rs780766451 Chr.1: 85581452-85581452 on Build GRCh38
rs764349811 Chr.1: 85581591-85581591 on Build GRCh38
rs761889399 Chr.1: 85581414-85581414 on Build GRCh38
rs754081825 Chr.1: 85581599-85581600 on Build GRCh38
rs746389670 Chr.1: 85581406-85581406 on Build GRCh38
rs868688488 Chr.1: 85581641-85581641 on Build GRCh38
rs926944706 Chr.1: 85581299-85581299 on Build GRCh38
rs773403271 Chr.1: 85581506-85581506 on Build GRCh38
rs971107057 Chr.1: 85581284-85581284 on Build GRCh38
rs750406551 Chr.1: 85581423-85581423 on Build GRCh38
rs767663303 Chr.1: 85581421-85581421 on Build GRCh38
rs1006868273 Chr.1: 85581318-85581318 on Build GRCh38
rs377546318 Chr.1: 85581587-85581587 on Build GRCh38
rs749823611 Chr.1: 85581599-85581599 on Build GRCh38
rs962479419 Chr.1: 85581242-85581242 on Build GRCh38
rs752609594 Chr.1: 85581428-85581428 on Build GRCh38
rs1051220565 Chr.1: 85581383-85581383 on Build GRCh38
rs752660470 Chr.1: 85581543-85581543 on Build GRCh38
rs372157931 Chr.1: 85581409-85581409 on Build GRCh38
rs752890662 Chr.1: 85581334-85581335 on Build GRCh38
rs1007209365 Chr.1: 85581217-85581217 on Build GRCh38
rs538510187 Chr.1: 85581272-85581272 on Build GRCh38
rs917598123 Chr.1: 85581467-85581467 on Build GRCh38
rs757325644 Chr.1: 85581358-85581358 on Build GRCh38
rs936541011 Chr.1: 85581265-85581265 on Build GRCh38
rs754959588 Chr.1: 85581424-85581424 on Build GRCh38
rs780704620 Chr.1: 85581561-85581561 on Build GRCh38
rs978000839 Chr.1: 85581586-85581586 on Build GRCh38
rs182829412 Chr.1: 85581650-85581650 on Build GRCh38
rs901177361 Chr.1: 85581699-85581699 on Build GRCh38
rs777563956 Chr.1: 85581399-85581399 on Build GRCh38
rs753831998 Chr.1: 85581382-85581382 on Build GRCh38
rs769026494 Chr.1: 85581490-85581490 on Build GRCh38
rs990461944 Chr.1: 85581441-85581441 on Build GRCh38
rs749764704 Chr.1: 85581511-85581511 on Build GRCh38
rs779339414 Chr.1: 85581605-85581606 on Build GRCh38
rs777618646 Chr.1: 85581438-85581438 on Build GRCh38
rs1041445624 Chr.1: 85581319-85581319 on Build GRCh38
rs770452661 Chr.1: 85581401-85581401 on Build GRCh38
rs376836267 Chr.1: 85581446-85581446 on Build GRCh38
rs747895300 Chr.1: 85581398-85581398 on Build GRCh38
rs764337279 Chr.1: 85581410-85581410 on Build GRCh38
rs558834066 Chr.1: 85581288-85581288 on Build GRCh38
rs369237188 Chr.1: 85581400-85581400 on Build GRCh38
rs760716323 Chr.1: 85581353-85581354 on Build GRCh38
rs145852662 Chr.1: 85581408-85581408 on Build GRCh38
rs748450946 Chr.1: 85581615-85581615 on Build GRCh38
rs575107561 Chr.1: 85581345-85581345 on Build GRCh38
rs762936508 Chr.1: 85581545-85581545 on Build GRCh38
rs138327040 Chr.1: 85581734-85581734 on Build GRCh38
rs191487668 Chr.1: 85581454-85581454 on Build GRCh38
rs774530780 Chr.1: 85581411-85581411 on Build GRCh38
rs946840087 Chr.1: 85581336-85581336 on Build GRCh38
rs981564198 Chr.1: 85581711-85581711 on Build GRCh38
rs749697975 Chr.1: 85581487-85581487 on Build GRCh38
rs779351827 Chr.1: 85581614-85581614 on Build GRCh38
rs751703185 Chr.1: 85581357-85581357 on Build GRCh38

View Full Product Details