Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 115725351-115725850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASQ2
Entrez Gene ID
845
Gene Name
calsequestrin 2
Gene Aliases
PDIB2
UniGene
Hs.57975
Gene Chromosome Location
Chr.1: 115700003-115768805 on Build GRCh38
Transcripts
NM_001232.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6018919 c.738-6_738-4delTTC p.? Chr.1: 115725557-115725559 on Build GRCh38
6584096 c.775G>A p.E259K Chr.1: 115725516-115725516 on Build GRCh38
5458086 c.758G>A p.R253H Chr.1: 115725533-115725533 on Build GRCh38
894356 c.752G>A p.R251H Chr.1: 115725539-115725539 on Build GRCh38
6432652 c.751C>T p.R251C Chr.1: 115725540-115725540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs76232874 Chr.1: 115725674-115725674 on Build GRCh38
rs1000057740 Chr.1: 115725715-115725715 on Build GRCh38
rs980666532 Chr.1: 115725430-115725430 on Build GRCh38
rs56889721 Chr.1: 115725579-115725580 on Build GRCh38
rs770233719 Chr.1: 115725573-115725573 on Build GRCh38
rs112950350 Chr.1: 115725756-115725756 on Build GRCh38
rs775955473 Chr.1: 115725574-115725574 on Build GRCh38
rs375131666 Chr.1: 115725587-115725587 on Build GRCh38
rs151232156 Chr.1: 115725658-115725658 on Build GRCh38
rs951417781 Chr.1: 115725811-115725811 on Build GRCh38
rs763687725 Chr.1: 115725636-115725636 on Build GRCh38
rs369499991 Chr.1: 115725463-115725463 on Build GRCh38
rs149191305 Chr.1: 115725581-115725581 on Build GRCh38
rs946228330 Chr.1: 115725493-115725493 on Build GRCh38
rs761205574 Chr.1: 115725482-115725482 on Build GRCh38
rs575264268 Chr.1: 115725428-115725428 on Build GRCh38
rs1017099302 Chr.1: 115725828-115725828 on Build GRCh38
rs777808216 Chr.1: 115725491-115725491 on Build GRCh38
rs190111142 Chr.1: 115725808-115725808 on Build GRCh38
rs978810907 Chr.1: 115725831-115725831 on Build GRCh38
rs772703632 Chr.1: 115725578-115725581 on Build GRCh38
rs769169039 Chr.1: 115725570-115725581 on Build GRCh38
rs780913821 Chr.1: 115725776-115725776 on Build GRCh38
rs56955144 Chr.1: 115725440-115725440 on Build GRCh38
rs567810196 Chr.1: 115725743-115725743 on Build GRCh38
rs60028701 Chr.1: 115725525-115725525 on Build GRCh38
rs551385087 Chr.1: 115725641-115725641 on Build GRCh38
rs1030086352 Chr.1: 115725805-115725805 on Build GRCh38
rs539532946 Chr.1: 115725794-115725794 on Build GRCh38
rs397735272 Chr.1: 115725580-115725581 on Build GRCh38
rs397516642 Chr.1: 115725510-115725510 on Build GRCh38
rs201847837 Chr.1: 115725581-115725581 on Build GRCh38
rs187306418 Chr.1: 115725542-115725542 on Build GRCh38
rs1018227257 Chr.1: 115725731-115725731 on Build GRCh38
rs376385940 Chr.1: 115725461-115725461 on Build GRCh38
rs767095892 Chr.1: 115725560-115725561 on Build GRCh38
rs763290471 Chr.1: 115725578-115725578 on Build GRCh38
rs941603963 Chr.1: 115725437-115725437 on Build GRCh38
rs900852572 Chr.1: 115725632-115725632 on Build GRCh38
rs181769947 Chr.1: 115725517-115725517 on Build GRCh38
rs878872937 Chr.1: 115725558-115725564 on Build GRCh38
rs982885646 Chr.1: 115725827-115725827 on Build GRCh38
rs748928919 Chr.1: 115725465-115725465 on Build GRCh38
rs773824666 Chr.1: 115725477-115725477 on Build GRCh38
rs151115064 Chr.1: 115725543-115725543 on Build GRCh38
rs727504510 Chr.1: 115725534-115725534 on Build GRCh38
rs200265771 Chr.1: 115725539-115725539 on Build GRCh38
rs558374907 Chr.1: 115725813-115725813 on Build GRCh38
rs750138764 Chr.1: 115725830-115725830 on Build GRCh38
rs533123916 Chr.1: 115725628-115725628 on Build GRCh38
rs876657750 Chr.1: 115725556-115725556 on Build GRCh38
rs547360790 Chr.1: 115725471-115725471 on Build GRCh38
rs556993377 Chr.1: 115725398-115725398 on Build GRCh38
rs4475752 Chr.1: 115725662-115725662 on Build GRCh38
rs937061457 Chr.1: 115725645-115725645 on Build GRCh38
rs765517257 Chr.1: 115725527-115725527 on Build GRCh38
rs756906492 Chr.1: 115725695-115725695 on Build GRCh38
rs904389154 Chr.1: 115725710-115725710 on Build GRCh38
rs76056486 Chr.1: 115725456-115725456 on Build GRCh38
rs545821699 Chr.1: 115725429-115725429 on Build GRCh38
rs770958200 Chr.1: 115725558-115725560 on Build GRCh38
rs114869970 Chr.1: 115725449-115725449 on Build GRCh38
rs542504128 Chr.1: 115725558-115725559 on Build GRCh38
rs922752705 Chr.1: 115725577-115725577 on Build GRCh38
rs781113955 Chr.1: 115725540-115725540 on Build GRCh38
rs762677092 Chr.1: 115725577-115725578 on Build GRCh38
rs910067612 Chr.1: 115725624-115725624 on Build GRCh38
rs942991368 Chr.1: 115725616-115725617 on Build GRCh38
rs562653885 Chr.1: 115725625-115725625 on Build GRCh38
rs933631673 Chr.1: 115725660-115725660 on Build GRCh38
rs768261470 Chr.1: 115725551-115725551 on Build GRCh38
rs748981981 Chr.1: 115725546-115725546 on Build GRCh38
rs876657751 Chr.1: 115725505-115725505 on Build GRCh38
rs747267070 Chr.1: 115725814-115725814 on Build GRCh38
rs113914107 Chr.1: 115725392-115725392 on Build GRCh38
rs762015552 Chr.1: 115725592-115725592 on Build GRCh38
rs775788421 Chr.1: 115725559-115725560 on Build GRCh38
rs766861780 Chr.1: 115725489-115725489 on Build GRCh38
rs535459644 Chr.1: 115725407-115725407 on Build GRCh38
rs759948112 Chr.1: 115725572-115725572 on Build GRCh38
rs772703928 Chr.1: 115725558-115725562 on Build GRCh38
rs777263692 Chr.1: 115725571-115725571 on Build GRCh38
rs749555206 Chr.1: 115725558-115725563 on Build GRCh38
rs753016201 Chr.1: 115725529-115725529 on Build GRCh38
rs756069840 Chr.1: 115725373-115725373 on Build GRCh38
rs899831114 Chr.1: 115725742-115725742 on Build GRCh38
rs58617078 Chr.1: 115725554-115725554 on Build GRCh38
rs746004006 Chr.1: 115725558-115725561 on Build GRCh38
rs747358070 Chr.1: 115725568-115725569 on Build GRCh38
rs1041508410 Chr.1: 115725698-115725698 on Build GRCh38
rs747600202 Chr.1: 115725552-115725552 on Build GRCh38
rs776874142 Chr.1: 115725508-115725508 on Build GRCh38
rs771347208 Chr.1: 115725567-115725567 on Build GRCh38
rs557052499 Chr.1: 115725810-115725810 on Build GRCh38
rs776183314 Chr.1: 115725558-115725558 on Build GRCh38
rs199571249 Chr.1: 115725533-115725533 on Build GRCh38

View Full Product Details