Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 229388115-229388520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNER
Entrez Gene ID
92737
Gene Name
delta/notch like EGF repeat containing
Gene Aliases
UNQ26, bet
UniGene
Hs.234074
Gene Chromosome Location
Chr.2: 229357629-229714570 on Build GRCh38
Transcripts
NM_139072.3 XM_005246950.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6157171 c.1766C>A p.P589H Chr.2: 229388354-229388354 on Build GRCh38
4902876 c.1780G>A p.G594R Chr.2: 229388340-229388340 on Build GRCh38
210323 c.1734C>T p.C578C Chr.2: 229388386-229388386 on Build GRCh38
5421203 c.1788C>T p.C596C Chr.2: 229388332-229388332 on Build GRCh38
1692122 c.1834G>A p.V612M Chr.2: 229388286-229388286 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561463040 Chr.2: 229388243-229388243 on Build GRCh38
rs746289573 Chr.2: 229388289-229388289 on Build GRCh38
rs766470085 Chr.2: 229388265-229388265 on Build GRCh38
rs893911270 Chr.2: 229388217-229388217 on Build GRCh38
rs765734328 Chr.2: 229388275-229388275 on Build GRCh38
rs749215233 Chr.2: 229388429-229388429 on Build GRCh38
rs370082704 Chr.2: 229388400-229388400 on Build GRCh38
rs747246282 Chr.2: 229388331-229388331 on Build GRCh38
rs112272021 Chr.2: 229388184-229388184 on Build GRCh38
rs751274945 Chr.2: 229388286-229388286 on Build GRCh38
rs943210242 Chr.2: 229388183-229388183 on Build GRCh38
rs917102534 Chr.2: 229388246-229388246 on Build GRCh38
rs200514482 Chr.2: 229388385-229388385 on Build GRCh38
rs1053792208 Chr.2: 229388349-229388349 on Build GRCh38
rs756959616 Chr.2: 229388392-229388392 on Build GRCh38
rs1029093836 Chr.2: 229388482-229388482 on Build GRCh38
rs144906958 Chr.2: 229388318-229388318 on Build GRCh38
rs776689590 Chr.2: 229388346-229388346 on Build GRCh38
rs551547290 Chr.2: 229388438-229388438 on Build GRCh38
rs542792046 Chr.2: 229388144-229388144 on Build GRCh38
rs371542012 Chr.2: 229388239-229388239 on Build GRCh38
rs753095191 Chr.2: 229388276-229388276 on Build GRCh38
rs973128647 Chr.2: 229388156-229388163 on Build GRCh38
rs376723054 Chr.2: 229388386-229388386 on Build GRCh38
rs773463006 Chr.2: 229388250-229388250 on Build GRCh38
rs769405660 Chr.2: 229388219-229388219 on Build GRCh38
rs754286370 Chr.2: 229388266-229388266 on Build GRCh38
rs765756411 Chr.2: 229388356-229388356 on Build GRCh38
rs562379428 Chr.2: 229388402-229388402 on Build GRCh38
rs878973633 Chr.2: 229388273-229388273 on Build GRCh38
rs755629131 Chr.2: 229388428-229388428 on Build GRCh38
rs561797653 Chr.2: 229388446-229388446 on Build GRCh38
rs572212590 Chr.2: 229388205-229388205 on Build GRCh38
rs779094906 Chr.2: 229388274-229388274 on Build GRCh38
rs141269091 Chr.2: 229388299-229388299 on Build GRCh38
rs751608883 Chr.2: 229388140-229388140 on Build GRCh38
rs775905924 Chr.2: 229388361-229388361 on Build GRCh38
rs375310744 Chr.2: 229388279-229388279 on Build GRCh38
rs555994251 Chr.2: 229388163-229388163 on Build GRCh38
rs202150174 Chr.2: 229388296-229388296 on Build GRCh38
rs1012383355 Chr.2: 229388362-229388362 on Build GRCh38
rs760706711 Chr.2: 229388264-229388264 on Build GRCh38
rs929224255 Chr.2: 229388151-229388151 on Build GRCh38
rs532716043 Chr.2: 229388347-229388347 on Build GRCh38
rs775349390 Chr.2: 229388221-229388221 on Build GRCh38
rs767590055 Chr.2: 229388382-229388382 on Build GRCh38
rs747992736 Chr.2: 229388448-229388448 on Build GRCh38
rs368405241 Chr.2: 229388297-229388297 on Build GRCh38
rs773374942 Chr.2: 229388320-229388320 on Build GRCh38
rs973145183 Chr.2: 229388403-229388403 on Build GRCh38
rs763324502 Chr.2: 229388381-229388381 on Build GRCh38
rs866594376 Chr.2: 229388339-229388339 on Build GRCh38
rs1038855459 Chr.2: 229388211-229388211 on Build GRCh38
rs962002788 Chr.2: 229388142-229388142 on Build GRCh38
rs770933741 Chr.2: 229388344-229388344 on Build GRCh38
rs777541195 Chr.2: 229388284-229388284 on Build GRCh38
rs1051721611 Chr.2: 229388162-229388162 on Build GRCh38

View Full Product Details