Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 236581600-236582177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACKR3
Entrez Gene ID
57007
Gene Name
atypical chemokine receptor 3
Gene Aliases
CMKOR1, CXC-R7, CXCR-7, CXCR7, GPR159, RDC-1, RDC1
UniGene
Hs.471751
Gene Chromosome Location
Chr.2: 236537131-236582358 on Build GRCh38
Transcripts
NM_020311.2 XM_005246097.2 XM_005246098.3 XM_017004516.1

SNP Detail

SNP ID Chromosome Location
rs943220864 Chr.2: 236581681-236581681 on Build GRCh38
rs764344543 Chr.2: 236582076-236582077 on Build GRCh38
rs139798730 Chr.2: 236581790-236581790 on Build GRCh38
rs886587651 Chr.2: 236581706-236581706 on Build GRCh38
rs182954328 Chr.2: 236581974-236581974 on Build GRCh38
rs753642800 Chr.2: 236582079-236582079 on Build GRCh38
rs998140266 Chr.2: 236581738-236581738 on Build GRCh38
rs930877621 Chr.2: 236581687-236581687 on Build GRCh38
rs923439519 Chr.2: 236582062-236582063 on Build GRCh38
rs923173925 Chr.2: 236582130-236582130 on Build GRCh38
rs918944775 Chr.2: 236581928-236581928 on Build GRCh38
rs1033115357 Chr.2: 236581834-236581834 on Build GRCh38
rs933268542 Chr.2: 236582137-236582137 on Build GRCh38
rs984910789 Chr.2: 236581983-236581983 on Build GRCh38
rs1041763159 Chr.2: 236582089-236582089 on Build GRCh38
rs1038895869 Chr.2: 236581715-236581715 on Build GRCh38
rs981136735 Chr.2: 236581645-236581645 on Build GRCh38
rs903099161 Chr.2: 236582135-236582135 on Build GRCh38
rs999315991 Chr.2: 236581767-236581767 on Build GRCh38
rs957731033 Chr.2: 236581844-236581844 on Build GRCh38
rs971677606 Chr.2: 236581881-236581881 on Build GRCh38
rs191607547 Chr.2: 236581740-236581740 on Build GRCh38
rs901930169 Chr.2: 236581727-236581728 on Build GRCh38
rs1011467622 Chr.2: 236581765-236581765 on Build GRCh38
rs760993497 Chr.2: 236581728-236581728 on Build GRCh38
rs967317204 Chr.2: 236581774-236581774 on Build GRCh38
rs1002689036 Chr.2: 236581782-236581782 on Build GRCh38
rs984980905 Chr.2: 236581980-236581980 on Build GRCh38
rs560845604 Chr.2: 236582144-236582144 on Build GRCh38
rs940972839 Chr.2: 236581990-236581990 on Build GRCh38
rs943200482 Chr.2: 236582078-236582078 on Build GRCh38
rs536816806 Chr.2: 236581819-236581819 on Build GRCh38
rs555188999 Chr.2: 236581836-236581836 on Build GRCh38
rs1023213667 Chr.2: 236581865-236581865 on Build GRCh38
rs552827097 Chr.2: 236581833-236581833 on Build GRCh38
rs990479354 Chr.2: 236581632-236581633 on Build GRCh38
rs184987816 Chr.2: 236581739-236581739 on Build GRCh38
rs775875655 Chr.2: 236581916-236581916 on Build GRCh38
rs538830584 Chr.2: 236581851-236581851 on Build GRCh38
rs909017425 Chr.2: 236581679-236581679 on Build GRCh38
rs1038883731 Chr.2: 236582099-236582099 on Build GRCh38
rs1048819192 Chr.2: 236581678-236581678 on Build GRCh38
rs952503451 Chr.2: 236581889-236581889 on Build GRCh38
rs182305985 Chr.2: 236581655-236581655 on Build GRCh38
rs929069271 Chr.2: 236581673-236581673 on Build GRCh38
rs556157398 Chr.2: 236581699-236581700 on Build GRCh38
rs766402558 Chr.2: 236581913-236581913 on Build GRCh38
rs571525641 Chr.2: 236581656-236581656 on Build GRCh38
rs937110307 Chr.2: 236581671-236581671 on Build GRCh38
rs1042298755 Chr.2: 236582138-236582138 on Build GRCh38
rs868808740 Chr.2: 236581644-236581644 on Build GRCh38
rs893106755 Chr.2: 236582078-236582085 on Build GRCh38
rs540554440 Chr.2: 236582066-236582075 on Build GRCh38
rs575056402 Chr.2: 236581718-236581718 on Build GRCh38
rs188100070 Chr.2: 236581988-236581988 on Build GRCh38
rs950434561 Chr.2: 236581964-236581964 on Build GRCh38
rs1050958442 Chr.2: 236582068-236582068 on Build GRCh38
rs1040509267 Chr.2: 236582048-236582048 on Build GRCh38
rs544747578 Chr.2: 236581861-236581861 on Build GRCh38
rs192909521 Chr.2: 236582021-236582021 on Build GRCh38
rs776718091 Chr.2: 236581979-236581979 on Build GRCh38

View Full Product Details