Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 36244479-36245017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAR2
Entrez Gene ID
25980
Gene Name
AAR2 splicing factor homolog
Gene Aliases
C20orf4, CGI-23
UniGene
Hs.11314
Gene Chromosome Location
Chr.20: 36236136-36256941 on Build GRCh38
Transcripts
NM_001271874.1 NM_015511.4 XM_006723770.3 XM_011528762.2 XM_011528763.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5527703 c.929C>T p.P310L Chr.20: 36244868-36244868 on Build GRCh38
5552056 c.962A>G p.D321G Chr.20: 36244901-36244901 on Build GRCh38
4954503 c.902T>G p.I301S Chr.20: 36244841-36244841 on Build GRCh38
301830 c.930C>A p.P310P Chr.20: 36244869-36244869 on Build GRCh38
6589281 c.857C>A p.A286E Chr.20: 36244796-36244796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746573248 Chr.20: 36244909-36244909 on Build GRCh38
rs1006565148 Chr.20: 36244552-36244552 on Build GRCh38
rs539592841 Chr.20: 36244896-36244896 on Build GRCh38
rs776514716 Chr.20: 36244677-36244679 on Build GRCh38
rs377157882 Chr.20: 36244889-36244889 on Build GRCh38
rs1006847964 Chr.20: 36244818-36244818 on Build GRCh38
rs983831798 Chr.20: 36244919-36244919 on Build GRCh38
rs753691544 Chr.20: 36244705-36244705 on Build GRCh38
rs532751767 Chr.20: 36244537-36244537 on Build GRCh38
rs543945528 Chr.20: 36244513-36244513 on Build GRCh38
rs113156393 Chr.20: 36244933-36244933 on Build GRCh38
rs989159070 Chr.20: 36244916-36244916 on Build GRCh38
rs767054441 Chr.20: 36244683-36244683 on Build GRCh38
rs770205897 Chr.20: 36244704-36244704 on Build GRCh38
rs928542035 Chr.20: 36244602-36244602 on Build GRCh38
rs778970346 Chr.20: 36244737-36244737 on Build GRCh38
rs917476138 Chr.20: 36244970-36244970 on Build GRCh38
rs562607102 Chr.20: 36244528-36244528 on Build GRCh38
rs1043152983 Chr.20: 36244684-36244684 on Build GRCh38
rs1001115790 Chr.20: 36244783-36244783 on Build GRCh38
rs761398969 Chr.20: 36244923-36244923 on Build GRCh38
rs117200013 Chr.20: 36244833-36244833 on Build GRCh38
rs953004216 Chr.20: 36244625-36244625 on Build GRCh38
rs750451424 Chr.20: 36244739-36244739 on Build GRCh38
rs374099380 Chr.20: 36244883-36244883 on Build GRCh38
rs752925227 Chr.20: 36244971-36244971 on Build GRCh38
rs1042212591 Chr.20: 36244757-36244757 on Build GRCh38
rs150912491 Chr.20: 36244744-36244744 on Build GRCh38
rs141514980 Chr.20: 36244646-36244646 on Build GRCh38
rs750338713 Chr.20: 36244856-36244856 on Build GRCh38
rs781449292 Chr.20: 36244870-36244870 on Build GRCh38
rs766373739 Chr.20: 36244956-36244956 on Build GRCh38
rs145635219 Chr.20: 36244766-36244766 on Build GRCh38
rs758286265 Chr.20: 36244857-36244857 on Build GRCh38
rs768875388 Chr.20: 36244773-36244773 on Build GRCh38
rs901436722 Chr.20: 36244746-36244746 on Build GRCh38
rs781465836 Chr.20: 36244786-36244786 on Build GRCh38
rs397864454 Chr.20: 36244684-36244685 on Build GRCh38
rs773817924 Chr.20: 36244801-36244801 on Build GRCh38
rs965067879 Chr.20: 36244592-36244592 on Build GRCh38
rs140703484 Chr.20: 36244794-36244794 on Build GRCh38
rs372921853 Chr.20: 36244743-36244743 on Build GRCh38
rs767696794 Chr.20: 36244968-36244968 on Build GRCh38
rs775840886 Chr.20: 36244977-36244979 on Build GRCh38
rs904578033 Chr.20: 36244770-36244770 on Build GRCh38
rs918316408 Chr.20: 36244608-36244608 on Build GRCh38
rs751627509 Chr.20: 36244958-36244958 on Build GRCh38
rs112647820 Chr.20: 36244681-36244682 on Build GRCh38
rs760425982 Chr.20: 36244702-36244702 on Build GRCh38
rs368599383 Chr.20: 36244716-36244716 on Build GRCh38
rs138478673 Chr.20: 36244824-36244824 on Build GRCh38
rs980014886 Chr.20: 36244598-36244598 on Build GRCh38
rs1032274733 Chr.20: 36244518-36244518 on Build GRCh38
rs771074176 Chr.20: 36244676-36244680 on Build GRCh38
rs376767059 Chr.20: 36244863-36244863 on Build GRCh38
rs35283785 Chr.20: 36244680-36244681 on Build GRCh38
rs202224152 Chr.20: 36244823-36244823 on Build GRCh38
rs762884456 Chr.20: 36244954-36244954 on Build GRCh38
rs763925130 Chr.20: 36244704-36244704 on Build GRCh38
rs1000680916 Chr.20: 36244509-36244509 on Build GRCh38
rs759141865 Chr.20: 36244673-36244673 on Build GRCh38
rs758937745 Chr.20: 36244802-36244802 on Build GRCh38
rs775133590 Chr.20: 36244908-36244908 on Build GRCh38
rs550754862 Chr.20: 36244848-36244848 on Build GRCh38
rs776383295 Chr.20: 36244917-36244917 on Build GRCh38
rs369290761 Chr.20: 36244869-36244869 on Build GRCh38
rs115016138 Chr.20: 36244579-36244579 on Build GRCh38
rs547595252 Chr.20: 36244526-36244526 on Build GRCh38
rs148948059 Chr.20: 36244924-36244924 on Build GRCh38
rs185526702 Chr.20: 36244964-36244964 on Build GRCh38
rs568801996 Chr.20: 36244653-36244655 on Build GRCh38
rs867724451 Chr.20: 36244705-36244705 on Build GRCh38
rs1057307448 Chr.20: 36244711-36244711 on Build GRCh38
rs768398840 Chr.20: 36244911-36244911 on Build GRCh38
rs759785059 Chr.20: 36244955-36244955 on Build GRCh38
rs144176613 Chr.20: 36244819-36244820 on Build GRCh38
rs984269726 Chr.20: 36244641-36244641 on Build GRCh38
rs147679615 Chr.20: 36244787-36244787 on Build GRCh38
rs748440774 Chr.20: 36244881-36244881 on Build GRCh38
rs201599475 Chr.20: 36244882-36244882 on Build GRCh38
rs763908537 Chr.20: 36244825-36244825 on Build GRCh38
rs917276647 Chr.20: 36244738-36244738 on Build GRCh38
rs752437824 Chr.20: 36244690-36244690 on Build GRCh38
rs972925602 Chr.20: 36244596-36244596 on Build GRCh38

View Full Product Details