Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 25891592-25892191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APP
Entrez Gene ID
351
Gene Name
amyloid beta precursor protein
Gene Aliases
AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2
UniGene
Hs.434980
Gene Chromosome Location
Chr.21: 25880550-26171128 on Build GRCh38
Transcripts
NM_000484.3 NM_001136016.3 NM_001136129.2 NM_001136130.2 NM_001136131.2 NM_001204301.1 NM_001204302.1 NM_001204303.1 NM_201413.2 NM_201414.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1413494 c.2180A>T p.Q727L Chr.21: 25891753-25891753 on Build GRCh38
2841330 c.2138C>T p.A713V Chr.21: 25891795-25891795 on Build GRCh38
1029634 c.2140A>G p.T714A Chr.21: 25891793-25891793 on Build GRCh38
3549993 c.2110G>A p.G704R Chr.21: 25891823-25891823 on Build GRCh38
7231872 c.2189C>A p.S730Y Chr.21: 25891744-25891744 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760838173 Chr.21: 25891829-25891829 on Build GRCh38
rs960074789 Chr.21: 25891976-25891976 on Build GRCh38
rs559574618 Chr.21: 25891933-25891933 on Build GRCh38
rs200093246 Chr.21: 25891696-25891696 on Build GRCh38
rs903760518 Chr.21: 25891620-25891621 on Build GRCh38
rs145564988 Chr.21: 25891785-25891785 on Build GRCh38
rs747973071 Chr.21: 25891882-25891887 on Build GRCh38
rs10638132 Chr.21: 25892159-25892160 on Build GRCh38
rs761736619 Chr.21: 25891880-25891880 on Build GRCh38
rs201269325 Chr.21: 25891808-25891808 on Build GRCh38
rs63750264 Chr.21: 25891784-25891784 on Build GRCh38
rs942324098 Chr.21: 25891697-25891697 on Build GRCh38
rs770140305 Chr.21: 25891701-25891702 on Build GRCh38
rs929848388 Chr.21: 25891616-25891616 on Build GRCh38
rs63749810 Chr.21: 25891853-25891853 on Build GRCh38
rs775632358 Chr.21: 25891707-25891707 on Build GRCh38
rs574315379 Chr.21: 25891618-25891618 on Build GRCh38
rs1016303518 Chr.21: 25892092-25892092 on Build GRCh38
rs1034409475 Chr.21: 25891962-25891962 on Build GRCh38
rs63750579 Chr.21: 25891856-25891856 on Build GRCh38
rs999364206 Chr.21: 25891961-25891961 on Build GRCh38
rs8127843 Chr.21: 25892116-25892116 on Build GRCh38
rs567117927 Chr.21: 25892145-25892145 on Build GRCh38
rs63750066 Chr.21: 25891796-25891796 on Build GRCh38
rs200388443 Chr.21: 25891818-25891818 on Build GRCh38
rs527350045 Chr.21: 25892144-25892144 on Build GRCh38
rs200059479 Chr.21: 25892047-25892048 on Build GRCh38
rs760281326 Chr.21: 25892048-25892049 on Build GRCh38
rs116650065 Chr.21: 25891800-25891800 on Build GRCh38
rs745948082 Chr.21: 25891814-25891814 on Build GRCh38
rs200499263 Chr.21: 25891675-25891675 on Build GRCh38
rs397867487 Chr.21: 25892065-25892065 on Build GRCh38
rs63749964 Chr.21: 25891783-25891783 on Build GRCh38
rs63750851 Chr.21: 25891786-25891786 on Build GRCh38
rs112984849 Chr.21: 25891871-25891871 on Build GRCh38
rs63750868 Chr.21: 25891789-25891789 on Build GRCh38
rs768446605 Chr.21: 25891854-25891854 on Build GRCh38
rs947816090 Chr.21: 25891932-25891932 on Build GRCh38
rs774394741 Chr.21: 25891689-25891689 on Build GRCh38
rs63750627 Chr.21: 25891788-25891788 on Build GRCh38
rs367709245 Chr.21: 25891634-25891639 on Build GRCh38
rs928855601 Chr.21: 25892127-25892127 on Build GRCh38
rs976119546 Chr.21: 25891665-25891665 on Build GRCh38
rs63750921 Chr.21: 25891820-25891820 on Build GRCh38
rs754569140 Chr.21: 25891902-25891902 on Build GRCh38
rs984467046 Chr.21: 25892076-25892076 on Build GRCh38
rs953453957 Chr.21: 25892063-25892066 on Build GRCh38
rs200791950 Chr.21: 25891887-25891887 on Build GRCh38
rs773042462 Chr.21: 25891890-25891891 on Build GRCh38
rs367620604 Chr.21: 25892066-25892066 on Build GRCh38
rs745475489 Chr.21: 25892047-25892049 on Build GRCh38
rs751177661 Chr.21: 25891896-25891896 on Build GRCh38
rs150115492 Chr.21: 25891674-25891674 on Build GRCh38
rs1029022213 Chr.21: 25892059-25892066 on Build GRCh38
rs778913571 Chr.21: 25891878-25891882 on Build GRCh38
rs528414329 Chr.21: 25891955-25891955 on Build GRCh38
rs573613532 Chr.21: 25891725-25891725 on Build GRCh38
rs771951164 Chr.21: 25891883-25891885 on Build GRCh38
rs749919142 Chr.21: 25891794-25891794 on Build GRCh38
rs148888161 Chr.21: 25891809-25891809 on Build GRCh38
rs63750643 Chr.21: 25891793-25891793 on Build GRCh38
rs201429297 Chr.21: 25891891-25891891 on Build GRCh38
rs201724975 Chr.21: 25891863-25891863 on Build GRCh38
rs63751039 Chr.21: 25891855-25891855 on Build GRCh38
rs762464558 Chr.21: 25891894-25891894 on Build GRCh38
rs201465309 Chr.21: 25891684-25891684 on Build GRCh38
rs63750399 Chr.21: 25891787-25891787 on Build GRCh38
rs760462028 Chr.21: 25891720-25891720 on Build GRCh38
rs951242739 Chr.21: 25892066-25892066 on Build GRCh38
rs969147992 Chr.21: 25891934-25891934 on Build GRCh38
rs544705359 Chr.21: 25891807-25891807 on Build GRCh38
rs775713536 Chr.21: 25891826-25891826 on Build GRCh38
rs63750151 Chr.21: 25891761-25891761 on Build GRCh38
rs942355083 Chr.21: 25892100-25892100 on Build GRCh38
rs768417419 Chr.21: 25891990-25891990 on Build GRCh38
rs35476811 Chr.21: 25892160-25892161 on Build GRCh38
rs199528897 Chr.21: 25891883-25891883 on Build GRCh38
rs571117086 Chr.21: 25892042-25892042 on Build GRCh38
rs63750734 Chr.21: 25891790-25891790 on Build GRCh38
rs756556593 Chr.21: 25891629-25891637 on Build GRCh38
rs776543778 Chr.21: 25891868-25891868 on Build GRCh38
rs1012062519 Chr.21: 25891617-25891617 on Build GRCh38
rs63750973 Chr.21: 25891792-25891792 on Build GRCh38
rs745774335 Chr.21: 25891625-25891625 on Build GRCh38
rs201020743 Chr.21: 25891743-25891743 on Build GRCh38
rs1048298298 Chr.21: 25891645-25891645 on Build GRCh38
rs909786462 Chr.21: 25892094-25892094 on Build GRCh38
rs63751122 Chr.21: 25891765-25891765 on Build GRCh38
rs966403398 Chr.21: 25891662-25891662 on Build GRCh38
rs773705711 Chr.21: 25891671-25891671 on Build GRCh38
rs962150459 Chr.21: 25892104-25892104 on Build GRCh38
rs111931176 Chr.21: 25891870-25891870 on Build GRCh38
rs200446085 Chr.21: 25891805-25891805 on Build GRCh38
rs573584712 Chr.21: 25891613-25891613 on Build GRCh38
rs546708356 Chr.21: 25891914-25891914 on Build GRCh38
rs63750671 Chr.21: 25891858-25891858 on Build GRCh38
rs114043297 Chr.21: 25891982-25891982 on Build GRCh38
rs10547971 Chr.21: 25892047-25892047 on Build GRCh38
rs550672423 Chr.21: 25892141-25892141 on Build GRCh38
rs1030998813 Chr.21: 25891651-25891651 on Build GRCh38
rs758297878 Chr.21: 25892064-25892066 on Build GRCh38
rs1800557 Chr.21: 25891795-25891795 on Build GRCh38

View Full Product Details