Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39522149-39522737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF4
Entrez Gene ID
468
Gene Name
activating transcription factor 4
Gene Aliases
CREB-2, CREB2, TAXREB67, TXREB
UniGene
Hs.496487
Gene Chromosome Location
Chr.22: 39519709-39522686 on Build GRCh38
Transcripts
NM_001675.4 NM_182810.2 XM_017028807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
287763 c.731G>T p.R244M Chr.22: 39522277-39522277 on Build GRCh38
1034311 c.912G>T p.E304D Chr.22: 39522458-39522458 on Build GRCh38
4663965 c.693T>A p.S231S Chr.22: 39522239-39522239 on Build GRCh38
3912828 c.680C>T p.S227F Chr.22: 39522226-39522226 on Build GRCh38
331238 c.631G>T p.E211* Chr.22: 39522177-39522177 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755371414 Chr.22: 39522619-39522619 on Build GRCh38
rs1803323 Chr.22: 39522318-39522318 on Build GRCh38
rs761949909 Chr.22: 39522598-39522598 on Build GRCh38
rs900144635 Chr.22: 39522630-39522630 on Build GRCh38
rs757998406 Chr.22: 39522509-39522512 on Build GRCh38
rs757538227 Chr.22: 39522382-39522382 on Build GRCh38
rs759406925 Chr.22: 39522429-39522429 on Build GRCh38
rs765638221 Chr.22: 39522527-39522527 on Build GRCh38
rs148305437 Chr.22: 39522603-39522603 on Build GRCh38
rs549806403 Chr.22: 39522653-39522653 on Build GRCh38
rs750748993 Chr.22: 39522560-39522560 on Build GRCh38
rs1044311654 Chr.22: 39522552-39522552 on Build GRCh38
rs558004697 Chr.22: 39522497-39522497 on Build GRCh38
rs761742541 Chr.22: 39522293-39522293 on Build GRCh38
rs200470722 Chr.22: 39522315-39522315 on Build GRCh38
rs772171586 Chr.22: 39522171-39522171 on Build GRCh38
rs768796191 Chr.22: 39522596-39522596 on Build GRCh38
rs760317005 Chr.22: 39522206-39522206 on Build GRCh38
rs568793240 Chr.22: 39522353-39522353 on Build GRCh38
rs147899546 Chr.22: 39522325-39522325 on Build GRCh38
rs781391904 Chr.22: 39522240-39522240 on Build GRCh38
rs374500320 Chr.22: 39522687-39522687 on Build GRCh38
rs950409361 Chr.22: 39522253-39522253 on Build GRCh38
rs764347691 Chr.22: 39522460-39522460 on Build GRCh38
rs758440007 Chr.22: 39522233-39522233 on Build GRCh38
rs773683906 Chr.22: 39522195-39522195 on Build GRCh38
rs369819561 Chr.22: 39522377-39522377 on Build GRCh38
rs200688485 Chr.22: 39522545-39522545 on Build GRCh38
rs772729966 Chr.22: 39522406-39522406 on Build GRCh38
rs753747328 Chr.22: 39522427-39522427 on Build GRCh38
rs750409615 Chr.22: 39522304-39522304 on Build GRCh38
rs907004587 Chr.22: 39522384-39522384 on Build GRCh38
rs1002567786 Chr.22: 39522221-39522221 on Build GRCh38
rs774987120 Chr.22: 39522691-39522691 on Build GRCh38
rs150046216 Chr.22: 39522256-39522256 on Build GRCh38
rs746575830 Chr.22: 39522330-39522330 on Build GRCh38
rs753752402 Chr.22: 39522218-39522218 on Build GRCh38
rs541765130 Chr.22: 39522546-39522546 on Build GRCh38
rs148936525 Chr.22: 39522342-39522342 on Build GRCh38
rs772997758 Chr.22: 39522347-39522347 on Build GRCh38
rs534223114 Chr.22: 39522637-39522638 on Build GRCh38
rs140871401 Chr.22: 39522312-39522312 on Build GRCh38
rs751431913 Chr.22: 39522356-39522356 on Build GRCh38
rs773948133 Chr.22: 39522508-39522508 on Build GRCh38
rs370215013 Chr.22: 39522316-39522316 on Build GRCh38
rs761885223 Chr.22: 39522525-39522525 on Build GRCh38
rs11556096 Chr.22: 39522452-39522452 on Build GRCh38
rs777967380 Chr.22: 39522379-39522379 on Build GRCh38
rs746468641 Chr.22: 39522615-39522617 on Build GRCh38
rs141463329 Chr.22: 39522407-39522407 on Build GRCh38
rs774454112 Chr.22: 39522327-39522328 on Build GRCh38
rs771506424 Chr.22: 39522698-39522698 on Build GRCh38
rs752301523 Chr.22: 39522509-39522510 on Build GRCh38
rs182791688 Chr.22: 39522173-39522173 on Build GRCh38
rs146633351 Chr.22: 39522170-39522170 on Build GRCh38
rs768305008 Chr.22: 39522387-39522387 on Build GRCh38
rs547803084 Chr.22: 39522690-39522690 on Build GRCh38
rs552036293 Chr.22: 39522280-39522280 on Build GRCh38
rs767533408 Chr.22: 39522366-39522366 on Build GRCh38
rs770163255 Chr.22: 39522322-39522322 on Build GRCh38
rs547982920 Chr.22: 39522231-39522231 on Build GRCh38
rs749762484 Chr.22: 39522324-39522324 on Build GRCh38
rs539627686 Chr.22: 39522184-39522184 on Build GRCh38
rs757102478 Chr.22: 39522226-39522226 on Build GRCh38
rs763453138 Chr.22: 39522601-39522601 on Build GRCh38
rs141451590 Chr.22: 39522611-39522611 on Build GRCh38
rs1803324 Chr.22: 39522512-39522512 on Build GRCh38
rs781774201 Chr.22: 39522638-39522639 on Build GRCh38
rs1026027404 Chr.22: 39522415-39522415 on Build GRCh38
rs765065019 Chr.22: 39522489-39522491 on Build GRCh38
rs780005904 Chr.22: 39522310-39522310 on Build GRCh38
rs779044628 Chr.22: 39522229-39522229 on Build GRCh38
rs748841276 Chr.22: 39522506-39522506 on Build GRCh38
rs771492320 Chr.22: 39522262-39522262 on Build GRCh38
rs769410051 Chr.22: 39522500-39522500 on Build GRCh38
rs769641664 Chr.22: 39522402-39522402 on Build GRCh38
rs746955630 Chr.22: 39522648-39522648 on Build GRCh38
rs747807293 Chr.22: 39522393-39522393 on Build GRCh38
rs754114703 Chr.22: 39522520-39522520 on Build GRCh38
rs1046801556 Chr.22: 39522701-39522701 on Build GRCh38
rs893040430 Chr.22: 39522349-39522349 on Build GRCh38
rs568102933 Chr.22: 39522670-39522670 on Build GRCh38
rs139408953 Chr.22: 39522196-39522196 on Build GRCh38
rs189076537 Chr.22: 39522636-39522636 on Build GRCh38
rs747215775 Chr.22: 39522174-39522174 on Build GRCh38
rs753343637 Chr.22: 39522621-39522621 on Build GRCh38
rs369015417 Chr.22: 39522466-39522466 on Build GRCh38
rs770355543 Chr.22: 39522186-39522186 on Build GRCh38
rs1017211406 Chr.22: 39522448-39522448 on Build GRCh38
rs377261776 Chr.22: 39522350-39522350 on Build GRCh38
rs189922789 Chr.22: 39522539-39522539 on Build GRCh38
rs56117154 Chr.22: 39522629-39522629 on Build GRCh38
rs373424533 Chr.22: 39522449-39522449 on Build GRCh38
rs2229273 Chr.22: 39522515-39522515 on Build GRCh38
rs749692770 Chr.22: 39522611-39522614 on Build GRCh38
rs780133133 Chr.22: 39522235-39522235 on Build GRCh38
rs774364445 Chr.22: 39522424-39522424 on Build GRCh38
rs934623226 Chr.22: 39522333-39522333 on Build GRCh38
rs562783366 Chr.22: 39522693-39522693 on Build GRCh38
rs777862308 Chr.22: 39522445-39522445 on Build GRCh38
rs752840127 Chr.22: 39522437-39522437 on Build GRCh38
rs1060000 Chr.22: 39522587-39522587 on Build GRCh38
rs147301363 Chr.22: 39522308-39522308 on Build GRCh38
rs779098982 Chr.22: 39522477-39522477 on Build GRCh38
rs890846106 Chr.22: 39522715-39522715 on Build GRCh38
rs955572638 Chr.22: 39522674-39522674 on Build GRCh38
rs777087053 Chr.22: 39522548-39522548 on Build GRCh38
rs765086046 Chr.22: 39522296-39522296 on Build GRCh38
rs746323130 Chr.22: 39522386-39522386 on Build GRCh38
rs770552438 Chr.22: 39522553-39522553 on Build GRCh38
rs774705715 Chr.22: 39522329-39522329 on Build GRCh38
rs562456003 Chr.22: 39522185-39522185 on Build GRCh38
rs775436808 Chr.22: 39522584-39522584 on Build GRCh38
rs1009492330 Chr.22: 39522717-39522717 on Build GRCh38
rs777947241 Chr.22: 39522529-39522529 on Build GRCh38
rs763800470 Chr.22: 39522215-39522215 on Build GRCh38
rs778049781 Chr.22: 39522323-39522323 on Build GRCh38
rs138696573 Chr.22: 39522517-39522517 on Build GRCh38
rs781238888 Chr.22: 39522378-39522378 on Build GRCh38
rs765439697 Chr.22: 39522599-39522599 on Build GRCh38
rs776455859 Chr.22: 39522337-39522337 on Build GRCh38
rs193150673 Chr.22: 39522637-39522637 on Build GRCh38
rs745712018 Chr.22: 39522571-39522571 on Build GRCh38
rs757673491 Chr.22: 39522470-39522470 on Build GRCh38
rs376820946 Chr.22: 39522632-39522632 on Build GRCh38
rs528891567 Chr.22: 39522683-39522683 on Build GRCh38
rs763863444 Chr.22: 39522279-39522279 on Build GRCh38
rs540001357 Chr.22: 39522494-39522494 on Build GRCh38
rs748971103 Chr.22: 39522549-39522549 on Build GRCh38
rs776223971 Chr.22: 39522392-39522392 on Build GRCh38
rs1006300408 Chr.22: 39522436-39522436 on Build GRCh38
rs151021575 Chr.22: 39522431-39522431 on Build GRCh38
rs1044236394 Chr.22: 39522384-39522386 on Build GRCh38
rs527474447 Chr.22: 39522270-39522270 on Build GRCh38
rs564042258 Chr.22: 39522633-39522633 on Build GRCh38
rs754832232 Chr.22: 39522436-39522438 on Build GRCh38
rs756804295 Chr.22: 39522636-39522637 on Build GRCh38
rs778987567 Chr.22: 39522169-39522169 on Build GRCh38
rs768929209 Chr.22: 39522182-39522182 on Build GRCh38
rs768859496 Chr.22: 39522236-39522236 on Build GRCh38
rs1028467420 Chr.22: 39522476-39522476 on Build GRCh38
rs572596597 Chr.22: 39522482-39522482 on Build GRCh38
rs760631397 Chr.22: 39522353-39522355 on Build GRCh38
rs761509481 Chr.22: 39522339-39522339 on Build GRCh38
rs776509275 Chr.22: 39522597-39522597 on Build GRCh38
rs756258147 Chr.22: 39522369-39522369 on Build GRCh38
rs766045339 Chr.22: 39522422-39522422 on Build GRCh38
rs183254244 Chr.22: 39522620-39522620 on Build GRCh38
rs777524346 Chr.22: 39522504-39522504 on Build GRCh38
rs560051439 Chr.22: 39522242-39522242 on Build GRCh38
rs746366547 Chr.22: 39522652-39522653 on Build GRCh38
rs746998189 Chr.22: 39522669-39522669 on Build GRCh38
rs766355529 Chr.22: 39522372-39522373 on Build GRCh38
rs376877986 Chr.22: 39522573-39522573 on Build GRCh38
rs906932678 Chr.22: 39522213-39522213 on Build GRCh38
rs764085250 Chr.22: 39522276-39522278 on Build GRCh38
rs779512289 Chr.22: 39522383-39522383 on Build GRCh38
rs372679887 Chr.22: 39522363-39522363 on Build GRCh38
rs11556100 Chr.22: 39522245-39522245 on Build GRCh38
rs749719594 Chr.22: 39522381-39522381 on Build GRCh38
rs749707544 Chr.22: 39522625-39522625 on Build GRCh38
rs566819541 Chr.22: 39522570-39522570 on Build GRCh38
rs779767311 Chr.22: 39522645-39522645 on Build GRCh38
rs751686315 Chr.22: 39522268-39522268 on Build GRCh38
rs768251819 Chr.22: 39522334-39522334 on Build GRCh38
rs373242816 Chr.22: 39522518-39522518 on Build GRCh38
rs760557862 Chr.22: 39522585-39522585 on Build GRCh38
rs770110284 Chr.22: 39522249-39522249 on Build GRCh38
rs752311404 Chr.22: 39522202-39522202 on Build GRCh38

View Full Product Details