Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24029424-24029981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDCP
Entrez Gene ID
80304
Gene Name
WD repeat and coiled coil containing
Gene Aliases
C2orf44, PP384
UniGene
Hs.24624
Gene Chromosome Location
Chr.2: 24029336-24047426 on Build GRCh38
Transcripts
NM_001142319.1 NM_025203.2

SNP Detail

SNP ID Chromosome Location
rs1034112302 Chr.2: 24029538-24029538 on Build GRCh38
rs767501819 Chr.2: 24029584-24029584 on Build GRCh38
rs1010527650 Chr.2: 24029453-24029453 on Build GRCh38
rs766554000 Chr.2: 24029878-24029878 on Build GRCh38
rs973682180 Chr.2: 24029802-24029802 on Build GRCh38
rs78427813 Chr.2: 24029834-24029835 on Build GRCh38
rs754393589 Chr.2: 24029728-24029728 on Build GRCh38
rs139467067 Chr.2: 24029821-24029821 on Build GRCh38
rs530204637 Chr.2: 24029861-24029861 on Build GRCh38
rs572167949 Chr.2: 24029536-24029536 on Build GRCh38
rs1029499209 Chr.2: 24029589-24029589 on Build GRCh38
rs760492119 Chr.2: 24029746-24029746 on Build GRCh38
rs538971329 Chr.2: 24029479-24029479 on Build GRCh38
rs3203 Chr.2: 24029547-24029547 on Build GRCh38
rs917399320 Chr.2: 24029881-24029881 on Build GRCh38
rs917368169 Chr.2: 24029468-24029468 on Build GRCh38
rs918566976 Chr.2: 24029701-24029701 on Build GRCh38
rs904211263 Chr.2: 24029480-24029480 on Build GRCh38
rs779960347 Chr.2: 24029909-24029909 on Build GRCh38
rs750166186 Chr.2: 24029657-24029657 on Build GRCh38
rs999921789 Chr.2: 24029483-24029483 on Build GRCh38
rs937464189 Chr.2: 24029452-24029452 on Build GRCh38
rs553677092 Chr.2: 24029495-24029495 on Build GRCh38
rs927500649 Chr.2: 24029825-24029825 on Build GRCh38
rs998393305 Chr.2: 24029571-24029571 on Build GRCh38
rs189525033 Chr.2: 24029567-24029567 on Build GRCh38
rs868368850 Chr.2: 24029577-24029577 on Build GRCh38
rs532683339 Chr.2: 24029762-24029762 on Build GRCh38
rs951134692 Chr.2: 24029613-24029613 on Build GRCh38
rs946165861 Chr.2: 24029945-24029945 on Build GRCh38
rs972036994 Chr.2: 24029606-24029606 on Build GRCh38
rs911316202 Chr.2: 24029786-24029786 on Build GRCh38
rs1003950955 Chr.2: 24029653-24029653 on Build GRCh38
rs962260696 Chr.2: 24029590-24029590 on Build GRCh38
rs958975979 Chr.2: 24029826-24029826 on Build GRCh38
rs3204 Chr.2: 24029566-24029566 on Build GRCh38
rs992998434 Chr.2: 24029868-24029868 on Build GRCh38
rs369104686 Chr.2: 24029828-24029828 on Build GRCh38
rs185009752 Chr.2: 24029557-24029557 on Build GRCh38
rs984142758 Chr.2: 24029778-24029778 on Build GRCh38
rs561342325 Chr.2: 24029552-24029552 on Build GRCh38
rs559797819 Chr.2: 24029829-24029829 on Build GRCh38

View Full Product Details