Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 51075134-51075564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK3
Entrez Gene ID
1795
Gene Name
dedicator of cytokinesis 3
Gene Aliases
MOCA, PBP
UniGene
Hs.476284
Gene Chromosome Location
Chr.3: 50674969-51384198 on Build GRCh38
Transcripts
NM_004947.4 XM_005264914.3 XM_005264915.3 XM_005264916.3 XM_005264917.3 XM_005264918.3 XM_006713008.3 XM_006713009.3 XM_006713010.3 XM_017005825.1 XM_017005826.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4118805 c.467A>C p.H156P Chr.3: 51075358-51075358 on Build GRCh38
7400829 c.549+5G>C p.? Chr.3: 51075445-51075445 on Build GRCh38
260544 c.535G>T p.D179Y Chr.3: 51075426-51075426 on Build GRCh38
3775195 c.543T>C p.Y181Y Chr.3: 51075434-51075434 on Build GRCh38
6511119 c.546G>A p.K182K Chr.3: 51075437-51075437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989679226 Chr.3: 51075486-51075486 on Build GRCh38
rs766404599 Chr.3: 51075434-51075434 on Build GRCh38
rs1009189452 Chr.3: 51075332-51075332 on Build GRCh38
rs747893689 Chr.3: 51075348-51075348 on Build GRCh38
rs762964297 Chr.3: 51075433-51075433 on Build GRCh38
rs957460121 Chr.3: 51075482-51075483 on Build GRCh38
rs751448866 Chr.3: 51075324-51075324 on Build GRCh38
rs569049127 Chr.3: 51075263-51075263 on Build GRCh38
rs375236026 Chr.3: 51075398-51075398 on Build GRCh38
rs754897931 Chr.3: 51075472-51075472 on Build GRCh38
rs945278674 Chr.3: 51075494-51075494 on Build GRCh38
rs902518892 Chr.3: 51075369-51075369 on Build GRCh38
rs369702617 Chr.3: 51075426-51075426 on Build GRCh38
rs34989289 Chr.3: 51075497-51075497 on Build GRCh38
rs777417594 Chr.3: 51075352-51075352 on Build GRCh38
rs994934218 Chr.3: 51075272-51075272 on Build GRCh38
rs1046051402 Chr.3: 51075160-51075160 on Build GRCh38
rs1018426989 Chr.3: 51075454-51075454 on Build GRCh38
rs1001906182 Chr.3: 51075163-51075163 on Build GRCh38
rs374712870 Chr.3: 51075306-51075306 on Build GRCh38
rs549005052 Chr.3: 51075493-51075496 on Build GRCh38
rs372196048 Chr.3: 51075396-51075396 on Build GRCh38
rs1019362298 Chr.3: 51075406-51075406 on Build GRCh38
rs780945033 Chr.3: 51075346-51075346 on Build GRCh38
rs940201230 Chr.3: 51075284-51075284 on Build GRCh38
rs539853165 Chr.3: 51075407-51075407 on Build GRCh38
rs777119939 Chr.3: 51075413-51075413 on Build GRCh38
rs758436551 Chr.3: 51075320-51075320 on Build GRCh38
rs192181274 Chr.3: 51075478-51075478 on Build GRCh38
rs929662031 Chr.3: 51075157-51075157 on Build GRCh38
rs775143931 Chr.3: 51075318-51075318 on Build GRCh38
rs199683497 Chr.3: 51075404-51075404 on Build GRCh38
rs111413708 Chr.3: 51075210-51075210 on Build GRCh38
rs748765805 Chr.3: 51075357-51075357 on Build GRCh38
rs928596811 Chr.3: 51075183-51075183 on Build GRCh38
rs999568216 Chr.3: 51075370-51075370 on Build GRCh38
rs951001400 Chr.3: 51075315-51075315 on Build GRCh38
rs766316595 Chr.3: 51075321-51075321 on Build GRCh38
rs879101547 Chr.3: 51075278-51075278 on Build GRCh38
rs141362459 Chr.3: 51075259-51075259 on Build GRCh38
rs762158144 Chr.3: 51075425-51075425 on Build GRCh38
rs200607525 Chr.3: 51075326-51075326 on Build GRCh38
rs973780832 Chr.3: 51075528-51075528 on Build GRCh38
rs755718820 Chr.3: 51075351-51075351 on Build GRCh38
rs774899243 Chr.3: 51075470-51075470 on Build GRCh38
rs1054824363 Chr.3: 51075247-51075247 on Build GRCh38
rs751536738 Chr.3: 51075467-51075467 on Build GRCh38
rs573067165 Chr.3: 51075364-51075364 on Build GRCh38
rs773425649 Chr.3: 51075432-51075432 on Build GRCh38
rs567472431 Chr.3: 51075220-51075220 on Build GRCh38
rs869061142 Chr.3: 51075531-51075532 on Build GRCh38
rs767367850 Chr.3: 51075473-51075473 on Build GRCh38
rs765377525 Chr.3: 51075431-51075431 on Build GRCh38
rs1043792383 Chr.3: 51075341-51075341 on Build GRCh38
rs769189456 Chr.3: 51075410-51075410 on Build GRCh38
rs777749638 Chr.3: 51075405-51075405 on Build GRCh38

View Full Product Details