Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73777929-73778507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP11
Entrez Gene ID
8446
Gene Name
dual specificity phosphatase 11
Gene Aliases
PIR1
UniGene
Hs.14611
Gene Chromosome Location
Chr.2: 73762184-73780157 on Build GRCh38
Transcripts
NM_003584.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5928266 c.136C>T p.P46S Chr.2: 73778342-73778342 on Build GRCh38
5928265 c.277C>T p.P93S Chr.2: 73778342-73778342 on Build GRCh38
5476395 c.271C>T p.R91W Chr.2: 73778348-73778348 on Build GRCh38
7206194 c.146G>A p.R49H Chr.2: 73778332-73778332 on Build GRCh38
3714311 c.113A>G p.Y38C Chr.2: 73778365-73778365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913678019 Chr.2: 73778234-73778234 on Build GRCh38
rs1014193960 Chr.2: 73778096-73778096 on Build GRCh38
rs757832879 Chr.2: 73778333-73778333 on Build GRCh38
rs774408546 Chr.2: 73778254-73778254 on Build GRCh38
rs61757364 Chr.2: 73778299-73778299 on Build GRCh38
rs750836592 Chr.2: 73778230-73778231 on Build GRCh38
rs995118865 Chr.2: 73778183-73778183 on Build GRCh38
rs1036362768 Chr.2: 73778392-73778392 on Build GRCh38
rs61758449 Chr.2: 73778296-73778296 on Build GRCh38
rs1026267740 Chr.2: 73778108-73778108 on Build GRCh38
rs752337805 Chr.2: 73778277-73778277 on Build GRCh38
rs987814141 Chr.2: 73778224-73778224 on Build GRCh38
rs1018547208 Chr.2: 73778198-73778198 on Build GRCh38
rs3738850 Chr.2: 73778389-73778389 on Build GRCh38
rs1008408956 Chr.2: 73778455-73778455 on Build GRCh38
rs1001505350 Chr.2: 73778087-73778087 on Build GRCh38
rs1052291036 Chr.2: 73778001-73778001 on Build GRCh38
rs917706660 Chr.2: 73778260-73778260 on Build GRCh38
rs558619375 Chr.2: 73778018-73778018 on Build GRCh38
rs552263243 Chr.2: 73778467-73778467 on Build GRCh38
rs1043225382 Chr.2: 73777999-73777999 on Build GRCh38
rs987982718 Chr.2: 73778258-73778258 on Build GRCh38
rs1055997490 Chr.2: 73778284-73778284 on Build GRCh38
rs994294696 Chr.2: 73778101-73778101 on Build GRCh38
rs1015451858 Chr.2: 73778116-73778116 on Build GRCh38
rs375284952 Chr.2: 73778241-73778241 on Build GRCh38
rs867471412 Chr.2: 73778287-73778287 on Build GRCh38
rs898610613 Chr.2: 73778098-73778098 on Build GRCh38
rs762167554 Chr.2: 73777958-73777958 on Build GRCh38
rs569186814 Chr.2: 73777992-73777992 on Build GRCh38
rs781726706 Chr.2: 73778410-73778410 on Build GRCh38
rs770932773 Chr.2: 73778379-73778379 on Build GRCh38
rs756633153 Chr.2: 73778424-73778424 on Build GRCh38
rs1045359994 Chr.2: 73778336-73778336 on Build GRCh38
rs967782090 Chr.2: 73778236-73778236 on Build GRCh38
rs761082658 Chr.2: 73778332-73778332 on Build GRCh38
rs763816897 Chr.2: 73778393-73778393 on Build GRCh38
rs750840451 Chr.2: 73778417-73778417 on Build GRCh38
rs540779010 Chr.2: 73778372-73778372 on Build GRCh38
rs905817966 Chr.2: 73778065-73778065 on Build GRCh38
rs376099907 Chr.2: 73778439-73778439 on Build GRCh38
rs567555052 Chr.2: 73778183-73778183 on Build GRCh38
rs1055737331 Chr.2: 73778057-73778057 on Build GRCh38
rs749572997 Chr.2: 73778361-73778361 on Build GRCh38
rs766200711 Chr.2: 73778257-73778257 on Build GRCh38
rs566467982 Chr.2: 73778191-73778191 on Build GRCh38
rs949360410 Chr.2: 73778026-73778026 on Build GRCh38
rs768832383 Chr.2: 73778369-73778369 on Build GRCh38
rs965731202 Chr.2: 73778194-73778194 on Build GRCh38
rs200053499 Chr.2: 73778395-73778395 on Build GRCh38
rs780270678 Chr.2: 73778358-73778358 on Build GRCh38
rs763502513 Chr.2: 73778265-73778265 on Build GRCh38
rs756686336 Chr.2: 73778407-73778407 on Build GRCh38
rs760703800 Chr.2: 73778256-73778256 on Build GRCh38
rs769989768 Chr.2: 73778348-73778348 on Build GRCh38
rs963679553 Chr.2: 73778219-73778219 on Build GRCh38
rs567302039 Chr.2: 73778184-73778184 on Build GRCh38
rs1038374355 Chr.2: 73778377-73778377 on Build GRCh38
rs953287699 Chr.2: 73778187-73778187 on Build GRCh38
rs937559185 Chr.2: 73778027-73778027 on Build GRCh38
rs897901988 Chr.2: 73778153-73778153 on Build GRCh38
rs758107996 Chr.2: 73778281-73778281 on Build GRCh38
rs895799576 Chr.2: 73778089-73778089 on Build GRCh38
rs1018855312 Chr.2: 73778193-73778193 on Build GRCh38
rs762735685 Chr.2: 73778390-73778390 on Build GRCh38
rs557296939 Chr.2: 73777963-73777963 on Build GRCh38
rs1044994869 Chr.2: 73778063-73778063 on Build GRCh38
rs746279320 Chr.2: 73778347-73778347 on Build GRCh38
rs759212007 Chr.2: 73778269-73778269 on Build GRCh38
rs891265122 Chr.2: 73778396-73778396 on Build GRCh38
rs774459988 Chr.2: 73778370-73778370 on Build GRCh38
rs935137403 Chr.2: 73777996-73777996 on Build GRCh38
rs979346257 Chr.2: 73778249-73778249 on Build GRCh38
rs754244295 Chr.2: 73778402-73778402 on Build GRCh38
rs139537978 Chr.2: 73778276-73778276 on Build GRCh38
rs372146613 Chr.2: 73778384-73778384 on Build GRCh38
rs201327566 Chr.2: 73778387-73778387 on Build GRCh38
rs146297318 Chr.2: 73777993-73777993 on Build GRCh38
rs753685568 Chr.2: 73778263-73778263 on Build GRCh38
rs533914328 Chr.2: 73778207-73778208 on Build GRCh38
rs765180107 Chr.2: 73778388-73778388 on Build GRCh38
rs781502977 Chr.2: 73778345-73778345 on Build GRCh38

View Full Product Details