Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 110641916-110642356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUB1
Entrez Gene ID
699
Gene Name
BUB1 mitotic checkpoint serine/threonine kinase
Gene Aliases
BUB1A, BUB1L, hBUB1
UniGene
Hs.469649
Gene Chromosome Location
Chr.2: 110637698-110678107 on Build GRCh38
Transcripts
NM_001278616.1 NM_001278617.1 NM_004336.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1005367 c.2452T>G p.F818V Chr.2: 110642130-110642130 on Build GRCh38
2982984 c.2440A>C p.N814H Chr.2: 110642142-110642142 on Build GRCh38
6323738 c.2358G>A p.L786L Chr.2: 110642224-110642224 on Build GRCh38
5524651 c.2349T>G p.G783G Chr.2: 110642233-110642233 on Build GRCh38
6613639 c.2391C>T p.A797A Chr.2: 110642191-110642191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs929317381 Chr.2: 110641997-110641997 on Build GRCh38
rs747195242 Chr.2: 110642055-110642055 on Build GRCh38
rs766243053 Chr.2: 110642111-110642111 on Build GRCh38
rs748836973 Chr.2: 110642272-110642276 on Build GRCh38
rs376808276 Chr.2: 110642237-110642237 on Build GRCh38
rs369687502 Chr.2: 110641979-110641979 on Build GRCh38
rs149429986 Chr.2: 110642209-110642209 on Build GRCh38
rs758161358 Chr.2: 110642276-110642281 on Build GRCh38
rs774922803 Chr.2: 110642196-110642196 on Build GRCh38
rs539595216 Chr.2: 110642132-110642132 on Build GRCh38
rs771426020 Chr.2: 110642179-110642179 on Build GRCh38
rs769693608 Chr.2: 110642317-110642317 on Build GRCh38
rs762664403 Chr.2: 110642247-110642247 on Build GRCh38
rs940424193 Chr.2: 110642187-110642187 on Build GRCh38
rs761502719 Chr.2: 110642075-110642075 on Build GRCh38
rs776173182 Chr.2: 110642228-110642228 on Build GRCh38
rs751407971 Chr.2: 110642255-110642255 on Build GRCh38
rs781312045 Chr.2: 110642136-110642136 on Build GRCh38
rs777879454 Chr.2: 110641972-110641972 on Build GRCh38
rs941252699 Chr.2: 110642031-110642031 on Build GRCh38
rs367877199 Chr.2: 110642086-110642086 on Build GRCh38
rs781034240 Chr.2: 110642108-110642108 on Build GRCh38
rs371721678 Chr.2: 110642176-110642176 on Build GRCh38
rs868404865 Chr.2: 110642124-110642124 on Build GRCh38
rs997999558 Chr.2: 110642052-110642052 on Build GRCh38
rs766152699 Chr.2: 110642253-110642253 on Build GRCh38
rs750176266 Chr.2: 110642087-110642087 on Build GRCh38
rs888375715 Chr.2: 110642025-110642025 on Build GRCh38
rs761257796 Chr.2: 110642235-110642235 on Build GRCh38
rs577822032 Chr.2: 110642016-110642016 on Build GRCh38
rs984620829 Chr.2: 110642040-110642040 on Build GRCh38
rs553324831 Chr.2: 110642289-110642289 on Build GRCh38
rs746371172 Chr.2: 110642197-110642197 on Build GRCh38
rs367545797 Chr.2: 110642164-110642164 on Build GRCh38
rs779946777 Chr.2: 110642203-110642205 on Build GRCh38
rs766008602 Chr.2: 110642283-110642284 on Build GRCh38
rs200385130 Chr.2: 110642175-110642175 on Build GRCh38
rs754932284 Chr.2: 110642118-110642118 on Build GRCh38
rs1039598735 Chr.2: 110642035-110642035 on Build GRCh38
rs957657111 Chr.2: 110642112-110642112 on Build GRCh38
rs1049016929 Chr.2: 110642010-110642010 on Build GRCh38
rs758207639 Chr.2: 110642095-110642095 on Build GRCh38
rs756161575 Chr.2: 110642280-110642280 on Build GRCh38
rs72942112 Chr.2: 110641938-110641938 on Build GRCh38
rs767387668 Chr.2: 110642268-110642268 on Build GRCh38
rs772860601 Chr.2: 110642238-110642238 on Build GRCh38
rs919320174 Chr.2: 110641973-110641973 on Build GRCh38
rs748219999 Chr.2: 110642160-110642160 on Build GRCh38
rs765280229 Chr.2: 110642133-110642134 on Build GRCh38
rs752775900 Chr.2: 110642269-110642269 on Build GRCh38
rs751498003 Chr.2: 110642114-110642114 on Build GRCh38
rs570878972 Chr.2: 110642042-110642042 on Build GRCh38
rs754915797 Chr.2: 110642266-110642266 on Build GRCh38

View Full Product Details