Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 177434101-177434579 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPS
Entrez Gene ID
8540
Gene Name
alkylglycerone phosphate synthase
Gene Aliases
ADAP-S, ADAS, ADHAPS, ADPS, ALDHPSY, RCDP3
UniGene
Hs.516543
Gene Chromosome Location
Chr.2: 177392743-177543836 on Build GRCh38
Transcripts
NM_003659.3 XM_011512041.2 XM_011512042.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6245648 c.355C>A p.P119T Chr.2: 177434331-177434331 on Build GRCh38
4453970 c.379A>G p.T127A Chr.2: 177434355-177434355 on Build GRCh38
6381305 c.430A>G p.T144A Chr.2: 177434406-177434406 on Build GRCh38
2894790 c.423G>A p.E141E Chr.2: 177434399-177434399 on Build GRCh38
1401477 c.402T>C p.N134N Chr.2: 177434378-177434378 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959431346 Chr.2: 177434424-177434424 on Build GRCh38
rs969799314 Chr.2: 177434314-177434314 on Build GRCh38
rs903839612 Chr.2: 177434212-177434212 on Build GRCh38
rs148111927 Chr.2: 177434494-177434494 on Build GRCh38
rs774373023 Chr.2: 177434402-177434402 on Build GRCh38
rs536406455 Chr.2: 177434385-177434385 on Build GRCh38
rs1024951809 Chr.2: 177434475-177434479 on Build GRCh38
rs907790983 Chr.2: 177434489-177434489 on Build GRCh38
rs766950054 Chr.2: 177434291-177434291 on Build GRCh38
rs202122306 Chr.2: 177434142-177434142 on Build GRCh38
rs1031249463 Chr.2: 177434248-177434248 on Build GRCh38
rs747488115 Chr.2: 177434373-177434373 on Build GRCh38
rs748300962 Chr.2: 177434390-177434390 on Build GRCh38
rs143730371 Chr.2: 177434269-177434269 on Build GRCh38
rs200594024 Chr.2: 177434330-177434330 on Build GRCh38
rs377177736 Chr.2: 177434303-177434303 on Build GRCh38
rs78131839 Chr.2: 177434193-177434193 on Build GRCh38
rs573032165 Chr.2: 177434546-177434546 on Build GRCh38
rs370074934 Chr.2: 177434322-177434322 on Build GRCh38
rs769887090 Chr.2: 177434394-177434394 on Build GRCh38
rs111382938 Chr.2: 177434129-177434129 on Build GRCh38
rs368286128 Chr.2: 177434416-177434416 on Build GRCh38
rs754114692 Chr.2: 177434287-177434287 on Build GRCh38
rs750397893 Chr.2: 177434196-177434196 on Build GRCh38
rs755713066 Chr.2: 177434317-177434317 on Build GRCh38
rs944512431 Chr.2: 177434125-177434125 on Build GRCh38
rs557061139 Chr.2: 177434289-177434289 on Build GRCh38
rs143834538 Chr.2: 177434551-177434551 on Build GRCh38
rs780475043 Chr.2: 177434336-177434336 on Build GRCh38
rs920285676 Chr.2: 177434380-177434380 on Build GRCh38
rs758795768 Chr.2: 177434333-177434333 on Build GRCh38
rs1000209265 Chr.2: 177434246-177434246 on Build GRCh38
rs879722367 Chr.2: 177434522-177434522 on Build GRCh38
rs983815847 Chr.2: 177434475-177434475 on Build GRCh38
rs773588383 Chr.2: 177434398-177434398 on Build GRCh38
rs144836649 Chr.2: 177434401-177434401 on Build GRCh38
rs552881569 Chr.2: 177434233-177434233 on Build GRCh38
rs76827775 Chr.2: 177434133-177434133 on Build GRCh38
rs879080371 Chr.2: 177434480-177434480 on Build GRCh38
rs571134475 Chr.2: 177434237-177434237 on Build GRCh38
rs760279900 Chr.2: 177434469-177434469 on Build GRCh38
rs1040248230 Chr.2: 177434535-177434535 on Build GRCh38
rs767554611 Chr.2: 177434433-177434433 on Build GRCh38
rs769268056 Chr.2: 177434379-177434379 on Build GRCh38
rs769202875 Chr.2: 177434321-177434322 on Build GRCh38
rs752115173 Chr.2: 177434435-177434435 on Build GRCh38
rs145828613 Chr.2: 177434160-177434160 on Build GRCh38

View Full Product Details