Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 222922584-222923156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL3
Entrez Gene ID
2181
Gene Name
acyl-CoA synthetase long-chain family member 3
Gene Aliases
ACS3, FACL3, PRO2194
UniGene
Hs.655772
Gene Chromosome Location
Chr.2: 222860984-222944624 on Build GRCh38
Transcripts
NM_004457.3 NM_203372.1 XM_017003584.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5904136 c.1081-1G>A p.? Chr.2: 222923077-222923077 on Build GRCh38
4991707 c.1103G>T p.S368I Chr.2: 222923100-222923100 on Build GRCh38
242786 c.1082C>T p.S361F Chr.2: 222923079-222923079 on Build GRCh38
278599 c.1087A>C p.K363Q Chr.2: 222923084-222923084 on Build GRCh38
4460038 c.1150C>T p.P384S Chr.2: 222923147-222923147 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748611651 Chr.2: 222922826-222922826 on Build GRCh38
rs1006829142 Chr.2: 222923115-222923115 on Build GRCh38
rs997549859 Chr.2: 222923058-222923058 on Build GRCh38
rs375798869 Chr.2: 222923074-222923074 on Build GRCh38
rs762419923 Chr.2: 222923047-222923048 on Build GRCh38
rs754721929 Chr.2: 222922666-222922666 on Build GRCh38
rs775251003 Chr.2: 222923114-222923114 on Build GRCh38
rs775804214 Chr.2: 222922693-222922693 on Build GRCh38
rs772524713 Chr.2: 222923032-222923032 on Build GRCh38
rs139999422 Chr.2: 222922794-222922794 on Build GRCh38
rs751579992 Chr.2: 222922869-222922869 on Build GRCh38
rs918204534 Chr.2: 222922902-222922902 on Build GRCh38
rs754516691 Chr.2: 222922966-222922966 on Build GRCh38
rs752282840 Chr.2: 222922847-222922847 on Build GRCh38
rs765899054 Chr.2: 222922753-222922753 on Build GRCh38
rs758127014 Chr.2: 222922782-222922782 on Build GRCh38
rs763951943 Chr.2: 222922861-222922861 on Build GRCh38
rs900970563 Chr.2: 222923021-222923021 on Build GRCh38
rs962840854 Chr.2: 222922829-222922829 on Build GRCh38
rs764004849 Chr.2: 222923130-222923130 on Build GRCh38
rs112352030 Chr.2: 222923056-222923056 on Build GRCh38
rs768200414 Chr.2: 222923018-222923018 on Build GRCh38
rs566961930 Chr.2: 222922982-222922982 on Build GRCh38
rs748007632 Chr.2: 222922668-222922668 on Build GRCh38
rs754703157 Chr.2: 222922771-222922771 on Build GRCh38
rs535801552 Chr.2: 222923023-222923023 on Build GRCh38
rs752787783 Chr.2: 222922881-222922881 on Build GRCh38
rs753504707 Chr.2: 222922664-222922664 on Build GRCh38
rs755760746 Chr.2: 222922658-222922658 on Build GRCh38
rs746924445 Chr.2: 222922793-222922793 on Build GRCh38
rs749323299 Chr.2: 222922875-222922875 on Build GRCh38
rs200158558 Chr.2: 222922796-222922796 on Build GRCh38
rs771365458 Chr.2: 222923064-222923064 on Build GRCh38
rs776996984 Chr.2: 222923040-222923053 on Build GRCh38
rs544850987 Chr.2: 222922610-222922610 on Build GRCh38
rs763298438 Chr.2: 222922699-222922699 on Build GRCh38
rs752473823 Chr.2: 222922777-222922777 on Build GRCh38
rs768137676 Chr.2: 222922724-222922724 on Build GRCh38
rs750935029 Chr.2: 222922739-222922739 on Build GRCh38
rs756205933 Chr.2: 222922917-222922917 on Build GRCh38
rs528004115 Chr.2: 222922958-222922958 on Build GRCh38
rs139854827 Chr.2: 222922765-222922765 on Build GRCh38
rs762475796 Chr.2: 222923117-222923117 on Build GRCh38
rs755244315 Chr.2: 222922747-222922747 on Build GRCh38
rs758266071 Chr.2: 222922851-222922851 on Build GRCh38
rs780295457 Chr.2: 222922873-222922873 on Build GRCh38
rs371238819 Chr.2: 222922667-222922667 on Build GRCh38
rs555864781 Chr.2: 222923061-222923061 on Build GRCh38
rs764633563 Chr.2: 222923100-222923100 on Build GRCh38
rs10932978 Chr.2: 222922973-222922973 on Build GRCh38
rs781123829 Chr.2: 222922673-222922673 on Build GRCh38
rs370161161 Chr.2: 222922831-222922831 on Build GRCh38
rs994022258 Chr.2: 222923050-222923050 on Build GRCh38
rs1025703687 Chr.2: 222923052-222923052 on Build GRCh38
rs768870128 Chr.2: 222922701-222922701 on Build GRCh38
rs948311747 Chr.2: 222922914-222922914 on Build GRCh38
rs371846611 Chr.2: 222922883-222922883 on Build GRCh38
rs377743525 Chr.2: 222922811-222922811 on Build GRCh38
rs368892612 Chr.2: 222923075-222923075 on Build GRCh38
rs200932353 Chr.2: 222922661-222922661 on Build GRCh38
rs991784665 Chr.2: 222922885-222922885 on Build GRCh38
rs75810699 Chr.2: 222922735-222922735 on Build GRCh38
rs755007755 Chr.2: 222922880-222922880 on Build GRCh38
rs745873011 Chr.2: 222922689-222922689 on Build GRCh38
rs541312035 Chr.2: 222922645-222922645 on Build GRCh38
rs543153547 Chr.2: 222922804-222922804 on Build GRCh38
rs762214053 Chr.2: 222922714-222922714 on Build GRCh38
rs777827095 Chr.2: 222922786-222922786 on Build GRCh38
rs778554898 Chr.2: 222922775-222922775 on Build GRCh38
rs1045229381 Chr.2: 222922616-222922616 on Build GRCh38
rs1054082764 Chr.2: 222922606-222922606 on Build GRCh38
rs572285434 Chr.2: 222922619-222922619 on Build GRCh38
rs1052522089 Chr.2: 222922996-222922996 on Build GRCh38
rs773522343 Chr.2: 222923053-222923053 on Build GRCh38
rs765011836 Chr.2: 222922844-222922844 on Build GRCh38
rs751158518 Chr.2: 222922923-222922923 on Build GRCh38
rs1007243543 Chr.2: 222922629-222922629 on Build GRCh38
rs759147663 Chr.2: 222923095-222923095 on Build GRCh38
rs558831982 Chr.2: 222922614-222922614 on Build GRCh38
rs141829448 Chr.2: 222922690-222922690 on Build GRCh38
rs759119779 Chr.2: 222922843-222922843 on Build GRCh38
rs531953984 Chr.2: 222922830-222922830 on Build GRCh38
rs927313197 Chr.2: 222922772-222922772 on Build GRCh38
rs202245769 Chr.2: 222922813-222922813 on Build GRCh38
rs772100177 Chr.2: 222922670-222922670 on Build GRCh38
rs116164403 Chr.2: 222922870-222922870 on Build GRCh38
rs946226410 Chr.2: 222922815-222922815 on Build GRCh38
rs866487232 Chr.2: 222922918-222922918 on Build GRCh38
rs11891410 Chr.2: 222922841-222922841 on Build GRCh38

View Full Product Details