Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 90406453-90407015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALM1
Entrez Gene ID
801
Gene Name
calmodulin 1
Gene Aliases
CALML2, CAMI, CPVT4, DD132, LQT14, PHKD, caM
UniGene
Hs.282410
Gene Chromosome Location
Chr.14: 90396983-90408275 on Build GRCh38
Transcripts
NM_006888.4 XM_006720258.3

SNP Detail

SNP ID Chromosome Location
rs552878782 Chr.14: 90406845-90406845 on Build GRCh38
rs1033969455 Chr.14: 90406866-90406866 on Build GRCh38
rs889083320 Chr.14: 90406501-90406501 on Build GRCh38
rs976507853 Chr.14: 90406981-90406981 on Build GRCh38
rs920402748 Chr.14: 90406722-90406722 on Build GRCh38
rs1040476221 Chr.14: 90406535-90406535 on Build GRCh38
rs35418035 Chr.14: 90406587-90406587 on Build GRCh38
rs1025053838 Chr.14: 90406831-90406831 on Build GRCh38
rs559970665 Chr.14: 90406572-90406572 on Build GRCh38
rs530199227 Chr.14: 90406551-90406551 on Build GRCh38
rs538040957 Chr.14: 90406705-90406705 on Build GRCh38
rs754281618 Chr.14: 90406595-90406595 on Build GRCh38
rs765709922 Chr.14: 90406695-90406695 on Build GRCh38
rs959416813 Chr.14: 90406573-90406575 on Build GRCh38
rs981797700 Chr.14: 90406634-90406634 on Build GRCh38
rs766898078 Chr.14: 90406905-90406905 on Build GRCh38
rs184371119 Chr.14: 90406935-90406935 on Build GRCh38
rs1038757399 Chr.14: 90406704-90406704 on Build GRCh38
rs930571066 Chr.14: 90406724-90406724 on Build GRCh38
rs553357792 Chr.14: 90406827-90406827 on Build GRCh38
rs923007518 Chr.14: 90406911-90406911 on Build GRCh38
rs575394798 Chr.14: 90406852-90406852 on Build GRCh38
rs886671594 Chr.14: 90406769-90406769 on Build GRCh38
rs181955035 Chr.14: 90406968-90406968 on Build GRCh38
rs539419419 Chr.14: 90406814-90406814 on Build GRCh38
rs1002417158 Chr.14: 90406605-90406605 on Build GRCh38
rs955952670 Chr.14: 90406869-90406869 on Build GRCh38
rs772899629 Chr.14: 90406702-90406704 on Build GRCh38
rs535805623 Chr.14: 90406583-90406583 on Build GRCh38
rs751584532 Chr.14: 90406910-90406910 on Build GRCh38
rs895300982 Chr.14: 90406804-90406804 on Build GRCh38
rs958392170 Chr.14: 90406644-90406644 on Build GRCh38
rs777819916 Chr.14: 90406899-90406899 on Build GRCh38
rs555446195 Chr.14: 90406849-90406849 on Build GRCh38
rs1006350838 Chr.14: 90406570-90406572 on Build GRCh38
rs892557783 Chr.14: 90406816-90406816 on Build GRCh38
rs374437556 Chr.14: 90406666-90406666 on Build GRCh38
rs1034022829 Chr.14: 90406632-90406632 on Build GRCh38
rs149760583 Chr.14: 90406843-90406843 on Build GRCh38
rs964506438 Chr.14: 90406909-90406909 on Build GRCh38
rs189778902 Chr.14: 90406920-90406920 on Build GRCh38
rs184803743 Chr.14: 90406802-90406802 on Build GRCh38
rs370139244 Chr.14: 90406853-90406853 on Build GRCh38
rs549051896 Chr.14: 90406985-90406985 on Build GRCh38
rs938351498 Chr.14: 90406925-90406925 on Build GRCh38
rs992490286 Chr.14: 90406658-90406658 on Build GRCh38
rs942285111 Chr.14: 90406942-90406942 on Build GRCh38
rs186848664 Chr.14: 90406559-90406559 on Build GRCh38
rs555800715 Chr.14: 90406886-90406886 on Build GRCh38
rs866590730 Chr.14: 90406518-90406518 on Build GRCh38
rs369706187 Chr.14: 90406556-90406556 on Build GRCh38
rs1026559753 Chr.14: 90406557-90406557 on Build GRCh38
rs961794722 Chr.14: 90406675-90406675 on Build GRCh38
rs1013944641 Chr.14: 90406893-90406893 on Build GRCh38
rs139692985 Chr.14: 90406681-90406681 on Build GRCh38
rs1006409192 Chr.14: 90406780-90406780 on Build GRCh38
rs944701803 Chr.14: 90406506-90406506 on Build GRCh38
rs1002363400 Chr.14: 90406856-90406856 on Build GRCh38
rs374154318 Chr.14: 90406880-90406881 on Build GRCh38
rs773056073 Chr.14: 90406545-90406545 on Build GRCh38
rs3814843 Chr.14: 90406669-90406669 on Build GRCh38
rs905583067 Chr.14: 90406855-90406855 on Build GRCh38
rs367855711 Chr.14: 90406956-90406956 on Build GRCh38
rs560712781 Chr.14: 90406586-90406589 on Build GRCh38
rs564452026 Chr.14: 90406478-90406478 on Build GRCh38
rs924517041 Chr.14: 90406649-90406649 on Build GRCh38
rs998216494 Chr.14: 90406546-90406547 on Build GRCh38
rs951921689 Chr.14: 90406955-90406955 on Build GRCh38
rs181805019 Chr.14: 90406928-90406928 on Build GRCh38
rs532760054 Chr.14: 90406491-90406491 on Build GRCh38
rs192452102 Chr.14: 90406733-90406733 on Build GRCh38
rs910467861 Chr.14: 90406743-90406743 on Build GRCh38
rs189147924 Chr.14: 90406967-90406967 on Build GRCh38
rs1010178768 Chr.14: 90406575-90406575 on Build GRCh38
rs1046918253 Chr.14: 90406985-90406986 on Build GRCh38

View Full Product Details