Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 51448204-51448745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMXL2
Entrez Gene ID
23312
Gene Name
Dmx like 2
Gene Aliases
PEPNS, RC3
UniGene
Hs.511386
Gene Chromosome Location
Chr.15: 51447724-51622833 on Build GRCh38
Transcripts
NM_001174116.1 NM_001174117.1 NM_015263.3 XM_005254255.1 XM_005254256.1 XM_017022034.1

SNP Detail

SNP ID Chromosome Location
rs982583963 Chr.15: 51448439-51448439 on Build GRCh38
rs1014992922 Chr.15: 51448570-51448570 on Build GRCh38
rs912885474 Chr.15: 51448408-51448408 on Build GRCh38
rs371704325 Chr.15: 51448338-51448338 on Build GRCh38
rs375021789 Chr.15: 51448673-51448673 on Build GRCh38
rs543909290 Chr.15: 51448624-51448624 on Build GRCh38
rs932134219 Chr.15: 51448502-51448502 on Build GRCh38
rs551046592 Chr.15: 51448293-51448293 on Build GRCh38
rs890658429 Chr.15: 51448641-51448641 on Build GRCh38
rs554926062 Chr.15: 51448499-51448499 on Build GRCh38
rs934791915 Chr.15: 51448592-51448592 on Build GRCh38
rs1053323665 Chr.15: 51448291-51448291 on Build GRCh38
rs557542675 Chr.15: 51448382-51448382 on Build GRCh38
rs546376560 Chr.15: 51448653-51448653 on Build GRCh38
rs749772723 Chr.15: 51448367-51448367 on Build GRCh38
rs180672401 Chr.15: 51448259-51448259 on Build GRCh38
rs1008869971 Chr.15: 51448252-51448252 on Build GRCh38
rs573317184 Chr.15: 51448503-51448503 on Build GRCh38
rs915545123 Chr.15: 51448540-51448540 on Build GRCh38
rs960245215 Chr.15: 51448461-51448461 on Build GRCh38
rs972613287 Chr.15: 51448295-51448295 on Build GRCh38
rs897824199 Chr.15: 51448607-51448607 on Build GRCh38
rs952575298 Chr.15: 51448320-51448320 on Build GRCh38
rs533855155 Chr.15: 51448479-51448479 on Build GRCh38
rs137899703 Chr.15: 51448674-51448674 on Build GRCh38
rs796468020 Chr.15: 51448352-51448353 on Build GRCh38
rs761455531 Chr.15: 51448545-51448545 on Build GRCh38
rs925694226 Chr.15: 51448590-51448590 on Build GRCh38
rs904690072 Chr.15: 51448276-51448276 on Build GRCh38
rs948438025 Chr.15: 51448234-51448234 on Build GRCh38
rs1039324966 Chr.15: 51448693-51448693 on Build GRCh38
rs373030881 Chr.15: 51448353-51448354 on Build GRCh38
rs12440288 Chr.15: 51448520-51448520 on Build GRCh38
rs934210444 Chr.15: 51448541-51448541 on Build GRCh38
rs952515555 Chr.15: 51448435-51448435 on Build GRCh38
rs1014894628 Chr.15: 51448267-51448267 on Build GRCh38
rs531917398 Chr.15: 51448469-51448469 on Build GRCh38
rs370444907 Chr.15: 51448647-51448647 on Build GRCh38
rs904774855 Chr.15: 51448501-51448501 on Build GRCh38
rs572766875 Chr.15: 51448465-51448465 on Build GRCh38
rs569299934 Chr.15: 51448345-51448345 on Build GRCh38
rs979931082 Chr.15: 51448676-51448676 on Build GRCh38
rs767129194 Chr.15: 51448629-51448630 on Build GRCh38
rs10654455 Chr.15: 51448548-51448549 on Build GRCh38
rs561509742 Chr.15: 51448379-51448379 on Build GRCh38
rs147387782 Chr.15: 51448649-51448649 on Build GRCh38
rs869159414 Chr.15: 51448549-51448550 on Build GRCh38
rs948408328 Chr.15: 51448542-51448542 on Build GRCh38
rs908434746 Chr.15: 51448459-51448459 on Build GRCh38
rs1043324472 Chr.15: 51448497-51448497 on Build GRCh38
rs890520645 Chr.15: 51448535-51448535 on Build GRCh38
rs767143430 Chr.15: 51448618-51448618 on Build GRCh38
rs771392474 Chr.15: 51448413-51448413 on Build GRCh38
rs919879954 Chr.15: 51448311-51448311 on Build GRCh38
rs186444496 Chr.15: 51448355-51448355 on Build GRCh38
rs535518224 Chr.15: 51448286-51448286 on Build GRCh38
rs573566133 Chr.15: 51448543-51448543 on Build GRCh38

View Full Product Details