Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121330387-121330944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC5
Entrez Gene ID
51433
Gene Name
anaphase promoting complex subunit 5
Gene Aliases
APC5
UniGene
Hs.7101
Gene Chromosome Location
Chr.12: 121308245-121354209 on Build GRCh38
Transcripts
NM_001137559.1 NM_016237.4 XM_005253900.3 XM_017019423.1 XM_017019424.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7166805 c.1121C>T p.P374L Chr.12: 121330584-121330584 on Build GRCh38
6599570 c.1035C>T p.S345S Chr.12: 121330670-121330670 on Build GRCh38
25303 c.1091_1118del28 p.S364fs*26 Chr.12: 121330587-121330614 on Build GRCh38
7290524 c.1097A>T p.K366M Chr.12: 121330608-121330608 on Build GRCh38
5966559 c.1102G>T p.A368S Chr.12: 121330603-121330603 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782154458 Chr.12: 121330718-121330718 on Build GRCh38
rs534294635 Chr.12: 121330780-121330780 on Build GRCh38
rs369753396 Chr.12: 121330689-121330689 on Build GRCh38
rs782059026 Chr.12: 121330702-121330702 on Build GRCh38
rs1055013633 Chr.12: 121330597-121330597 on Build GRCh38
rs540659440 Chr.12: 121330449-121330449 on Build GRCh38
rs376710093 Chr.12: 121330656-121330656 on Build GRCh38
rs372504555 Chr.12: 121330703-121330703 on Build GRCh38
rs550818210 Chr.12: 121330499-121330499 on Build GRCh38
rs782005793 Chr.12: 121330551-121330551 on Build GRCh38
rs782502655 Chr.12: 121330612-121330612 on Build GRCh38
rs781986829 Chr.12: 121330670-121330670 on Build GRCh38
rs782595703 Chr.12: 121330651-121330651 on Build GRCh38
rs782012994 Chr.12: 121330705-121330705 on Build GRCh38
rs114635317 Chr.12: 121330608-121330608 on Build GRCh38
rs782647462 Chr.12: 121330637-121330637 on Build GRCh38
rs552581046 Chr.12: 121330790-121330790 on Build GRCh38
rs782753405 Chr.12: 121330567-121330567 on Build GRCh38
rs782763427 Chr.12: 121330515-121330515 on Build GRCh38
rs149057486 Chr.12: 121330640-121330640 on Build GRCh38
rs951778731 Chr.12: 121330446-121330446 on Build GRCh38
rs377332140 Chr.12: 121330909-121330909 on Build GRCh38
rs782488179 Chr.12: 121330643-121330643 on Build GRCh38
rs561034773 Chr.12: 121330421-121330421 on Build GRCh38
rs781865469 Chr.12: 121330740-121330740 on Build GRCh38
rs782092193 Chr.12: 121330552-121330552 on Build GRCh38
rs907290763 Chr.12: 121330696-121330696 on Build GRCh38
rs1037049970 Chr.12: 121330789-121330789 on Build GRCh38
rs1047067700 Chr.12: 121330606-121330606 on Build GRCh38
rs187119656 Chr.12: 121330834-121330834 on Build GRCh38
rs565363127 Chr.12: 121330468-121330468 on Build GRCh38
rs370468197 Chr.12: 121330498-121330498 on Build GRCh38
rs985879899 Chr.12: 121330478-121330478 on Build GRCh38
rs782438051 Chr.12: 121330667-121330667 on Build GRCh38
rs182826414 Chr.12: 121330680-121330680 on Build GRCh38
rs782773565 Chr.12: 121330739-121330739 on Build GRCh38
rs113139253 Chr.12: 121330819-121330819 on Build GRCh38
rs372451754 Chr.12: 121330598-121330598 on Build GRCh38
rs116135448 Chr.12: 121330632-121330632 on Build GRCh38
rs910209645 Chr.12: 121330488-121330488 on Build GRCh38
rs191636855 Chr.12: 121330883-121330883 on Build GRCh38
rs781974176 Chr.12: 121330699-121330699 on Build GRCh38
rs782121964 Chr.12: 121330542-121330542 on Build GRCh38
rs781783544 Chr.12: 121330571-121330571 on Build GRCh38
rs2280067 Chr.12: 121330445-121330445 on Build GRCh38
rs782744057 Chr.12: 121330742-121330742 on Build GRCh38
rs975776040 Chr.12: 121330514-121330514 on Build GRCh38
rs201038764 Chr.12: 121330584-121330584 on Build GRCh38

View Full Product Details