Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 82990119-82990713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf40
Entrez Gene ID
123207
Gene Name
chromosome 15 open reading frame 40
Gene Aliases
-
UniGene
Hs.352541
Gene Chromosome Location
Chr.15: 82988297-83012298 on Build GRCh38
Transcripts
NM_001160115.1 NM_001160116.1 XM_011521213.2 NM_031452.3

Gene Symbol
FAM103A1
Entrez Gene ID
83640
Gene Name
family with sequence similarity 103 member A1
Gene Aliases
C15orf18, HsT19360, RAM
UniGene
Hs.80624
Gene Chromosome Location
Chr.15: 82986203-82991057 on Build GRCh38
Transcripts
NM_001160115.1 NM_001160116.1 XM_011521213.2 NM_031452.3

SNP Detail

SNP ID Chromosome Location
rs397854504 Chr.15: 82990557-82990558 on Build GRCh38
rs928671212 Chr.15: 82990585-82990585 on Build GRCh38
rs897393287 Chr.15: 82990483-82990483 on Build GRCh38
rs543329557 Chr.15: 82990354-82990354 on Build GRCh38
rs538049575 Chr.15: 82990674-82990676 on Build GRCh38
rs997463145 Chr.15: 82990300-82990300 on Build GRCh38
rs1038978511 Chr.15: 82990464-82990464 on Build GRCh38
rs903609204 Chr.15: 82990146-82990146 on Build GRCh38
rs747383732 Chr.15: 82990630-82990631 on Build GRCh38
rs371615072 Chr.15: 82990604-82990604 on Build GRCh38
rs866790956 Chr.15: 82990301-82990301 on Build GRCh38
rs970548298 Chr.15: 82990501-82990501 on Build GRCh38
rs979779021 Chr.15: 82990671-82990671 on Build GRCh38
rs1026630684 Chr.15: 82990324-82990324 on Build GRCh38
rs1018319386 Chr.15: 82990535-82990535 on Build GRCh38
rs927039740 Chr.15: 82990451-82990451 on Build GRCh38
rs947630977 Chr.15: 82990635-82990635 on Build GRCh38
rs965674211 Chr.15: 82990229-82990229 on Build GRCh38
rs576044610 Chr.15: 82990506-82990506 on Build GRCh38
rs759578867 Chr.15: 82990543-82990544 on Build GRCh38
rs988661932 Chr.15: 82990632-82990632 on Build GRCh38
rs938668175 Chr.15: 82990592-82990592 on Build GRCh38
rs980130749 Chr.15: 82990531-82990531 on Build GRCh38
rs1006903541 Chr.15: 82990505-82990505 on Build GRCh38
rs968575425 Chr.15: 82990601-82990601 on Build GRCh38
rs1007329040 Chr.15: 82990669-82990669 on Build GRCh38
rs544961770 Chr.15: 82990573-82990573 on Build GRCh38
rs985021540 Chr.15: 82990352-82990352 on Build GRCh38
rs1033285518 Chr.15: 82990615-82990615 on Build GRCh38
rs1044092730 Chr.15: 82990438-82990438 on Build GRCh38
rs1007278149 Chr.15: 82990200-82990201 on Build GRCh38
rs36034211 Chr.15: 82990544-82990545 on Build GRCh38
rs747046285 Chr.15: 82990654-82990654 on Build GRCh38
rs955936372 Chr.15: 82990620-82990620 on Build GRCh38
rs941537145 Chr.15: 82990458-82990458 on Build GRCh38
rs527779061 Chr.15: 82990611-82990611 on Build GRCh38
rs994444825 Chr.15: 82990488-82990488 on Build GRCh38
rs950617668 Chr.15: 82990343-82990343 on Build GRCh38
rs574710727 Chr.15: 82990217-82990217 on Build GRCh38
rs186257867 Chr.15: 82990663-82990663 on Build GRCh38
rs1051364148 Chr.15: 82990495-82990495 on Build GRCh38
rs563571481 Chr.15: 82990500-82990500 on Build GRCh38
rs1017300056 Chr.15: 82990201-82990205 on Build GRCh38
rs879132671 Chr.15: 82990544-82990544 on Build GRCh38

View Full Product Details