Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 100400221-100400492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERS3
Entrez Gene ID
204219
Gene Name
ceramide synthase 3
Gene Aliases
ARCI9, LASS3
UniGene
Hs.662371
Gene Chromosome Location
Chr.15: 100400395-100544982 on Build GRCh38
Transcripts
NM_001290341.2 NM_001290342.2 NM_001290343.1 NM_178842.4 XM_011521355.2 XM_011521357.2 XM_011521358.1 XM_017022002.1 XM_017022003.1 XM_017022004.1 NR_120374.1

Gene Symbol
CERS3-AS1
Entrez Gene ID
102723320
Gene Name
CERS3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.416099
Gene Chromosome Location
Chr.15: 100372939-100437914 on Build GRCh38
Transcripts
NM_001290341.2 NM_001290342.2 NM_001290343.1 NM_178842.4 XM_011521355.2 XM_011521357.2 XM_011521358.1 XM_017022002.1 XM_017022003.1 XM_017022004.1 NR_120374.1

SNP Detail

SNP ID Chromosome Location
rs888361789 Chr.15: 100400418-100400418 on Build GRCh38
rs1001223132 Chr.15: 100400316-100400316 on Build GRCh38
rs575350283 Chr.15: 100400283-100400283 on Build GRCh38
rs932481959 Chr.15: 100400372-100400372 on Build GRCh38
rs748115132 Chr.15: 100400264-100400264 on Build GRCh38
rs771126717 Chr.15: 100400351-100400351 on Build GRCh38
rs941044310 Chr.15: 100400422-100400422 on Build GRCh38
rs773212090 Chr.15: 100400300-100400300 on Build GRCh38
rs967308683 Chr.15: 100400434-100400434 on Build GRCh38
rs995524044 Chr.15: 100400285-100400285 on Build GRCh38
rs959787531 Chr.15: 100400349-100400349 on Build GRCh38
rs772109091 Chr.15: 100400284-100400284 on Build GRCh38
rs1050135384 Chr.15: 100400384-100400384 on Build GRCh38
rs747240393 Chr.15: 100400317-100400317 on Build GRCh38
rs139941505 Chr.15: 100400362-100400362 on Build GRCh38
rs1048039020 Chr.15: 100400296-100400296 on Build GRCh38
rs896796017 Chr.15: 100400450-100400450 on Build GRCh38
rs1014031798 Chr.15: 100400453-100400453 on Build GRCh38

View Full Product Details